Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111298246-111298740 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CUX2
Entrez Gene ID
23316
Gene Name
cut like homeobox 2
Gene Aliases
CDP2, CUTL2
UniGene
Hs.124953
Gene Chromosome Location
Chr.12: 111034024-111350556 on Build GRCh38
Transcripts
NM_015267.3 XM_011538061.2 XM_011538063.2 XM_011538069.2 XM_011538070.2 XM_017019080.1 XM_017019081.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6494113 c.705-2A>C p.? Chr.12: 111298539-111298539 on Build GRCh38
243923 c.745G>A p.A249T Chr.12: 111298581-111298581 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs922960600 Chr.12: 111298302-111298302 on Build GRCh38
rs1037908766 Chr.12: 111298679-111298679 on Build GRCh38
rs866178813 Chr.12: 111298550-111298550 on Build GRCh38
rs763177862 Chr.12: 111298603-111298603 on Build GRCh38
rs765115719 Chr.12: 111298500-111298500 on Build GRCh38
rs768316702 Chr.12: 111298641-111298641 on Build GRCh38
rs866977401 Chr.12: 111298587-111298587 on Build GRCh38
rs370133313 Chr.12: 111298586-111298586 on Build GRCh38
rs756419901 Chr.12: 111298521-111298521 on Build GRCh38
rs373751946 Chr.12: 111298627-111298627 on Build GRCh38
rs925186848 Chr.12: 111298711-111298711 on Build GRCh38
rs778094108 Chr.12: 111298522-111298522 on Build GRCh38
rs758175938 Chr.12: 111298504-111298504 on Build GRCh38
rs1038341798 Chr.12: 111298385-111298385 on Build GRCh38
rs1034392100 Chr.12: 111298421-111298421 on Build GRCh38
rs1032664641 Chr.12: 111298490-111298490 on Build GRCh38
rs780397353 Chr.12: 111298615-111298615 on Build GRCh38
rs1022950008 Chr.12: 111298695-111298708 on Build GRCh38
rs1014358890 Chr.12: 111298443-111298443 on Build GRCh38
rs571437106 Chr.12: 111298489-111298489 on Build GRCh38
rs886622212 Chr.12: 111298367-111298367 on Build GRCh38
rs1015543429 Chr.12: 111298532-111298532 on Build GRCh38
rs913308342 Chr.12: 111298414-111298414 on Build GRCh38
rs138009341 Chr.12: 111298600-111298600 on Build GRCh38
rs879141608 Chr.12: 111298479-111298479 on Build GRCh38
rs1024787687 Chr.12: 111298467-111298467 on Build GRCh38
rs899444727 Chr.12: 111298397-111298397 on Build GRCh38
rs150355586 Chr.12: 111298514-111298514 on Build GRCh38
rs567332099 Chr.12: 111298386-111298386 on Build GRCh38
rs1045715153 Chr.12: 111298485-111298485 on Build GRCh38
rs538837671 Chr.12: 111298501-111298501 on Build GRCh38
rs373503790 Chr.12: 111298592-111298592 on Build GRCh38
rs375736198 Chr.12: 111298556-111298556 on Build GRCh38
rs527267833 Chr.12: 111298325-111298325 on Build GRCh38
rs376337237 Chr.12: 111298440-111298440 on Build GRCh38
rs933073774 Chr.12: 111298357-111298357 on Build GRCh38
rs11065854 Chr.12: 111298274-111298274 on Build GRCh38
rs921683068 Chr.12: 111298353-111298353 on Build GRCh38
rs953250587 Chr.12: 111298354-111298354 on Build GRCh38
rs1001934477 Chr.12: 111298398-111298398 on Build GRCh38
rs779340550 Chr.12: 111298584-111298584 on Build GRCh38
rs750342664 Chr.12: 111298503-111298503 on Build GRCh38
rs895867482 Chr.12: 111298436-111298436 on Build GRCh38
rs371327937 Chr.12: 111298506-111298506 on Build GRCh38
rs757331678 Chr.12: 111298554-111298554 on Build GRCh38
rs907232248 Chr.12: 111298486-111298486 on Build GRCh38
rs1032433105 Chr.12: 111298496-111298496 on Build GRCh38
rs531337871 Chr.12: 111298413-111298413 on Build GRCh38
rs4766459 Chr.12: 111298470-111298470 on Build GRCh38
rs987200476 Chr.12: 111298616-111298616 on Build GRCh38
rs749413126 Chr.12: 111298541-111298541 on Build GRCh38
rs112918730 Chr.12: 111298613-111298613 on Build GRCh38
rs12313589 Chr.12: 111298358-111298358 on Build GRCh38
rs974467697 Chr.12: 111298321-111298321 on Build GRCh38
rs751333910 Chr.12: 111298512-111298512 on Build GRCh38
rs977978451 Chr.12: 111298483-111298483 on Build GRCh38
rs762004141 Chr.12: 111298377-111298377 on Build GRCh38

View Full Product Details