Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111784891-111785389 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH2
Entrez Gene ID
217
Gene Name
aldehyde dehydrogenase 2 family (mitochondrial)
Gene Aliases
ALDH-E2, ALDHI, ALDM
UniGene
Hs.604551
Gene Chromosome Location
Chr.12: 111766887-111809985 on Build GRCh38
Transcripts
NM_000690.3 NM_001204889.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1947903 c.372C>T p.T124T Chr.12: 111785278-111785278 on Build GRCh38
3772257 c.440+8C>T p.? Chr.12: 111785354-111785354 on Build GRCh38
6331203 c.394G>T p.V132F Chr.12: 111785300-111785300 on Build GRCh38
5124417 c.414T>G p.D138E Chr.12: 111785320-111785320 on Build GRCh38
5458325 c.440+7G>A p.? Chr.12: 111785353-111785353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1033627886 Chr.12: 111785028-111785028 on Build GRCh38
rs371993659 Chr.12: 111785346-111785346 on Build GRCh38
rs1041935924 Chr.12: 111784924-111784924 on Build GRCh38
rs749478901 Chr.12: 111785364-111785364 on Build GRCh38
rs900004707 Chr.12: 111784983-111784983 on Build GRCh38
rs556864953 Chr.12: 111784928-111784928 on Build GRCh38
rs141373324 Chr.12: 111785326-111785326 on Build GRCh38
rs560781954 Chr.12: 111785137-111785137 on Build GRCh38
rs377343804 Chr.12: 111785238-111785238 on Build GRCh38
rs112230847 Chr.12: 111785136-111785137 on Build GRCh38
rs954905025 Chr.12: 111785129-111785129 on Build GRCh38
rs190633288 Chr.12: 111785261-111785261 on Build GRCh38
rs367691794 Chr.12: 111785357-111785357 on Build GRCh38
rs752335420 Chr.12: 111784967-111784967 on Build GRCh38
rs374555554 Chr.12: 111785237-111785237 on Build GRCh38
rs751931265 Chr.12: 111785345-111785345 on Build GRCh38
rs532463245 Chr.12: 111785188-111785188 on Build GRCh38
rs4648328 Chr.12: 111784984-111784984 on Build GRCh38
rs753520756 Chr.12: 111785337-111785337 on Build GRCh38
rs771960765 Chr.12: 111785312-111785312 on Build GRCh38
rs893893753 Chr.12: 111784962-111784962 on Build GRCh38
rs958585550 Chr.12: 111785154-111785154 on Build GRCh38
rs781322365 Chr.12: 111785245-111785245 on Build GRCh38
rs13306165 Chr.12: 111785352-111785352 on Build GRCh38
rs997054274 Chr.12: 111785286-111785286 on Build GRCh38
rs777614177 Chr.12: 111785147-111785147 on Build GRCh38
rs1014846978 Chr.12: 111785107-111785107 on Build GRCh38
rs1054278049 Chr.12: 111784948-111784949 on Build GRCh38
rs759512257 Chr.12: 111785301-111785301 on Build GRCh38
rs1026424714 Chr.12: 111785124-111785124 on Build GRCh38
rs753675078 Chr.12: 111785156-111785156 on Build GRCh38
rs760105168 Chr.12: 111785315-111785315 on Build GRCh38
rs945423353 Chr.12: 111785187-111785187 on Build GRCh38
rs536895009 Chr.12: 111785228-111785228 on Build GRCh38
rs949701064 Chr.12: 111785196-111785196 on Build GRCh38
rs375313806 Chr.12: 111785259-111785259 on Build GRCh38
rs751690737 Chr.12: 111785312-111785313 on Build GRCh38
rs3832868 Chr.12: 111785063-111785063 on Build GRCh38
rs768784052 Chr.12: 111784954-111784954 on Build GRCh38
rs758863490 Chr.12: 111785229-111785229 on Build GRCh38
rs774052763 Chr.12: 111785298-111785298 on Build GRCh38
rs1029761295 Chr.12: 111785068-111785068 on Build GRCh38
rs780616608 Chr.12: 111785232-111785232 on Build GRCh38
rs1042523900 Chr.12: 111785195-111785195 on Build GRCh38
rs116758112 Chr.12: 111785169-111785169 on Build GRCh38
rs148600666 Chr.12: 111785069-111785069 on Build GRCh38
rs201141937 Chr.12: 111785064-111785064 on Build GRCh38
rs766849182 Chr.12: 111785338-111785338 on Build GRCh38
rs1009373745 Chr.12: 111784981-111784981 on Build GRCh38
rs576228549 Chr.12: 111785146-111785146 on Build GRCh38
rs547685237 Chr.12: 111785199-111785199 on Build GRCh38
rs777877167 Chr.12: 111785358-111785358 on Build GRCh38
rs781556042 Chr.12: 111785353-111785353 on Build GRCh38
rs577898083 Chr.12: 111784966-111784966 on Build GRCh38
rs910844304 Chr.12: 111785138-111785138 on Build GRCh38
rs374472479 Chr.12: 111785355-111785355 on Build GRCh38
rs1041404387 Chr.12: 111785197-111785197 on Build GRCh38
rs1003447305 Chr.12: 111785003-111785003 on Build GRCh38
rs943418719 Chr.12: 111784947-111784947 on Build GRCh38
rs397804637 Chr.12: 111785064-111785064 on Build GRCh38
rs755001438 Chr.12: 111785314-111785314 on Build GRCh38
rs138443541 Chr.12: 111785296-111785296 on Build GRCh38
rs370855783 Chr.12: 111785251-111785251 on Build GRCh38
rs936215416 Chr.12: 111785250-111785250 on Build GRCh38
rs766248728 Chr.12: 111785062-111785062 on Build GRCh38
rs113539902 Chr.12: 111785157-111785157 on Build GRCh38
rs894799092 Chr.12: 111785039-111785039 on Build GRCh38
rs571679923 Chr.12: 111785075-111785075 on Build GRCh38
rs367911769 Chr.12: 111785285-111785285 on Build GRCh38
rs779179243 Chr.12: 111785226-111785226 on Build GRCh38
rs961169574 Chr.12: 111785076-111785076 on Build GRCh38
rs115350402 Chr.12: 111785093-111785093 on Build GRCh38

View Full Product Details