Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 111799828-111800336 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALDH2
Entrez Gene ID
217
Gene Name
aldehyde dehydrogenase 2 family (mitochondrial)
Gene Aliases
ALDH-E2, ALDHI, ALDM
UniGene
Hs.604551
Gene Chromosome Location
Chr.12: 111766887-111809985 on Build GRCh38
Transcripts
NM_000690.3 NM_001204889.1 NR_106819.1

Gene Symbol
MIR6761
Entrez Gene ID
102465456
Gene Name
microRNA 6761
Gene Aliases
hsa-mir-6761
UniGene
Gene Chromosome Location
Chr.12: 111799834-111799905 on Build GRCh38
Transcripts
NM_000690.3 NM_001204889.1 NR_106819.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6598240 c.1366G>A p.A456T Chr.12: 111800023-111800023 on Build GRCh38
7187652 c.1257G>A p.G419G Chr.12: 111799914-111799914 on Build GRCh38
6574939 c.1284G>T p.K428N Chr.12: 111799941-111799941 on Build GRCh38
5104737 c.1253T>C p.F418S Chr.12: 111799910-111799910 on Build GRCh38
3456130 c.1294G>A p.E432K Chr.12: 111799951-111799951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs189647488 Chr.12: 111800105-111800105 on Build GRCh38
rs774697819 Chr.12: 111799898-111799898 on Build GRCh38
rs745937439 Chr.12: 111800074-111800074 on Build GRCh38
rs771996854 Chr.12: 111800273-111800273 on Build GRCh38
rs747280625 Chr.12: 111800103-111800103 on Build GRCh38
rs201405479 Chr.12: 111800038-111800038 on Build GRCh38
rs775286182 Chr.12: 111800010-111800010 on Build GRCh38
rs947644216 Chr.12: 111800262-111800262 on Build GRCh38
rs373059460 Chr.12: 111799876-111799876 on Build GRCh38
rs999097746 Chr.12: 111799924-111799924 on Build GRCh38
rs753357413 Chr.12: 111799867-111799867 on Build GRCh38
rs764691327 Chr.12: 111800281-111800281 on Build GRCh38
rs1056245071 Chr.12: 111800292-111800292 on Build GRCh38
rs750085443 Chr.12: 111799945-111799945 on Build GRCh38
rs928280347 Chr.12: 111800190-111800190 on Build GRCh38
rs369822900 Chr.12: 111799868-111799868 on Build GRCh38
rs4646779 Chr.12: 111800177-111800177 on Build GRCh38
rs200408280 Chr.12: 111799887-111799887 on Build GRCh38
rs550693989 Chr.12: 111800224-111800224 on Build GRCh38
rs139552439 Chr.12: 111799883-111799883 on Build GRCh38
rs764713289 Chr.12: 111800027-111800027 on Build GRCh38
rs778449798 Chr.12: 111799965-111799965 on Build GRCh38
rs527914959 Chr.12: 111799882-111799882 on Build GRCh38
rs372806989 Chr.12: 111799892-111799892 on Build GRCh38
rs867446901 Chr.12: 111799984-111799984 on Build GRCh38
rs747287850 Chr.12: 111799880-111799880 on Build GRCh38
rs1045404081 Chr.12: 111799889-111799889 on Build GRCh38
rs542666146 Chr.12: 111800310-111800310 on Build GRCh38
rs146167264 Chr.12: 111800102-111800102 on Build GRCh38
rs889843446 Chr.12: 111799964-111799964 on Build GRCh38
rs558809393 Chr.12: 111800182-111800182 on Build GRCh38
rs199811756 Chr.12: 111799911-111799911 on Build GRCh38
rs764776351 Chr.12: 111799944-111799944 on Build GRCh38
rs896373176 Chr.12: 111800293-111800293 on Build GRCh38
rs183108437 Chr.12: 111799896-111799896 on Build GRCh38
rs371851241 Chr.12: 111800031-111800031 on Build GRCh38
rs756781003 Chr.12: 111799926-111799926 on Build GRCh38
rs368435076 Chr.12: 111799963-111799963 on Build GRCh38
rs376773752 Chr.12: 111799888-111799888 on Build GRCh38
rs999339893 Chr.12: 111800280-111800280 on Build GRCh38
rs1012586095 Chr.12: 111800301-111800301 on Build GRCh38
rs760416164 Chr.12: 111800014-111800014 on Build GRCh38
rs972388995 Chr.12: 111800056-111800056 on Build GRCh38
rs910583371 Chr.12: 111800247-111800247 on Build GRCh38
rs1039003577 Chr.12: 111800196-111800196 on Build GRCh38
rs764022056 Chr.12: 111799914-111799914 on Build GRCh38
rs761778133 Chr.12: 111800022-111800022 on Build GRCh38
rs536321330 Chr.12: 111800046-111800046 on Build GRCh38
rs547207199 Chr.12: 111800019-111800019 on Build GRCh38
rs772193708 Chr.12: 111799992-111799992 on Build GRCh38
rs550022818 Chr.12: 111799910-111799910 on Build GRCh38
rs772296175 Chr.12: 111800082-111800082 on Build GRCh38
rs776526118 Chr.12: 111800020-111800020 on Build GRCh38
rs746086528 Chr.12: 111799983-111799983 on Build GRCh38
rs757893333 Chr.12: 111800051-111800051 on Build GRCh38
rs753304028 Chr.12: 111799923-111799923 on Build GRCh38
rs866854424 Chr.12: 111800213-111800213 on Build GRCh38
rs753058436 Chr.12: 111800043-111800043 on Build GRCh38
rs763578676 Chr.12: 111799857-111799857 on Build GRCh38
rs771107373 Chr.12: 111799979-111799979 on Build GRCh38
rs560252444 Chr.12: 111800225-111800225 on Build GRCh38
rs530839095 Chr.12: 111800195-111800195 on Build GRCh38
rs1053410643 Chr.12: 111799927-111799927 on Build GRCh38
rs777079274 Chr.12: 111799912-111799912 on Build GRCh38
rs144598865 Chr.12: 111799948-111799948 on Build GRCh38
rs141556759 Chr.12: 111799980-111799980 on Build GRCh38
rs765981127 Chr.12: 111800041-111800041 on Build GRCh38
rs74429409 Chr.12: 111800244-111800244 on Build GRCh38
rs754387972 Chr.12: 111800050-111800050 on Build GRCh38
rs138505396 Chr.12: 111799959-111799959 on Build GRCh38
rs374377652 Chr.12: 111799946-111799946 on Build GRCh38
rs900507099 Chr.12: 111800220-111800220 on Build GRCh38
rs778990792 Chr.12: 111800052-111800052 on Build GRCh38
rs1022047610 Chr.12: 111799907-111799907 on Build GRCh38
rs755347814 Chr.12: 111799881-111799881 on Build GRCh38
rs893285841 Chr.12: 111799938-111799938 on Build GRCh38
rs915749712 Chr.12: 111800169-111800169 on Build GRCh38
rs749863724 Chr.12: 111799967-111799967 on Build GRCh38
rs1054444379 Chr.12: 111800068-111800068 on Build GRCh38
rs920756648 Chr.12: 111800089-111800089 on Build GRCh38

View Full Product Details