Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112161286-112161794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD4
Entrez Gene ID
283450
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Gene Aliases
C12orf51, POTAGE
UniGene
Hs.530943
Gene Chromosome Location
Chr.12: 112160188-112382092 on Build GRCh38
Transcripts
NM_001109662.3

SNP Detail

SNP ID Chromosome Location
rs1052476993 Chr.12: 112161716-112161716 on Build GRCh38
rs900888394 Chr.12: 112161668-112161668 on Build GRCh38
rs7968188 Chr.12: 112161589-112161589 on Build GRCh38
rs1007632844 Chr.12: 112161409-112161409 on Build GRCh38
rs555658241 Chr.12: 112161579-112161579 on Build GRCh38
rs35601228 Chr.12: 112161314-112161315 on Build GRCh38
rs769961233 Chr.12: 112161472-112161472 on Build GRCh38
rs371553368 Chr.12: 112161355-112161355 on Build GRCh38
rs953708469 Chr.12: 112161385-112161385 on Build GRCh38
rs7978730 Chr.12: 112161337-112161337 on Build GRCh38
rs568017657 Chr.12: 112161620-112161620 on Build GRCh38
rs1031183419 Chr.12: 112161552-112161552 on Build GRCh38
rs80096478 Chr.12: 112161461-112161461 on Build GRCh38
rs933568926 Chr.12: 112161672-112161672 on Build GRCh38
rs1032391014 Chr.12: 112161741-112161741 on Build GRCh38
rs570034884 Chr.12: 112161348-112161348 on Build GRCh38
rs957993897 Chr.12: 112161471-112161471 on Build GRCh38
rs183487457 Chr.12: 112161338-112161338 on Build GRCh38
rs12462 Chr.12: 112161652-112161652 on Build GRCh38
rs989713771 Chr.12: 112161566-112161566 on Build GRCh38
rs762977364 Chr.12: 112161557-112161557 on Build GRCh38
rs1030498765 Chr.12: 112161379-112161379 on Build GRCh38
rs897687149 Chr.12: 112161683-112161683 on Build GRCh38
rs888440707 Chr.12: 112161740-112161740 on Build GRCh38
rs944510001 Chr.12: 112161653-112161653 on Build GRCh38
rs970628368 Chr.12: 112161481-112161481 on Build GRCh38
rs79726326 Chr.12: 112161479-112161479 on Build GRCh38
rs774852064 Chr.12: 112161625-112161625 on Build GRCh38
rs535129710 Chr.12: 112161624-112161624 on Build GRCh38
rs760768742 Chr.12: 112161734-112161734 on Build GRCh38
rs1036659383 Chr.12: 112161664-112161664 on Build GRCh38
rs956969021 Chr.12: 112161554-112161554 on Build GRCh38
rs552343506 Chr.12: 112161439-112161439 on Build GRCh38
rs1036216353 Chr.12: 112161658-112161658 on Build GRCh38
rs776631771 Chr.12: 112161394-112161394 on Build GRCh38
rs982252212 Chr.12: 112161549-112161549 on Build GRCh38
rs144286398 Chr.12: 112161721-112161721 on Build GRCh38
rs531144158 Chr.12: 112161354-112161354 on Build GRCh38
rs1036544640 Chr.12: 112161314-112161314 on Build GRCh38
rs931701451 Chr.12: 112161605-112161605 on Build GRCh38
rs898091904 Chr.12: 112161336-112161336 on Build GRCh38
rs1012740425 Chr.12: 112161404-112161404 on Build GRCh38
rs552804702 Chr.12: 112161594-112161594 on Build GRCh38
rs7968051 Chr.12: 112161445-112161445 on Build GRCh38
rs930536253 Chr.12: 112161713-112161713 on Build GRCh38

View Full Product Details