Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 112161695-112162206 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD4
Entrez Gene ID
283450
Gene Name
HECT domain E3 ubiquitin protein ligase 4
Gene Aliases
C12orf51, POTAGE
UniGene
Hs.530943
Gene Chromosome Location
Chr.12: 112160188-112382092 on Build GRCh38
Transcripts
NM_001109662.3

SNP Detail

SNP ID Chromosome Location
rs948022319 Chr.12: 112162001-112162001 on Build GRCh38
rs530076240 Chr.12: 112161979-112161979 on Build GRCh38
rs919082349 Chr.12: 112161959-112161959 on Build GRCh38
rs559607796 Chr.12: 112161875-112161875 on Build GRCh38
rs377559858 Chr.12: 112161862-112161862 on Build GRCh38
rs35555620 Chr.12: 112161975-112161976 on Build GRCh38
rs894402463 Chr.12: 112161998-112161998 on Build GRCh38
rs933677747 Chr.12: 112162056-112162056 on Build GRCh38
rs1052001016 Chr.12: 112162108-112162108 on Build GRCh38
rs906632181 Chr.12: 112161808-112161808 on Build GRCh38
rs889372532 Chr.12: 112162120-112162120 on Build GRCh38
rs1045092291 Chr.12: 112162031-112162031 on Build GRCh38
rs922260256 Chr.12: 112162033-112162033 on Build GRCh38
rs1032391014 Chr.12: 112161741-112161741 on Build GRCh38
rs970745902 Chr.12: 112161835-112161835 on Build GRCh38
rs893896570 Chr.12: 112161788-112161788 on Build GRCh38
rs574495673 Chr.12: 112161790-112161790 on Build GRCh38
rs572954626 Chr.12: 112161863-112161863 on Build GRCh38
rs758364554 Chr.12: 112161982-112161982 on Build GRCh38
rs971834626 Chr.12: 112161946-112161946 on Build GRCh38
rs956899584 Chr.12: 112161840-112161840 on Build GRCh38
rs552456285 Chr.12: 112162183-112162183 on Build GRCh38
rs188884732 Chr.12: 112161871-112161871 on Build GRCh38
rs375866854 Chr.12: 112161812-112161812 on Build GRCh38
rs1024103832 Chr.12: 112161807-112161807 on Build GRCh38
rs888440707 Chr.12: 112161740-112161740 on Build GRCh38
rs541806228 Chr.12: 112162036-112162036 on Build GRCh38
rs756901747 Chr.12: 112161903-112161903 on Build GRCh38
rs760768742 Chr.12: 112161734-112161734 on Build GRCh38
rs985825924 Chr.12: 112161892-112161892 on Build GRCh38
rs1003644430 Chr.12: 112161828-112161828 on Build GRCh38
rs964395301 Chr.12: 112161970-112161970 on Build GRCh38
rs368374249 Chr.12: 112162135-112162136 on Build GRCh38
rs546157835 Chr.12: 112161824-112161824 on Build GRCh38
rs944419078 Chr.12: 112161932-112161932 on Build GRCh38
rs531108735 Chr.12: 112162070-112162070 on Build GRCh38
rs144286398 Chr.12: 112161721-112161721 on Build GRCh38
rs774474068 Chr.12: 112162005-112162005 on Build GRCh38
rs3830633 Chr.12: 112162132-112162133 on Build GRCh38
rs1031079380 Chr.12: 112162167-112162167 on Build GRCh38
rs1034031776 Chr.12: 112161886-112161886 on Build GRCh38
rs749899185 Chr.12: 112162014-112162014 on Build GRCh38
rs767864934 Chr.12: 112161831-112161831 on Build GRCh38
rs193152225 Chr.12: 112162052-112162052 on Build GRCh38
rs757856913 Chr.12: 112162029-112162029 on Build GRCh38
rs1003570815 Chr.12: 112162164-112162164 on Build GRCh38
rs764829036 Chr.12: 112161926-112161926 on Build GRCh38

View Full Product Details