Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113154292-113154791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP73
Entrez Gene ID
387885
Gene Name
cilia and flagella associated protein 73
Gene Aliases
CCDC42B, MIA2
UniGene
Hs.486856
Gene Chromosome Location
Chr.12: 113149162-113159276 on Build GRCh38
Transcripts
NM_001144872.1 XM_011538327.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6194675 c.501C>G p.F167L Chr.12: 113154446-113154446 on Build GRCh38
1358949 c.512C>T p.A171V Chr.12: 113154457-113154457 on Build GRCh38
6394204 c.615G>T p.E205D Chr.12: 113154560-113154560 on Build GRCh38
6429250 c.506G>A p.G169D Chr.12: 113154451-113154451 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs756598920 Chr.12: 113154719-113154720 on Build GRCh38
rs928365491 Chr.12: 113154686-113154686 on Build GRCh38
rs754566301 Chr.12: 113154643-113154643 on Build GRCh38
rs77610415 Chr.12: 113154748-113154748 on Build GRCh38
rs569409543 Chr.12: 113154649-113154649 on Build GRCh38
rs906958932 Chr.12: 113154443-113154443 on Build GRCh38
rs956291749 Chr.12: 113154599-113154599 on Build GRCh38
rs978062265 Chr.12: 113154609-113154609 on Build GRCh38
rs889657799 Chr.12: 113154349-113154349 on Build GRCh38
rs897052500 Chr.12: 113154456-113154456 on Build GRCh38
rs758336329 Chr.12: 113154510-113154510 on Build GRCh38
rs764054583 Chr.12: 113154436-113154436 on Build GRCh38
rs556701233 Chr.12: 113154725-113154725 on Build GRCh38
rs900426425 Chr.12: 113154768-113154768 on Build GRCh38
rs886339334 Chr.12: 113154529-113154529 on Build GRCh38
rs189148376 Chr.12: 113154744-113154744 on Build GRCh38
rs1049555031 Chr.12: 113154328-113154328 on Build GRCh38
rs948261094 Chr.12: 113154425-113154425 on Build GRCh38
rs900307644 Chr.12: 113154582-113154582 on Build GRCh38
rs760878157 Chr.12: 113154400-113154400 on Build GRCh38
rs760145852 Chr.12: 113154383-113154383 on Build GRCh38
rs772653103 Chr.12: 113154388-113154388 on Build GRCh38
rs547786243 Chr.12: 113154647-113154647 on Build GRCh38
rs373830521 Chr.12: 113154470-113154470 on Build GRCh38
rs916764167 Chr.12: 113154351-113154351 on Build GRCh38
rs1047376326 Chr.12: 113154731-113154731 on Build GRCh38
rs774891726 Chr.12: 113154625-113154625 on Build GRCh38
rs914536821 Chr.12: 113154602-113154602 on Build GRCh38
rs970662836 Chr.12: 113154575-113154575 on Build GRCh38
rs1044144882 Chr.12: 113154440-113154440 on Build GRCh38
rs888203830 Chr.12: 113154545-113154545 on Build GRCh38
rs888152764 Chr.12: 113154750-113154750 on Build GRCh38
rs536788003 Chr.12: 113154659-113154659 on Build GRCh38
rs747086199 Chr.12: 113154660-113154660 on Build GRCh38
rs989227927 Chr.12: 113154601-113154601 on Build GRCh38
rs775718623 Chr.12: 113154399-113154399 on Build GRCh38
rs994258229 Chr.12: 113154467-113154467 on Build GRCh38
rs948163440 Chr.12: 113154654-113154654 on Build GRCh38
rs368029965 Chr.12: 113154720-113154720 on Build GRCh38
rs1036375311 Chr.12: 113154595-113154595 on Build GRCh38
rs535805558 Chr.12: 113154552-113154552 on Build GRCh38
rs1023386770 Chr.12: 113154561-113154561 on Build GRCh38
rs761897292 Chr.12: 113154451-113154451 on Build GRCh38
rs985545459 Chr.12: 113154626-113154626 on Build GRCh38
rs200344876 Chr.12: 113154501-113154502 on Build GRCh38
rs532416340 Chr.12: 113154450-113154450 on Build GRCh38
rs764039546 Chr.12: 113154428-113154428 on Build GRCh38
rs1016193020 Chr.12: 113154574-113154574 on Build GRCh38
rs35881201 Chr.12: 113154615-113154616 on Build GRCh38
rs554471177 Chr.12: 113154761-113154761 on Build GRCh38
rs981618701 Chr.12: 113154585-113154585 on Build GRCh38
rs751437694 Chr.12: 113154604-113154604 on Build GRCh38

View Full Product Details