Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 113191791-113192292 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RITA1
Entrez Gene ID
84934
Gene Name
RBPJ interacting and tubulin associated 1
Gene Aliases
C12orf52, RITA
UniGene
Hs.524762
Gene Chromosome Location
Chr.12: 113185526-113192368 on Build GRCh38
Transcripts
NM_001286215.1 NM_032848.2 XM_005253972.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
259716 c.804G>T p.W268C Chr.12: 113191811-113191811 on Build GRCh38
1677365 c.797C>T p.P266L Chr.12: 113191804-113191804 on Build GRCh38
5051356 c.796C>T p.P266S Chr.12: 113191803-113191803 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs910987659 Chr.12: 113192107-113192107 on Build GRCh38
rs571942580 Chr.12: 113191820-113191820 on Build GRCh38
rs1000200284 Chr.12: 113191908-113191908 on Build GRCh38
rs1051009072 Chr.12: 113192199-113192199 on Build GRCh38
rs1000000451 Chr.12: 113192184-113192184 on Build GRCh38
rs765924256 Chr.12: 113191863-113191863 on Build GRCh38
rs901070662 Chr.12: 113192141-113192141 on Build GRCh38
rs775398585 Chr.12: 113191840-113191840 on Build GRCh38
rs753127642 Chr.12: 113191865-113191865 on Build GRCh38
rs958671974 Chr.12: 113191945-113191945 on Build GRCh38
rs935649968 Chr.12: 113192106-113192106 on Build GRCh38
rs1014840785 Chr.12: 113191900-113191900 on Build GRCh38
rs749409309 Chr.12: 113191814-113191814 on Build GRCh38
rs996372488 Chr.12: 113192164-113192164 on Build GRCh38
rs776514177 Chr.12: 113191849-113191849 on Build GRCh38
rs531658790 Chr.12: 113192186-113192186 on Build GRCh38
rs962350832 Chr.12: 113192009-113192009 on Build GRCh38
rs972635026 Chr.12: 113192048-113192048 on Build GRCh38
rs942317722 Chr.12: 113192113-113192116 on Build GRCh38
rs528139015 Chr.12: 113191961-113191961 on Build GRCh38
rs772030531 Chr.12: 113192008-113192008 on Build GRCh38
rs1043845 Chr.12: 113192144-113192144 on Build GRCh38
rs760367270 Chr.12: 113191842-113191842 on Build GRCh38
rs113499502 Chr.12: 113192155-113192155 on Build GRCh38
rs149502906 Chr.12: 113191891-113191891 on Build GRCh38
rs1041411424 Chr.12: 113192092-113192092 on Build GRCh38
rs969207419 Chr.12: 113191901-113191901 on Build GRCh38
rs962111632 Chr.12: 113191902-113191902 on Build GRCh38
rs924307176 Chr.12: 113192096-113192096 on Build GRCh38
rs758115487 Chr.12: 113191857-113191857 on Build GRCh38
rs917473568 Chr.12: 113191988-113191988 on Build GRCh38
rs904333482 Chr.12: 113192158-113192158 on Build GRCh38
rs920656427 Chr.12: 113191966-113191966 on Build GRCh38
rs549872050 Chr.12: 113192220-113192220 on Build GRCh38
rs199873710 Chr.12: 113191843-113191843 on Build GRCh38
rs11832314 Chr.12: 113191850-113191850 on Build GRCh38
rs1020143692 Chr.12: 113191890-113191890 on Build GRCh38
rs758608716 Chr.12: 113191833-113191834 on Build GRCh38
rs958592626 Chr.12: 113192014-113192014 on Build GRCh38
rs560967904 Chr.12: 113192175-113192175 on Build GRCh38
rs920811653 Chr.12: 113192053-113192054 on Build GRCh38
rs576235202 Chr.12: 113191851-113191851 on Build GRCh38
rs1032753606 Chr.12: 113191812-113191812 on Build GRCh38
rs949908714 Chr.12: 113192091-113192091 on Build GRCh38
rs772177154 Chr.12: 113191839-113191839 on Build GRCh38
rs917189271 Chr.12: 113192055-113192055 on Build GRCh38
rs144042449 Chr.12: 113191958-113191958 on Build GRCh38
rs16942608 Chr.12: 113191852-113191852 on Build GRCh38
rs750699779 Chr.12: 113191830-113191830 on Build GRCh38
rs578225282 Chr.12: 113191950-113191950 on Build GRCh38
rs1040737909 Chr.12: 113192129-113192129 on Build GRCh38
rs1034406054 Chr.12: 113191932-113191932 on Build GRCh38
rs560877884 Chr.12: 113192006-113192006 on Build GRCh38
rs73208734 Chr.12: 113191832-113191832 on Build GRCh38
rs991701032 Chr.12: 113192015-113192015 on Build GRCh38

View Full Product Details