Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 132853295-132853786 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHFR
Entrez Gene ID
55743
Gene Name
checkpoint with forkhead and ring finger domains, E3 ubiquitin protein ligase
Gene Aliases
RNF116, RNF196
UniGene
Hs.720197
Gene Chromosome Location
Chr.12: 132840352-132887618 on Build GRCh38
Transcripts
NM_001161344.1 NM_001161345.1 NM_001161346.1 NM_001161347.1 NM_018223.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3688108 c.1389G>T p.T463T Chr.12: 132853450-132853450 on Build GRCh38
6359958 c.1339C>T p.P447S Chr.12: 132853500-132853500 on Build GRCh38
6628323 c.1392C>T p.S464S Chr.12: 132853447-132853447 on Build GRCh38
6414459 c.1273A>T p.S425C Chr.12: 132853443-132853443 on Build GRCh38
4645839 c.1286G>A p.R429Q Chr.12: 132853553-132853553 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs965810108 Chr.12: 132853399-132853399 on Build GRCh38
rs865808693 Chr.12: 132853384-132853384 on Build GRCh38
rs759439729 Chr.12: 132853600-132853600 on Build GRCh38
rs138207644 Chr.12: 132853460-132853460 on Build GRCh38
rs146337818 Chr.12: 132853751-132853751 on Build GRCh38
rs374006779 Chr.12: 132853492-132853492 on Build GRCh38
rs748419509 Chr.12: 132853567-132853567 on Build GRCh38
rs117363206 Chr.12: 132853358-132853358 on Build GRCh38
rs778169456 Chr.12: 132853439-132853439 on Build GRCh38
rs1054935485 Chr.12: 132853647-132853647 on Build GRCh38
rs775426850 Chr.12: 132853608-132853608 on Build GRCh38
rs143653967 Chr.12: 132853672-132853672 on Build GRCh38
rs1019707998 Chr.12: 132853730-132853730 on Build GRCh38
rs374229656 Chr.12: 132853526-132853526 on Build GRCh38
rs367968934 Chr.12: 132853493-132853493 on Build GRCh38
rs150627547 Chr.12: 132853560-132853560 on Build GRCh38
rs746360566 Chr.12: 132853553-132853553 on Build GRCh38
rs546118391 Chr.12: 132853746-132853746 on Build GRCh38
rs902210721 Chr.12: 132853679-132853679 on Build GRCh38
rs574255301 Chr.12: 132853602-132853602 on Build GRCh38
rs563852229 Chr.12: 132853522-132853522 on Build GRCh38
rs763278073 Chr.12: 132853418-132853418 on Build GRCh38
rs117696161 Chr.12: 132853706-132853706 on Build GRCh38
rs536396076 Chr.12: 132853435-132853435 on Build GRCh38
rs140026593 Chr.12: 132853619-132853619 on Build GRCh38
rs752473859 Chr.12: 132853638-132853638 on Build GRCh38
rs902265625 Chr.12: 132853580-132853580 on Build GRCh38
rs770621972 Chr.12: 132853577-132853577 on Build GRCh38
rs765209279 Chr.12: 132853607-132853607 on Build GRCh38
rs975768269 Chr.12: 132853417-132853417 on Build GRCh38
rs775767314 Chr.12: 132853455-132853455 on Build GRCh38
rs148176220 Chr.12: 132853323-132853323 on Build GRCh38
rs993756830 Chr.12: 132853349-132853349 on Build GRCh38
rs191849035 Chr.12: 132853315-132853315 on Build GRCh38
rs141835819 Chr.12: 132853446-132853446 on Build GRCh38
rs768814594 Chr.12: 132853563-132853563 on Build GRCh38
rs1041545091 Chr.12: 132853486-132853486 on Build GRCh38
rs1056433107 Chr.12: 132853721-132853721 on Build GRCh38
rs1048859786 Chr.12: 132853350-132853350 on Build GRCh38
rs749373830 Chr.12: 132853561-132853561 on Build GRCh38
rs927350519 Chr.12: 132853639-132853639 on Build GRCh38
rs781699438 Chr.12: 132853544-132853544 on Build GRCh38
rs924051400 Chr.12: 132853488-132853488 on Build GRCh38
rs1020597739 Chr.12: 132853569-132853569 on Build GRCh38
rs972717972 Chr.12: 132853626-132853626 on Build GRCh38
rs918549955 Chr.12: 132853627-132853627 on Build GRCh38
rs868539292 Chr.12: 132853340-132853340 on Build GRCh38
rs764723102 Chr.12: 132853510-132853510 on Build GRCh38
rs1013270057 Chr.12: 132853565-132853565 on Build GRCh38
rs747380214 Chr.12: 132853442-132853442 on Build GRCh38
rs944033912 Chr.12: 132853317-132853317 on Build GRCh38
rs375812260 Chr.12: 132853421-132853421 on Build GRCh38
rs762328534 Chr.12: 132853611-132853611 on Build GRCh38
rs770024875 Chr.12: 132853403-132853403 on Build GRCh38
rs372270460 Chr.12: 132853566-132853566 on Build GRCh38
rs191413650 Chr.12: 132853378-132853378 on Build GRCh38
rs377661913 Chr.12: 132853422-132853422 on Build GRCh38
rs777240371 Chr.12: 132853525-132853525 on Build GRCh38
rs575456726 Chr.12: 132853727-132853727 on Build GRCh38
rs767325947 Chr.12: 132853419-132853419 on Build GRCh38
rs139904993 Chr.12: 132853762-132853762 on Build GRCh38
rs1013238955 Chr.12: 132853375-132853375 on Build GRCh38
rs757496767 Chr.12: 132853541-132853541 on Build GRCh38
rs887701014 Chr.12: 132853352-132853352 on Build GRCh38
rs368755833 Chr.12: 132853558-132853558 on Build GRCh38
rs1049043829 Chr.12: 132853735-132853736 on Build GRCh38
rs183988735 Chr.12: 132853321-132853321 on Build GRCh38
rs777595033 Chr.12: 132853373-132853373 on Build GRCh38
rs758130662 Chr.12: 132853646-132853646 on Build GRCh38
rs768798394 Chr.12: 132853482-132853482 on Build GRCh38
rs769933100 Chr.12: 132853451-132853451 on Build GRCh38
rs1019843304 Chr.12: 132853601-132853601 on Build GRCh38
rs1032115826 Chr.12: 132853704-132853704 on Build GRCh38
rs965803102 Chr.12: 132853618-132853618 on Build GRCh38
rs766543527 Chr.12: 132853396-132853396 on Build GRCh38
rs375336559 Chr.12: 132853595-132853595 on Build GRCh38
rs746380972 Chr.12: 132853450-132853450 on Build GRCh38
rs867070196 Chr.12: 132853631-132853631 on Build GRCh38
rs534389499 Chr.12: 132853594-132853594 on Build GRCh38
rs896411382 Chr.12: 132853651-132853651 on Build GRCh38
rs371541325 Chr.12: 132853523-132853523 on Build GRCh38
rs564562551 Chr.12: 132853357-132853357 on Build GRCh38
rs187999239 Chr.12: 132853371-132853371 on Build GRCh38
rs949244374 Chr.12: 132853659-132853659 on Build GRCh38
rs374668913 Chr.12: 132853423-132853423 on Build GRCh38
rs61412200 Chr.12: 132853596-132853596 on Build GRCh38
rs760789709 Chr.12: 132853372-132853372 on Build GRCh38
rs759529335 Chr.12: 132853498-132853498 on Build GRCh38
rs781511228 Chr.12: 132853447-132853447 on Build GRCh38
rs754422976 Chr.12: 132853424-132853424 on Build GRCh38
rs948088249 Chr.12: 132853728-132853728 on Build GRCh38
rs752333209 Chr.12: 132853436-132853436 on Build GRCh38
rs1000926941 Chr.12: 132853697-132853697 on Build GRCh38
rs967308971 Chr.12: 132853649-132853649 on Build GRCh38
rs775787800 Chr.12: 132853407-132853407 on Build GRCh38
rs753721064 Chr.12: 132853497-132853497 on Build GRCh38
rs939453079 Chr.12: 132853700-132853700 on Build GRCh38
rs937875968 Chr.12: 132853641-132853641 on Build GRCh38
rs776748296 Chr.12: 132853380-132853380 on Build GRCh38
rs1000659512 Chr.12: 132853587-132853587 on Build GRCh38
rs35167652 Chr.12: 132853725-132853726 on Build GRCh38
rs774658371 Chr.12: 132853491-132853491 on Build GRCh38
rs372547607 Chr.12: 132853316-132853316 on Build GRCh38

View Full Product Details