Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20141859-20142362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GJA3
Entrez Gene ID
2700
Gene Name
gap junction protein alpha 3
Gene Aliases
CTRCT14, CX46, CZP3
UniGene
Hs.130313
Gene Chromosome Location
Chr.13: 20138252-20161327 on Build GRCh38
Transcripts
NM_021954.3 XM_011535048.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5716239 c.928C>T p.Q310* Chr.13: 20142361-20142361 on Build GRCh38
5732526 c.1075T>C p.S359P Chr.13: 20142214-20142214 on Build GRCh38
1365738 c.1017G>A p.A339A Chr.13: 20142272-20142272 on Build GRCh38
4461040 c.1190C>T p.A397V Chr.13: 20142099-20142099 on Build GRCh38
6850177 c.1177G>A p.E393K Chr.13: 20142112-20142112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750903354 Chr.13: 20142111-20142111 on Build GRCh38
rs776453557 Chr.13: 20142235-20142235 on Build GRCh38
rs368180843 Chr.13: 20142203-20142203 on Build GRCh38
rs763561000 Chr.13: 20141896-20141896 on Build GRCh38
rs770822237 Chr.13: 20141979-20141979 on Build GRCh38
rs547364696 Chr.13: 20142266-20142266 on Build GRCh38
rs900736864 Chr.13: 20142076-20142076 on Build GRCh38
rs762438384 Chr.13: 20142255-20142255 on Build GRCh38
rs756171884 Chr.13: 20142156-20142156 on Build GRCh38
rs906390729 Chr.13: 20142114-20142114 on Build GRCh38
rs767950248 Chr.13: 20142087-20142087 on Build GRCh38
rs763735876 Chr.13: 20142065-20142065 on Build GRCh38
rs1027325918 Chr.13: 20142197-20142197 on Build GRCh38
rs745647356 Chr.13: 20142308-20142308 on Build GRCh38
rs1025143356 Chr.13: 20142182-20142182 on Build GRCh38
rs936228357 Chr.13: 20142082-20142082 on Build GRCh38
rs75630774 Chr.13: 20141934-20141934 on Build GRCh38
rs757015149 Chr.13: 20142306-20142306 on Build GRCh38
rs963846840 Chr.13: 20142230-20142230 on Build GRCh38
rs922529783 Chr.13: 20142010-20142010 on Build GRCh38
rs886050019 Chr.13: 20141880-20141880 on Build GRCh38
rs942579142 Chr.13: 20141999-20141999 on Build GRCh38
rs901224776 Chr.13: 20142214-20142214 on Build GRCh38
rs779251021 Chr.13: 20142206-20142206 on Build GRCh38
rs906802114 Chr.13: 20142192-20142192 on Build GRCh38
rs111570432 Chr.13: 20141881-20141881 on Build GRCh38
rs780112656 Chr.13: 20142167-20142167 on Build GRCh38
rs920227881 Chr.13: 20141975-20141975 on Build GRCh38
rs201955819 Chr.13: 20142055-20142055 on Build GRCh38
rs929587384 Chr.13: 20142025-20142025 on Build GRCh38
rs1010364645 Chr.13: 20142109-20142109 on Build GRCh38
rs929067280 Chr.13: 20141976-20141976 on Build GRCh38
rs1047204525 Chr.13: 20142049-20142049 on Build GRCh38
rs895291478 Chr.13: 20142115-20142115 on Build GRCh38
rs775049943 Chr.13: 20142254-20142254 on Build GRCh38
rs34389531 Chr.13: 20142129-20142130 on Build GRCh38
rs756709065 Chr.13: 20142275-20142275 on Build GRCh38
rs763785204 Chr.13: 20142263-20142263 on Build GRCh38
rs561316104 Chr.13: 20142009-20142009 on Build GRCh38
rs11617415 Chr.13: 20142272-20142272 on Build GRCh38
rs755073578 Chr.13: 20142284-20142284 on Build GRCh38
rs989998887 Chr.13: 20142224-20142224 on Build GRCh38
rs989325196 Chr.13: 20141958-20141958 on Build GRCh38
rs769829236 Chr.13: 20142331-20142331 on Build GRCh38
rs891750293 Chr.13: 20142106-20142106 on Build GRCh38
rs950387904 Chr.13: 20142201-20142201 on Build GRCh38
rs779308349 Chr.13: 20142289-20142289 on Build GRCh38
rs746840779 Chr.13: 20142226-20142226 on Build GRCh38
rs768118379 Chr.13: 20142341-20142341 on Build GRCh38
rs377190812 Chr.13: 20142340-20142340 on Build GRCh38
rs973109214 Chr.13: 20141945-20141945 on Build GRCh38
rs958396385 Chr.13: 20141937-20141937 on Build GRCh38
rs970679339 Chr.13: 20141886-20141886 on Build GRCh38
rs1024387568 Chr.13: 20142110-20142110 on Build GRCh38
rs776596399 Chr.13: 20141986-20141986 on Build GRCh38
rs973696142 Chr.13: 20142190-20142190 on Build GRCh38
rs746787477 Chr.13: 20141947-20141947 on Build GRCh38
rs757596277 Chr.13: 20142086-20142086 on Build GRCh38
rs200588691 Chr.13: 20142290-20142290 on Build GRCh38
rs141882615 Chr.13: 20142178-20142178 on Build GRCh38
rs770718484 Chr.13: 20142229-20142229 on Build GRCh38
rs764752482 Chr.13: 20142045-20142045 on Build GRCh38
rs911069225 Chr.13: 20141982-20141982 on Build GRCh38
rs752918056 Chr.13: 20142294-20142294 on Build GRCh38
rs769194838 Chr.13: 20142252-20142252 on Build GRCh38
rs958128474 Chr.13: 20142211-20142211 on Build GRCh38
rs780971190 Chr.13: 20142307-20142307 on Build GRCh38
rs774018241 Chr.13: 20142342-20142342 on Build GRCh38
rs758996859 Chr.13: 20142018-20142018 on Build GRCh38
rs766975987 Chr.13: 20142276-20142276 on Build GRCh38
rs745758244 Chr.13: 20142244-20142244 on Build GRCh38
rs565869270 Chr.13: 20142271-20142271 on Build GRCh38
rs974958945 Chr.13: 20142238-20142238 on Build GRCh38
rs543711227 Chr.13: 20142056-20142056 on Build GRCh38
rs754015946 Chr.13: 20142280-20142280 on Build GRCh38
rs886050020 Chr.13: 20142207-20142207 on Build GRCh38
rs1033585708 Chr.13: 20141933-20141933 on Build GRCh38
rs751848890 Chr.13: 20142069-20142069 on Build GRCh38

View Full Product Details