Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 20413332-20413863 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYL1
Entrez Gene ID
51084
Gene Name
crystallin lambda 1
Gene Aliases
GDH, HEL30, lambda-CRY
UniGene
Hs.370703
Gene Chromosome Location
Chr.13: 20403667-20525873 on Build GRCh38
Transcripts
NM_015974.2 XM_005266415.3 XM_017020624.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
695835 c.661G>T p.D221Y Chr.13: 20413360-20413360 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1037855428 Chr.13: 20413455-20413455 on Build GRCh38
rs926883284 Chr.13: 20413715-20413715 on Build GRCh38
rs991275949 Chr.13: 20413737-20413737 on Build GRCh38
rs758743245 Chr.13: 20413379-20413379 on Build GRCh38
rs1031062490 Chr.13: 20413470-20413470 on Build GRCh38
rs925062804 Chr.13: 20413368-20413368 on Build GRCh38
rs771731265 Chr.13: 20413429-20413429 on Build GRCh38
rs560097700 Chr.13: 20413441-20413441 on Build GRCh38
rs755903919 Chr.13: 20413398-20413398 on Build GRCh38
rs78984065 Chr.13: 20413406-20413406 on Build GRCh38
rs9509206 Chr.13: 20413589-20413589 on Build GRCh38
rs547425646 Chr.13: 20413665-20413665 on Build GRCh38
rs146695972 Chr.13: 20413480-20413481 on Build GRCh38
rs868272756 Chr.13: 20413383-20413383 on Build GRCh38
rs535805943 Chr.13: 20413519-20413519 on Build GRCh38
rs778322164 Chr.13: 20413416-20413416 on Build GRCh38
rs1046718917 Chr.13: 20413447-20413447 on Build GRCh38
rs893907221 Chr.13: 20413492-20413492 on Build GRCh38
rs754939005 Chr.13: 20413415-20413415 on Build GRCh38
rs925095620 Chr.13: 20413577-20413577 on Build GRCh38
rs1010829821 Chr.13: 20413508-20413508 on Build GRCh38
rs539999492 Chr.13: 20413593-20413593 on Build GRCh38
rs532656902 Chr.13: 20413658-20413658 on Build GRCh38
rs987876378 Chr.13: 20413588-20413588 on Build GRCh38
rs781566563 Chr.13: 20413820-20413820 on Build GRCh38
rs71426008 Chr.13: 20413538-20413538 on Build GRCh38
rs958042415 Chr.13: 20413579-20413579 on Build GRCh38
rs527940173 Chr.13: 20413499-20413499 on Build GRCh38
rs981757713 Chr.13: 20413689-20413689 on Build GRCh38
rs915573402 Chr.13: 20413742-20413742 on Build GRCh38
rs905434460 Chr.13: 20413449-20413449 on Build GRCh38
rs34494861 Chr.13: 20413469-20413470 on Build GRCh38
rs566072470 Chr.13: 20413745-20413745 on Build GRCh38
rs758780645 Chr.13: 20413416-20413418 on Build GRCh38
rs146532854 Chr.13: 20413544-20413544 on Build GRCh38
rs765305591 Chr.13: 20413373-20413373 on Build GRCh38
rs941091809 Chr.13: 20413735-20413735 on Build GRCh38
rs993566343 Chr.13: 20413510-20413510 on Build GRCh38
rs752790617 Chr.13: 20413378-20413378 on Build GRCh38
rs1019524980 Chr.13: 20413531-20413531 on Build GRCh38
rs1026740243 Chr.13: 20413522-20413522 on Build GRCh38
rs764364499 Chr.13: 20413391-20413391 on Build GRCh38
rs761275981 Chr.13: 20413836-20413837 on Build GRCh38
rs1002399440 Chr.13: 20413454-20413454 on Build GRCh38
rs951221585 Chr.13: 20413549-20413549 on Build GRCh38
rs116131020 Chr.13: 20413524-20413524 on Build GRCh38
rs747630994 Chr.13: 20413421-20413421 on Build GRCh38
rs1047121288 Chr.13: 20413394-20413394 on Build GRCh38
rs773860829 Chr.13: 20413441-20413441 on Build GRCh38
rs543335059 Chr.13: 20413523-20413523 on Build GRCh38
rs35966420 Chr.13: 20413461-20413462 on Build GRCh38
rs938220253 Chr.13: 20413730-20413730 on Build GRCh38
rs536290158 Chr.13: 20413836-20413836 on Build GRCh38
rs750276767 Chr.13: 20413392-20413392 on Build GRCh38
rs374268998 Chr.13: 20413407-20413407 on Build GRCh38
rs769264226 Chr.13: 20413721-20413721 on Build GRCh38
rs949112520 Chr.13: 20413683-20413683 on Build GRCh38
rs74036319 Chr.13: 20413357-20413357 on Build GRCh38

View Full Product Details