Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 11828018-11828519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCN6
Entrez Gene ID
1185
Gene Name
chloride voltage-gated channel 6
Gene Aliases
CLC-6
UniGene
Hs.193043
Gene Chromosome Location
Chr.1: 11806096-11843144 on Build GRCh38
Transcripts
NM_001256959.1 NM_001286.3 NR_046428.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5353153 c.883C>T p.R295C Chr.1: 11828148-11828148 on Build GRCh38
5715477 c.882C>T p.F294F Chr.1: 11828147-11828147 on Build GRCh38
6629895 c.883C>A p.R295S Chr.1: 11828148-11828148 on Build GRCh38
3472090 c.982C>T p.L328F Chr.1: 11828485-11828485 on Build GRCh38
234545 c.1011C>T p.F337F Chr.1: 11828514-11828514 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766982803 Chr.1: 11828258-11828258 on Build GRCh38
rs756256656 Chr.1: 11828393-11828393 on Build GRCh38
rs772513160 Chr.1: 11828171-11828171 on Build GRCh38
rs374439855 Chr.1: 11828101-11828101 on Build GRCh38
rs562629579 Chr.1: 11828073-11828073 on Build GRCh38
rs775875902 Chr.1: 11828464-11828464 on Build GRCh38
rs754280746 Chr.1: 11828299-11828299 on Build GRCh38
rs1017936780 Chr.1: 11828242-11828242 on Build GRCh38
rs533407817 Chr.1: 11828077-11828077 on Build GRCh38
rs566826626 Chr.1: 11828186-11828186 on Build GRCh38
rs748466916 Chr.1: 11828419-11828419 on Build GRCh38
rs779699582 Chr.1: 11828418-11828418 on Build GRCh38
rs778779004 Chr.1: 11828330-11828330 on Build GRCh38
rs937326132 Chr.1: 11828357-11828357 on Build GRCh38
rs748520221 Chr.1: 11828167-11828167 on Build GRCh38
rs111743209 Chr.1: 11828356-11828356 on Build GRCh38
rs755089605 Chr.1: 11828070-11828070 on Build GRCh38
rs750073103 Chr.1: 11828266-11828266 on Build GRCh38
rs765120680 Chr.1: 11828210-11828210 on Build GRCh38
rs762604276 Chr.1: 11828442-11828442 on Build GRCh38
rs780854681 Chr.1: 11828136-11828136 on Build GRCh38
rs762854886 Chr.1: 11828226-11828226 on Build GRCh38
rs973397317 Chr.1: 11828353-11828353 on Build GRCh38
rs774319486 Chr.1: 11828497-11828497 on Build GRCh38
rs368875103 Chr.1: 11828148-11828148 on Build GRCh38
rs768983318 Chr.1: 11828149-11828149 on Build GRCh38
rs773734538 Chr.1: 11828183-11828183 on Build GRCh38
rs751480840 Chr.1: 11828239-11828239 on Build GRCh38
rs987960466 Chr.1: 11828314-11828314 on Build GRCh38
rs998524162 Chr.1: 11828270-11828270 on Build GRCh38
rs747521070 Chr.1: 11828423-11828423 on Build GRCh38
rs976142394 Chr.1: 11828368-11828368 on Build GRCh38
rs770083031 Chr.1: 11828446-11828446 on Build GRCh38
rs538662051 Chr.1: 11828475-11828475 on Build GRCh38
rs577642663 Chr.1: 11828417-11828417 on Build GRCh38
rs762794667 Chr.1: 11828487-11828487 on Build GRCh38
rs198401 Chr.1: 11828327-11828327 on Build GRCh38
rs760769936 Chr.1: 11828185-11828185 on Build GRCh38
rs764018909 Chr.1: 11828493-11828493 on Build GRCh38
rs745448194 Chr.1: 11828341-11828341 on Build GRCh38
rs150459389 Chr.1: 11828100-11828100 on Build GRCh38
rs540067231 Chr.1: 11828227-11828227 on Build GRCh38
rs777856169 Chr.1: 11828092-11828092 on Build GRCh38
rs771722467 Chr.1: 11828371-11828371 on Build GRCh38
rs929582897 Chr.1: 11828403-11828403 on Build GRCh38
rs144929575 Chr.1: 11828211-11828211 on Build GRCh38
rs759735117 Chr.1: 11828196-11828196 on Build GRCh38
rs751021591 Chr.1: 11828094-11828094 on Build GRCh38
rs775384402 Chr.1: 11828232-11828233 on Build GRCh38
rs751838678 Chr.1: 11828056-11828056 on Build GRCh38
rs771030960 Chr.1: 11828424-11828424 on Build GRCh38
rs779303268 Chr.1: 11828164-11828164 on Build GRCh38
rs899475520 Chr.1: 11828260-11828260 on Build GRCh38
rs758797555 Chr.1: 11828420-11828420 on Build GRCh38
rs776642689 Chr.1: 11828429-11828429 on Build GRCh38
rs766577739 Chr.1: 11828058-11828058 on Build GRCh38
rs960647131 Chr.1: 11828301-11828301 on Build GRCh38
rs1020823549 Chr.1: 11828322-11828322 on Build GRCh38
rs140927950 Chr.1: 11828331-11828331 on Build GRCh38
rs879439807 Chr.1: 11828325-11828325 on Build GRCh38
rs776891800 Chr.1: 11828189-11828189 on Build GRCh38
rs367685242 Chr.1: 11828089-11828089 on Build GRCh38
rs1002088984 Chr.1: 11828201-11828201 on Build GRCh38
rs745946653 Chr.1: 11828437-11828437 on Build GRCh38
rs748232280 Chr.1: 11828386-11828387 on Build GRCh38
rs756757065 Chr.1: 11828241-11828241 on Build GRCh38
rs779569368 Chr.1: 11828404-11828404 on Build GRCh38
rs963415814 Chr.1: 11828340-11828340 on Build GRCh38
rs754041624 Chr.1: 11828062-11828062 on Build GRCh38
rs369653657 Chr.1: 11828291-11828291 on Build GRCh38

View Full Product Details