Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15301140-15301591 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
UniGene
Hs.659997
Gene Chromosome Location
Chr.1: 15236559-15400283 on Build GRCh38
Transcripts
NM_052929.1 XM_011540575.2 XM_011540576.2 XM_011540577.2 XM_011540580.2 XM_011540581.2 XM_011540582.2 XM_011540584.2 XM_011540588.2 XM_011540589.2 XM_011540590.2 XM_011540592.2 XM_011540593.2 XM_011540595.2 XM_011540597.2 XM_011540598.2 XM_017000198.1 XM_017000199.1 XM_017000200.1 XM_017000201.1 XM_017000202.1 XM_017000203.1 XM_017000204.1 XM_017000205.1 XM_017000206.1 XM_017000207.1 XM_017000208.1 XM_017000209.1 XM_017000210.1 XM_017000211.1 XM_017000212.1 XM_017000213.1 XM_017000214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5413659 c.810G>T p.E270D Chr.1: 15301336-15301336 on Build GRCh38
897189 c.876C>T p.I292I Chr.1: 15301402-15301402 on Build GRCh38
4222635 c.830A>G p.N277S Chr.1: 15301356-15301356 on Build GRCh38
4824440 c.716G>A p.R239H Chr.1: 15301242-15301242 on Build GRCh38
897186 c.751G>A p.D251N Chr.1: 15301277-15301277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371115593 Chr.1: 15301460-15301460 on Build GRCh38
rs547058385 Chr.1: 15301307-15301307 on Build GRCh38
rs745658743 Chr.1: 15301371-15301371 on Build GRCh38
rs772923462 Chr.1: 15301174-15301174 on Build GRCh38
rs990419066 Chr.1: 15301356-15301356 on Build GRCh38
rs977778835 Chr.1: 15301496-15301496 on Build GRCh38
rs12039283 Chr.1: 15301280-15301280 on Build GRCh38
rs914884936 Chr.1: 15301490-15301490 on Build GRCh38
rs971005434 Chr.1: 15301264-15301264 on Build GRCh38
rs1012646814 Chr.1: 15301241-15301241 on Build GRCh38
rs1056692297 Chr.1: 15301530-15301530 on Build GRCh38
rs149253803 Chr.1: 15301538-15301538 on Build GRCh38
rs566811075 Chr.1: 15301341-15301341 on Build GRCh38
rs894241035 Chr.1: 15301230-15301230 on Build GRCh38
rs946376212 Chr.1: 15301493-15301493 on Build GRCh38
rs551228538 Chr.1: 15301253-15301253 on Build GRCh38
rs186350869 Chr.1: 15301487-15301487 on Build GRCh38
rs181294222 Chr.1: 15301410-15301410 on Build GRCh38
rs953869823 Chr.1: 15301557-15301557 on Build GRCh38
rs200533304 Chr.1: 15301414-15301414 on Build GRCh38
rs894613328 Chr.1: 15301443-15301443 on Build GRCh38
rs758448498 Chr.1: 15301309-15301309 on Build GRCh38
rs986204492 Chr.1: 15301374-15301374 on Build GRCh38
rs768186300 Chr.1: 15301279-15301279 on Build GRCh38
rs77096706 Chr.1: 15301537-15301537 on Build GRCh38
rs576965055 Chr.1: 15301531-15301531 on Build GRCh38
rs533310005 Chr.1: 15301306-15301306 on Build GRCh38
rs531204038 Chr.1: 15301242-15301242 on Build GRCh38
rs117301380 Chr.1: 15301173-15301173 on Build GRCh38
rs1013085764 Chr.1: 15301477-15301477 on Build GRCh38
rs944728261 Chr.1: 15301391-15301391 on Build GRCh38
rs997788477 Chr.1: 15301560-15301560 on Build GRCh38
rs748993451 Chr.1: 15301367-15301367 on Build GRCh38
rs374819143 Chr.1: 15301348-15301348 on Build GRCh38
rs775148844 Chr.1: 15301400-15301400 on Build GRCh38
rs777348957 Chr.1: 15301463-15301463 on Build GRCh38
rs998513154 Chr.1: 15301511-15301511 on Build GRCh38
rs759119181 Chr.1: 15301532-15301532 on Build GRCh38
rs976656704 Chr.1: 15301423-15301423 on Build GRCh38
rs757209918 Chr.1: 15301347-15301347 on Build GRCh38
rs202211467 Chr.1: 15301328-15301328 on Build GRCh38
rs541870600 Chr.1: 15301565-15301565 on Build GRCh38
rs768170370 Chr.1: 15301404-15301404 on Build GRCh38
rs778730401 Chr.1: 15301433-15301433 on Build GRCh38
rs80173107 Chr.1: 15301550-15301550 on Build GRCh38
rs762537863 Chr.1: 15301190-15301190 on Build GRCh38
rs868352825 Chr.1: 15301359-15301361 on Build GRCh38
rs562673863 Chr.1: 15301402-15301402 on Build GRCh38
rs766796840 Chr.1: 15301183-15301183 on Build GRCh38
rs939168724 Chr.1: 15301260-15301260 on Build GRCh38
rs188187202 Chr.1: 15301167-15301167 on Build GRCh38
rs911904280 Chr.1: 15301385-15301385 on Build GRCh38
rs902312238 Chr.1: 15301254-15301254 on Build GRCh38
rs1028589043 Chr.1: 15301540-15301540 on Build GRCh38
rs556906560 Chr.1: 15301510-15301510 on Build GRCh38
rs748894561 Chr.1: 15301403-15301403 on Build GRCh38
rs769632640 Chr.1: 15301372-15301372 on Build GRCh38

View Full Product Details