Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15308522-15309024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
UniGene
Hs.659997
Gene Chromosome Location
Chr.1: 15236559-15400283 on Build GRCh38
Transcripts
NM_052929.1 XM_011540575.2 XM_011540576.2 XM_011540577.2 XM_011540580.2 XM_011540581.2 XM_011540582.2 XM_011540584.2 XM_011540588.2 XM_011540589.2 XM_011540590.2 XM_011540592.2 XM_011540593.2 XM_011540595.2 XM_011540597.2 XM_011540598.2 XM_017000198.1 XM_017000199.1 XM_017000200.1 XM_017000201.1 XM_017000202.1 XM_017000203.1 XM_017000204.1 XM_017000205.1 XM_017000206.1 XM_017000207.1 XM_017000208.1 XM_017000209.1 XM_017000210.1 XM_017000211.1 XM_017000212.1 XM_017000213.1 XM_017000214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6051748 c.1012T>G p.L338V Chr.1: 15308709-15308709 on Build GRCh38
4651970 c.956A>C p.Q319P Chr.1: 15308653-15308653 on Build GRCh38
4997545 c.34-4A>G p.? Chr.1: 15308609-15308609 on Build GRCh38
1600958 c.1035A>G p.T345T Chr.1: 15308732-15308732 on Build GRCh38
4997543 c.916-4A>G p.? Chr.1: 15308609-15308609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs866999910 Chr.1: 15308615-15308615 on Build GRCh38
rs776105854 Chr.1: 15308780-15308780 on Build GRCh38
rs763424963 Chr.1: 15308656-15308656 on Build GRCh38
rs1017910073 Chr.1: 15308842-15308842 on Build GRCh38
rs949067050 Chr.1: 15308664-15308664 on Build GRCh38
rs977003394 Chr.1: 15308699-15308699 on Build GRCh38
rs771644797 Chr.1: 15308763-15308763 on Build GRCh38
rs148232755 Chr.1: 15308585-15308585 on Build GRCh38
rs201389821 Chr.1: 15308640-15308640 on Build GRCh38
rs906104746 Chr.1: 15308550-15308550 on Build GRCh38
rs976657875 Chr.1: 15308907-15308907 on Build GRCh38
rs1044359122 Chr.1: 15308756-15308756 on Build GRCh38
rs548937894 Chr.1: 15308551-15308551 on Build GRCh38
rs747931259 Chr.1: 15308675-15308676 on Build GRCh38
rs756025985 Chr.1: 15308722-15308722 on Build GRCh38
rs758103101 Chr.1: 15308710-15308711 on Build GRCh38
rs951251414 Chr.1: 15308939-15308939 on Build GRCh38
rs575919945 Chr.1: 15308781-15308781 on Build GRCh38
rs1017606148 Chr.1: 15308776-15308776 on Build GRCh38
rs773116127 Chr.1: 15308832-15308832 on Build GRCh38
rs374360835 Chr.1: 15308667-15308667 on Build GRCh38
rs114852757 Chr.1: 15308798-15308798 on Build GRCh38
rs926330580 Chr.1: 15308989-15308989 on Build GRCh38
rs747819323 Chr.1: 15308758-15308758 on Build GRCh38
rs535466884 Chr.1: 15308774-15308774 on Build GRCh38
rs566528481 Chr.1: 15308737-15308737 on Build GRCh38
rs368985318 Chr.1: 15308723-15308723 on Build GRCh38
rs754872796 Chr.1: 15308752-15308752 on Build GRCh38
rs909326567 Chr.1: 15308979-15308979 on Build GRCh38
rs924799466 Chr.1: 15308581-15308581 on Build GRCh38
rs541350300 Chr.1: 15308753-15308753 on Build GRCh38
rs79547319 Chr.1: 15308775-15308775 on Build GRCh38
rs965682057 Chr.1: 15308845-15308845 on Build GRCh38
rs750317323 Chr.1: 15308718-15308718 on Build GRCh38
rs771769226 Chr.1: 15308571-15308571 on Build GRCh38
rs529500015 Chr.1: 15308727-15308727 on Build GRCh38
rs1018271462 Chr.1: 15308556-15308556 on Build GRCh38
rs942098087 Chr.1: 15308985-15308985 on Build GRCh38
rs755306609 Chr.1: 15308693-15308693 on Build GRCh38
rs577824017 Chr.1: 15308802-15308802 on Build GRCh38
rs767588298 Chr.1: 15308714-15308714 on Build GRCh38
rs41269435 Chr.1: 15308800-15308800 on Build GRCh38
rs550782073 Chr.1: 15308690-15308690 on Build GRCh38
rs749236561 Chr.1: 15308851-15308851 on Build GRCh38
rs969004829 Chr.1: 15308567-15308567 on Build GRCh38
rs747370305 Chr.1: 15308668-15308668 on Build GRCh38
rs770180074 Chr.1: 15308778-15308778 on Build GRCh38
rs770158092 Chr.1: 15308600-15308600 on Build GRCh38
rs10489961 Chr.1: 15309004-15309004 on Build GRCh38
rs988518428 Chr.1: 15308584-15308584 on Build GRCh38
rs368262816 Chr.1: 15308572-15308572 on Build GRCh38
rs540380444 Chr.1: 15308834-15308834 on Build GRCh38
rs987835871 Chr.1: 15308586-15308586 on Build GRCh38
rs1006579716 Chr.1: 15308837-15308837 on Build GRCh38
rs201939988 Chr.1: 15308702-15308702 on Build GRCh38
rs918498344 Chr.1: 15308935-15308935 on Build GRCh38
rs546729897 Chr.1: 15308729-15308729 on Build GRCh38
rs980808583 Chr.1: 15308959-15308959 on Build GRCh38
rs772859709 Chr.1: 15308663-15308663 on Build GRCh38
rs983907103 Chr.1: 15308960-15308960 on Build GRCh38

View Full Product Details