Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 15312971-15313476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FHAD1
Entrez Gene ID
114827
Gene Name
forkhead associated phosphopeptide binding domain 1
Gene Aliases
-
UniGene
Hs.659997
Gene Chromosome Location
Chr.1: 15236559-15400283 on Build GRCh38
Transcripts
NM_052929.1 XM_011540575.2 XM_011540576.2 XM_011540577.2 XM_011540580.2 XM_011540581.2 XM_011540582.2 XM_011540584.2 XM_011540588.2 XM_011540589.2 XM_011540590.2 XM_011540592.2 XM_011540593.2 XM_011540595.2 XM_011540596.2 XM_011540597.2 XM_011540598.2 XM_017000198.1 XM_017000199.1 XM_017000200.1 XM_017000201.1 XM_017000202.1 XM_017000203.1 XM_017000204.1 XM_017000205.1 XM_017000206.1 XM_017000207.1 XM_017000208.1 XM_017000209.1 XM_017000210.1 XM_017000211.1 XM_017000212.1 XM_017000213.1 XM_017000214.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6533580 c.1055T>G p.L352W Chr.1: 15313072-15313072 on Build GRCh38
6362148 c.272A>G p.E91G Chr.1: 15313171-15313171 on Build GRCh38
5698094 c.225G>T p.M75I Chr.1: 15313061-15313061 on Build GRCh38
5911330 c.1040-6T>A p.? Chr.1: 15313051-15313051 on Build GRCh38
5698093 c.1044G>T p.M348I Chr.1: 15313061-15313061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs534842505 Chr.1: 15313428-15313428 on Build GRCh38
rs376689986 Chr.1: 15313340-15313340 on Build GRCh38
rs745667006 Chr.1: 15313144-15313144 on Build GRCh38
rs565608881 Chr.1: 15313427-15313427 on Build GRCh38
rs892537104 Chr.1: 15313374-15313374 on Build GRCh38
rs916879595 Chr.1: 15313354-15313354 on Build GRCh38
rs753874975 Chr.1: 15313172-15313172 on Build GRCh38
rs777416009 Chr.1: 15313172-15313178 on Build GRCh38
rs550458698 Chr.1: 15313425-15313425 on Build GRCh38
rs754673665 Chr.1: 15313193-15313193 on Build GRCh38
rs776925191 Chr.1: 15312996-15312996 on Build GRCh38
rs934478260 Chr.1: 15313362-15313362 on Build GRCh38
rs966176957 Chr.1: 15313088-15313088 on Build GRCh38
rs530894965 Chr.1: 15313251-15313251 on Build GRCh38
rs972194936 Chr.1: 15313449-15313449 on Build GRCh38
rs948626318 Chr.1: 15313220-15313220 on Build GRCh38
rs577658369 Chr.1: 15313043-15313043 on Build GRCh38
rs556661286 Chr.1: 15313002-15313002 on Build GRCh38
rs536738679 Chr.1: 15313434-15313434 on Build GRCh38
rs968108592 Chr.1: 15313282-15313286 on Build GRCh38
rs765131238 Chr.1: 15313196-15313196 on Build GRCh38
rs978040834 Chr.1: 15313328-15313328 on Build GRCh38
rs79291577 Chr.1: 15313430-15313430 on Build GRCh38
rs377501179 Chr.1: 15313372-15313372 on Build GRCh38
rs367713922 Chr.1: 15313133-15313133 on Build GRCh38
rs948196235 Chr.1: 15313355-15313355 on Build GRCh38
rs777618645 Chr.1: 15313217-15313217 on Build GRCh38
rs1022996774 Chr.1: 15313041-15313041 on Build GRCh38
rs554790366 Chr.1: 15313327-15313327 on Build GRCh38
rs907069871 Chr.1: 15313277-15313277 on Build GRCh38
rs112338079 Chr.1: 15313205-15313205 on Build GRCh38
rs960493456 Chr.1: 15313350-15313350 on Build GRCh38
rs568183746 Chr.1: 15313433-15313433 on Build GRCh38
rs766225372 Chr.1: 15313148-15313148 on Build GRCh38
rs775950935 Chr.1: 15313202-15313215 on Build GRCh38
rs930335951 Chr.1: 15313176-15313176 on Build GRCh38
rs562229056 Chr.1: 15313234-15313234 on Build GRCh38
rs1034478697 Chr.1: 15313343-15313343 on Build GRCh38
rs956960394 Chr.1: 15313011-15313011 on Build GRCh38
rs939396537 Chr.1: 15313349-15313349 on Build GRCh38
rs1005316920 Chr.1: 15313373-15313373 on Build GRCh38
rs769537613 Chr.1: 15313206-15313206 on Build GRCh38
rs373780564 Chr.1: 15313100-15313100 on Build GRCh38
rs10489962 Chr.1: 15313085-15313085 on Build GRCh38
rs749846767 Chr.1: 15313221-15313221 on Build GRCh38
rs774488009 Chr.1: 15313018-15313018 on Build GRCh38
rs1001807147 Chr.1: 15313335-15313335 on Build GRCh38
rs770501771 Chr.1: 15313186-15313189 on Build GRCh38
rs1049267866 Chr.1: 15313371-15313371 on Build GRCh38
rs746574716 Chr.1: 15313181-15313184 on Build GRCh38
rs746519179 Chr.1: 15313033-15313033 on Build GRCh38
rs752359916 Chr.1: 15313199-15313199 on Build GRCh38
rs767533354 Chr.1: 15313080-15313080 on Build GRCh38
rs1012446491 Chr.1: 15313003-15313003 on Build GRCh38
rs201999060 Chr.1: 15313055-15313055 on Build GRCh38
rs149382716 Chr.1: 15313224-15313224 on Build GRCh38
rs971830072 Chr.1: 15313126-15313126 on Build GRCh38
rs1005181038 Chr.1: 15313039-15313039 on Build GRCh38
rs79093484 Chr.1: 15313424-15313424 on Build GRCh38
rs568840040 Chr.1: 15313448-15313448 on Build GRCh38

View Full Product Details