Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 24692700-24693195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP12A
Entrez Gene ID
479
Gene Name
ATPase H+/K+ transporting non-gastric alpha2 subunit
Gene Aliases
#945, ATP1AL1, HK
UniGene
Hs.147111
Gene Chromosome Location
Chr.13: 24680411-24711785 on Build GRCh38
Transcripts
NM_001185085.1 NM_001676.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7091981 c.1357G>A p.E453K Chr.13: 24692858-24692858 on Build GRCh38
222265 c.1342T>C p.F448L Chr.13: 24692861-24692861 on Build GRCh38
3885111 c.1304C>T p.S435F Chr.13: 24692823-24692823 on Build GRCh38
3765685 c.1275C>A p.V425V Chr.13: 24692794-24692794 on Build GRCh38
3793209 c.1339G>A p.E447K Chr.13: 24692858-24692858 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1032913457 Chr.13: 24692733-24692733 on Build GRCh38
rs536811573 Chr.13: 24692908-24692908 on Build GRCh38
rs535965576 Chr.13: 24693105-24693105 on Build GRCh38
rs771535571 Chr.13: 24692954-24692954 on Build GRCh38
rs56291145 Chr.13: 24692879-24692879 on Build GRCh38
rs759577074 Chr.13: 24692852-24692852 on Build GRCh38
rs954419461 Chr.13: 24693015-24693015 on Build GRCh38
rs1013210768 Chr.13: 24693135-24693135 on Build GRCh38
rs147362225 Chr.13: 24693122-24693122 on Build GRCh38
rs776660323 Chr.13: 24692849-24692849 on Build GRCh38
rs779263873 Chr.13: 24692829-24692829 on Build GRCh38
rs957944443 Chr.13: 24693081-24693081 on Build GRCh38
rs371308376 Chr.13: 24692786-24692786 on Build GRCh38
rs144196547 Chr.13: 24693114-24693114 on Build GRCh38
rs756823647 Chr.13: 24692758-24692758 on Build GRCh38
rs867916190 Chr.13: 24692885-24692885 on Build GRCh38
rs963984 Chr.13: 24692794-24692794 on Build GRCh38
rs750867795 Chr.13: 24692902-24692902 on Build GRCh38
rs201568944 Chr.13: 24692740-24692740 on Build GRCh38
rs1040414665 Chr.13: 24692968-24692968 on Build GRCh38
rs761287267 Chr.13: 24692895-24692895 on Build GRCh38
rs749471714 Chr.13: 24692826-24692826 on Build GRCh38
rs369272793 Chr.13: 24692743-24692743 on Build GRCh38
rs767048918 Chr.13: 24692896-24692896 on Build GRCh38
rs570411456 Chr.13: 24692948-24692948 on Build GRCh38
rs374909596 Chr.13: 24692934-24692934 on Build GRCh38
rs202206800 Chr.13: 24692788-24692788 on Build GRCh38
rs769564297 Chr.13: 24692853-24692853 on Build GRCh38
rs755413180 Chr.13: 24692929-24692929 on Build GRCh38
rs1017235775 Chr.13: 24693072-24693072 on Build GRCh38
rs878891335 Chr.13: 24692989-24692989 on Build GRCh38
rs774310407 Chr.13: 24692736-24692736 on Build GRCh38
rs754150188 Chr.13: 24692925-24692925 on Build GRCh38
rs764833751 Chr.13: 24692831-24692831 on Build GRCh38
rs963983 Chr.13: 24692950-24692950 on Build GRCh38
rs267603786 Chr.13: 24692876-24692876 on Build GRCh38
rs989200767 Chr.13: 24693085-24693085 on Build GRCh38
rs762516608 Chr.13: 24692750-24692750 on Build GRCh38
rs763598668 Chr.13: 24692753-24692753 on Build GRCh38
rs763793066 Chr.13: 24692754-24692754 on Build GRCh38
rs1055628156 Chr.13: 24693148-24693148 on Build GRCh38
rs1044093651 Chr.13: 24693145-24693145 on Build GRCh38
rs748885621 Chr.13: 24692773-24692774 on Build GRCh38
rs552937237 Chr.13: 24693018-24693018 on Build GRCh38
rs867713966 Chr.13: 24693020-24693020 on Build GRCh38
rs981893064 Chr.13: 24693113-24693113 on Build GRCh38
rs781638957 Chr.13: 24692780-24692780 on Build GRCh38
rs913813292 Chr.13: 24693093-24693093 on Build GRCh38
rs776269833 Chr.13: 24692724-24692724 on Build GRCh38
rs938118126 Chr.13: 24692819-24692819 on Build GRCh38
rs200821826 Chr.13: 24692913-24692913 on Build GRCh38
rs934501517 Chr.13: 24693052-24693052 on Build GRCh38
rs74643383 Chr.13: 24692889-24692889 on Build GRCh38
rs766417528 Chr.13: 24692741-24692741 on Build GRCh38
rs963985 Chr.13: 24692748-24692748 on Build GRCh38
rs768749384 Chr.13: 24692827-24692827 on Build GRCh38
rs772327642 Chr.13: 24692832-24692832 on Build GRCh38
rs1015859368 Chr.13: 24692901-24692901 on Build GRCh38
rs553316691 Chr.13: 24692958-24692958 on Build GRCh38
rs775159643 Chr.13: 24692859-24692859 on Build GRCh38
rs147563292 Chr.13: 24692891-24692891 on Build GRCh38
rs752353022 Chr.13: 24692749-24692749 on Build GRCh38
rs142939089 Chr.13: 24692801-24692801 on Build GRCh38

View Full Product Details