Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 26393144-26393668 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDK8
Entrez Gene ID
1024
Gene Name
cyclin dependent kinase 8
Gene Aliases
K35
UniGene
Hs.382306
Gene Chromosome Location
Chr.13: 26254104-26405238 on Build GRCh38
Transcripts
NM_001260.2 NM_001318368.1 XM_005266216.4 XM_011534864.1 XM_011534865.2 XM_017020317.1 XM_017020318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625603 c.757G>A p.D253N Chr.13: 26393477-26393477 on Build GRCh38
6925938 c.647-1G>C p.? Chr.13: 26393366-26393366 on Build GRCh38
1200579 c.709C>T p.R237* Chr.13: 26393429-26393429 on Build GRCh38
5376571 c.686C>T p.T229M Chr.13: 26393406-26393406 on Build GRCh38
1477168 c.790G>A p.D264N Chr.13: 26393510-26393510 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143032653 Chr.13: 26393550-26393550 on Build GRCh38
rs779887937 Chr.13: 26393322-26393322 on Build GRCh38
rs200393346 Chr.13: 26393240-26393240 on Build GRCh38
rs761586414 Chr.13: 26393352-26393352 on Build GRCh38
rs766303862 Chr.13: 26393548-26393548 on Build GRCh38
rs199692476 Chr.13: 26393317-26393317 on Build GRCh38
rs773127078 Chr.13: 26393382-26393382 on Build GRCh38
rs746239579 Chr.13: 26393352-26393352 on Build GRCh38
rs371948919 Chr.13: 26393327-26393327 on Build GRCh38
rs756446183 Chr.13: 26393479-26393479 on Build GRCh38
rs750825295 Chr.13: 26393548-26393550 on Build GRCh38
rs373367197 Chr.13: 26393480-26393480 on Build GRCh38
rs993713272 Chr.13: 26393499-26393499 on Build GRCh38
rs1010042898 Chr.13: 26393356-26393358 on Build GRCh38
rs1032140813 Chr.13: 26393537-26393537 on Build GRCh38
rs910056757 Chr.13: 26393572-26393572 on Build GRCh38
rs901631610 Chr.13: 26393296-26393296 on Build GRCh38
rs543336636 Chr.13: 26393573-26393573 on Build GRCh38
rs781580601 Chr.13: 26393520-26393520 on Build GRCh38
rs377752027 Chr.13: 26393221-26393221 on Build GRCh38
rs746177103 Chr.13: 26393523-26393523 on Build GRCh38
rs771389314 Chr.13: 26393348-26393348 on Build GRCh38
rs375527784 Chr.13: 26393416-26393416 on Build GRCh38
rs775575000 Chr.13: 26393361-26393361 on Build GRCh38
rs201316683 Chr.13: 26393524-26393524 on Build GRCh38
rs757970759 Chr.13: 26393470-26393470 on Build GRCh38
rs778368115 Chr.13: 26393344-26393344 on Build GRCh38
rs1049193920 Chr.13: 26393181-26393181 on Build GRCh38
rs374970075 Chr.13: 26393531-26393531 on Build GRCh38
rs1004601353 Chr.13: 26393240-26393240 on Build GRCh38
rs1006936817 Chr.13: 26393253-26393253 on Build GRCh38
rs765513398 Chr.13: 26393523-26393524 on Build GRCh38
rs888500469 Chr.13: 26393247-26393247 on Build GRCh38
rs190021037 Chr.13: 26393341-26393341 on Build GRCh38
rs747654897 Chr.13: 26393336-26393336 on Build GRCh38
rs942784424 Chr.13: 26393593-26393593 on Build GRCh38
rs1057404975 Chr.13: 26393216-26393216 on Build GRCh38
rs761906791 Chr.13: 26393546-26393546 on Build GRCh38
rs754765308 Chr.13: 26393414-26393414 on Build GRCh38
rs762959271 Chr.13: 26393363-26393363 on Build GRCh38
rs5802375 Chr.13: 26393593-26393593 on Build GRCh38
rs753623415 Chr.13: 26393413-26393413 on Build GRCh38
rs533603123 Chr.13: 26393491-26393491 on Build GRCh38
rs749575359 Chr.13: 26393528-26393528 on Build GRCh38
rs778290917 Chr.13: 26393544-26393544 on Build GRCh38
rs770017335 Chr.13: 26393358-26393358 on Build GRCh38
rs913286900 Chr.13: 26393588-26393588 on Build GRCh38
rs753403293 Chr.13: 26393323-26393323 on Build GRCh38
rs981818981 Chr.13: 26393640-26393640 on Build GRCh38
rs768217582 Chr.13: 26393340-26393341 on Build GRCh38
rs747494868 Chr.13: 26393514-26393514 on Build GRCh38
rs1015080289 Chr.13: 26393286-26393286 on Build GRCh38
rs112787342 Chr.13: 26393396-26393396 on Build GRCh38
rs988028327 Chr.13: 26393565-26393565 on Build GRCh38
rs760223405 Chr.13: 26393341-26393341 on Build GRCh38
rs778456747 Chr.13: 26393330-26393330 on Build GRCh38
rs747578844 Chr.13: 26393347-26393347 on Build GRCh38
rs764212936 Chr.13: 26393364-26393364 on Build GRCh38
rs182781177 Chr.13: 26393407-26393407 on Build GRCh38
rs891725757 Chr.13: 26393347-26393358 on Build GRCh38
rs369889152 Chr.13: 26393429-26393429 on Build GRCh38
rs753784914 Chr.13: 26393333-26393336 on Build GRCh38
rs757789047 Chr.13: 26393517-26393517 on Build GRCh38
rs930231461 Chr.13: 26393549-26393549 on Build GRCh38
rs777100485 Chr.13: 26393350-26393350 on Build GRCh38
rs17083996 Chr.13: 26393425-26393425 on Build GRCh38
rs760647749 Chr.13: 26393401-26393401 on Build GRCh38
rs549264774 Chr.13: 26393293-26393293 on Build GRCh38
rs753115898 Chr.13: 26393194-26393194 on Build GRCh38
rs369692411 Chr.13: 26393226-26393226 on Build GRCh38
rs950189269 Chr.13: 26393605-26393605 on Build GRCh38
rs751415326 Chr.13: 26393557-26393557 on Build GRCh38
rs376033714 Chr.13: 26393351-26393352 on Build GRCh38

View Full Product Details