Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28014372-28014872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3
Entrez Gene ID
2322
Gene Name
fms related tyrosine kinase 3
Gene Aliases
CD135, FLK-2, FLK2, STK1
UniGene
Hs.507590
Gene Chromosome Location
Chr.13: 28003274-28100592 on Build GRCh38
Transcripts
NM_004119.2 NR_130706.1 XM_011535015.2 XM_011535017.2 XM_011535018.2 XM_017020486.1 XM_017020487.1 XM_017020488.1 XM_017020489.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6138849 c.2857G>A p.A953T Chr.13: 28014454-28014454 on Build GRCh38
6138848 c.2797C>A p.R933R Chr.13: 28014514-28014514 on Build GRCh38
6915669 c.2780C>T p.A927V Chr.13: 28014531-28014531 on Build GRCh38
4987973 c.2770T>A p.S924T Chr.13: 28014541-28014541 on Build GRCh38
696846 c.2817G>T p.L939F Chr.13: 28014494-28014494 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781248096 Chr.13: 28014473-28014473 on Build GRCh38
rs763324475 Chr.13: 28014550-28014550 on Build GRCh38
rs943415452 Chr.13: 28014477-28014477 on Build GRCh38
rs548609046 Chr.13: 28014489-28014489 on Build GRCh38
rs530893616 Chr.13: 28014676-28014676 on Build GRCh38
rs770630954 Chr.13: 28014514-28014514 on Build GRCh38
rs1053006340 Chr.13: 28014632-28014632 on Build GRCh38
rs771849609 Chr.13: 28014588-28014590 on Build GRCh38
rs764308917 Chr.13: 28014557-28014557 on Build GRCh38
rs975408931 Chr.13: 28014773-28014773 on Build GRCh38
rs370451886 Chr.13: 28014403-28014403 on Build GRCh38
rs960956649 Chr.13: 28014715-28014715 on Build GRCh38
rs553135703 Chr.13: 28014778-28014778 on Build GRCh38
rs966303363 Chr.13: 28014769-28014769 on Build GRCh38
rs201025099 Chr.13: 28014488-28014488 on Build GRCh38
rs758146765 Chr.13: 28014502-28014502 on Build GRCh38
rs978507448 Chr.13: 28014795-28014795 on Build GRCh38
rs1043201739 Chr.13: 28014581-28014581 on Build GRCh38
rs777510370 Chr.13: 28014506-28014506 on Build GRCh38
rs370386010 Chr.13: 28014570-28014570 on Build GRCh38
rs923240583 Chr.13: 28014818-28014818 on Build GRCh38
rs753080745 Chr.13: 28014438-28014438 on Build GRCh38
rs1044201565 Chr.13: 28014688-28014688 on Build GRCh38
rs376897394 Chr.13: 28014689-28014689 on Build GRCh38
rs7994155 Chr.13: 28014411-28014411 on Build GRCh38
rs777210242 Chr.13: 28014427-28014427 on Build GRCh38
rs774626768 Chr.13: 28014559-28014559 on Build GRCh38
rs367849862 Chr.13: 28014519-28014519 on Build GRCh38
rs749569272 Chr.13: 28014562-28014562 on Build GRCh38
rs974980406 Chr.13: 28014546-28014546 on Build GRCh38
rs754904480 Chr.13: 28014576-28014576 on Build GRCh38
rs990805991 Chr.13: 28014611-28014621 on Build GRCh38
rs1026899249 Chr.13: 28014724-28014724 on Build GRCh38
rs568364150 Chr.13: 28014640-28014640 on Build GRCh38
rs778614556 Chr.13: 28014500-28014500 on Build GRCh38
rs771726006 Chr.13: 28014410-28014410 on Build GRCh38
rs869140671 Chr.13: 28014423-28014423 on Build GRCh38
rs983637380 Chr.13: 28014465-28014465 on Build GRCh38
rs934343630 Chr.13: 28014631-28014631 on Build GRCh38
rs925587322 Chr.13: 28014583-28014583 on Build GRCh38
rs1028165452 Chr.13: 28014717-28014717 on Build GRCh38
rs750486890 Chr.13: 28014573-28014573 on Build GRCh38
rs746521853 Chr.13: 28014509-28014509 on Build GRCh38
rs191829695 Chr.13: 28014713-28014713 on Build GRCh38
rs746716105 Chr.13: 28014595-28014595 on Build GRCh38
rs747936727 Chr.13: 28014501-28014501 on Build GRCh38
rs775744646 Chr.13: 28014539-28014539 on Build GRCh38
rs976101910 Chr.13: 28014799-28014799 on Build GRCh38
rs912176426 Chr.13: 28014470-28014470 on Build GRCh38
rs570582806 Chr.13: 28014694-28014694 on Build GRCh38
rs934574752 Chr.13: 28014827-28014827 on Build GRCh38
rs765576611 Chr.13: 28014431-28014431 on Build GRCh38
rs145998293 Chr.13: 28014453-28014453 on Build GRCh38
rs750975846 Chr.13: 28014777-28014777 on Build GRCh38
rs762012802 Chr.13: 28014562-28014562 on Build GRCh38
rs771577500 Chr.13: 28014424-28014424 on Build GRCh38
rs546562710 Chr.13: 28014793-28014793 on Build GRCh38
rs746408964 Chr.13: 28014521-28014521 on Build GRCh38
rs1014088787 Chr.13: 28014672-28014672 on Build GRCh38
rs764257600 Chr.13: 28014448-28014448 on Build GRCh38
rs372911679 Chr.13: 28014578-28014578 on Build GRCh38
rs752448694 Chr.13: 28014590-28014590 on Build GRCh38
rs531218543 Chr.13: 28014762-28014762 on Build GRCh38
rs751656163 Chr.13: 28014452-28014452 on Build GRCh38
rs756945583 Chr.13: 28014596-28014596 on Build GRCh38
rs1004865447 Chr.13: 28014763-28014763 on Build GRCh38
rs892937122 Chr.13: 28014584-28014584 on Build GRCh38
rs1019154997 Chr.13: 28014768-28014768 on Build GRCh38
rs760081935 Chr.13: 28014430-28014430 on Build GRCh38
rs375185557 Chr.13: 28014439-28014439 on Build GRCh38
rs925715591 Chr.13: 28014846-28014846 on Build GRCh38
rs368093531 Chr.13: 28014409-28014409 on Build GRCh38
rs758200027 Chr.13: 28014591-28014591 on Build GRCh38
rs111892726 Chr.13: 28014455-28014455 on Build GRCh38
rs770301265 Chr.13: 28014526-28014526 on Build GRCh38

View Full Product Details