Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 28301430-28301942 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT1
Entrez Gene ID
2321
Gene Name
fms related tyrosine kinase 1
Gene Aliases
FLT, FLT-1, VEGFR-1, VEGFR1
UniGene
Hs.594454
Gene Chromosome Location
Chr.13: 28300346-28495128 on Build GRCh38
Transcripts
NM_002019.4 XM_017020485.1

SNP Detail

SNP ID Chromosome Location
rs1042161031 Chr.13: 28301659-28301659 on Build GRCh38
rs757220180 Chr.13: 28301473-28301473 on Build GRCh38
rs761841033 Chr.13: 28301660-28301661 on Build GRCh38
rs567443851 Chr.13: 28301558-28301558 on Build GRCh38
rs555668854 Chr.13: 28301500-28301501 on Build GRCh38
rs1051376421 Chr.13: 28301696-28301696 on Build GRCh38
rs3829380 Chr.13: 28301899-28301899 on Build GRCh38
rs762870766 Chr.13: 28301564-28301564 on Build GRCh38
rs889712758 Chr.13: 28301475-28301475 on Build GRCh38
rs377153576 Chr.13: 28301641-28301641 on Build GRCh38
rs765107250 Chr.13: 28301525-28301525 on Build GRCh38
rs994147798 Chr.13: 28301782-28301782 on Build GRCh38
rs966 Chr.13: 28301652-28301652 on Build GRCh38
rs142404928 Chr.13: 28301706-28301706 on Build GRCh38
rs889700040 Chr.13: 28301701-28301701 on Build GRCh38
rs927534387 Chr.13: 28301584-28301584 on Build GRCh38
rs753281462 Chr.13: 28301465-28301465 on Build GRCh38
rs766225552 Chr.13: 28301647-28301647 on Build GRCh38
rs538858081 Chr.13: 28301665-28301665 on Build GRCh38
rs1016880317 Chr.13: 28301723-28301723 on Build GRCh38
rs547412297 Chr.13: 28301542-28301542 on Build GRCh38
rs767747786 Chr.13: 28301889-28301892 on Build GRCh38
rs571722084 Chr.13: 28301746-28301746 on Build GRCh38
rs561581929 Chr.13: 28301739-28301739 on Build GRCh38
rs374118849 Chr.13: 28301632-28301632 on Build GRCh38
rs971567211 Chr.13: 28301575-28301575 on Build GRCh38
rs982031547 Chr.13: 28301580-28301580 on Build GRCh38
rs972822469 Chr.13: 28301539-28301539 on Build GRCh38
rs937579231 Chr.13: 28301622-28301622 on Build GRCh38
rs1016932591 Chr.13: 28301522-28301522 on Build GRCh38
rs533930910 Chr.13: 28301912-28301912 on Build GRCh38
rs374604624 Chr.13: 28301839-28301839 on Build GRCh38
rs9554314 Chr.13: 28301652-28301652 on Build GRCh38
rs530043739 Chr.13: 28301573-28301573 on Build GRCh38
rs933809656 Chr.13: 28301687-28301687 on Build GRCh38
rs1025699223 Chr.13: 28301841-28301841 on Build GRCh38
rs190047650 Chr.13: 28301456-28301456 on Build GRCh38
rs971897984 Chr.13: 28301915-28301915 on Build GRCh38
rs549982355 Chr.13: 28301616-28301616 on Build GRCh38
rs914943630 Chr.13: 28301648-28301648 on Build GRCh38

View Full Product Details