Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 35769683-35770193 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCLK1
Entrez Gene ID
9201
Gene Name
doublecortin like kinase 1
Gene Aliases
CL1, CLICK1, DCAMKL1, DCDC3A, DCLK
UniGene
Hs.507755
Gene Chromosome Location
Chr.13: 35768652-36132020 on Build GRCh38
Transcripts
NM_001195415.1 NM_001195416.1 NM_004734.4 XM_005266592.2 XM_006719893.2 XM_017020847.1

SNP Detail

SNP ID Chromosome Location
rs775692978 Chr.13: 35769974-35769974 on Build GRCh38
rs1045176915 Chr.13: 35770135-35770135 on Build GRCh38
rs1041906777 Chr.13: 35770069-35770069 on Build GRCh38
rs188415033 Chr.13: 35770073-35770073 on Build GRCh38
rs561771292 Chr.13: 35769928-35769928 on Build GRCh38
rs1012325292 Chr.13: 35769772-35769772 on Build GRCh38
rs1014879751 Chr.13: 35770035-35770035 on Build GRCh38
rs973230682 Chr.13: 35770061-35770061 on Build GRCh38
rs951712072 Chr.13: 35769829-35769829 on Build GRCh38
rs951401483 Chr.13: 35769984-35769984 on Build GRCh38
rs760164270 Chr.13: 35769860-35769860 on Build GRCh38
rs982306723 Chr.13: 35770106-35770106 on Build GRCh38
rs970904722 Chr.13: 35769799-35769799 on Build GRCh38
rs1053820218 Chr.13: 35770097-35770097 on Build GRCh38
rs1029630206 Chr.13: 35769923-35769923 on Build GRCh38
rs915469280 Chr.13: 35770137-35770139 on Build GRCh38
rs946973517 Chr.13: 35770137-35770137 on Build GRCh38
rs906583652 Chr.13: 35770138-35770138 on Build GRCh38
rs922330629 Chr.13: 35770033-35770033 on Build GRCh38
rs1004798031 Chr.13: 35769985-35769986 on Build GRCh38
rs1006202255 Chr.13: 35770162-35770162 on Build GRCh38
rs146406427 Chr.13: 35770054-35770054 on Build GRCh38
rs963191641 Chr.13: 35770057-35770057 on Build GRCh38
rs142207103 Chr.13: 35769952-35769952 on Build GRCh38
rs1046029008 Chr.13: 35769710-35769710 on Build GRCh38
rs116097087 Chr.13: 35770107-35770107 on Build GRCh38
rs1032749549 Chr.13: 35769759-35769759 on Build GRCh38
rs975143666 Chr.13: 35770003-35770003 on Build GRCh38
rs982238406 Chr.13: 35769811-35769811 on Build GRCh38
rs142793851 Chr.13: 35770125-35770125 on Build GRCh38
rs1003750245 Chr.13: 35770143-35770143 on Build GRCh38
rs935582204 Chr.13: 35769762-35769762 on Build GRCh38
rs28633348 Chr.13: 35770025-35770025 on Build GRCh38
rs367796449 Chr.13: 35769764-35769764 on Build GRCh38
rs927634366 Chr.13: 35769761-35769761 on Build GRCh38
rs951688496 Chr.13: 35770151-35770151 on Build GRCh38
rs1011096753 Chr.13: 35769803-35769803 on Build GRCh38
rs998685847 Chr.13: 35769877-35769877 on Build GRCh38
rs565452932 Chr.13: 35770065-35770065 on Build GRCh38
rs1026233296 Chr.13: 35770149-35770149 on Build GRCh38
rs942479310 Chr.13: 35769932-35769932 on Build GRCh38
rs138214408 Chr.13: 35770092-35770092 on Build GRCh38
rs183410317 Chr.13: 35769862-35769862 on Build GRCh38
rs113095988 Chr.13: 35769769-35769769 on Build GRCh38
rs1023697400 Chr.13: 35769780-35769780 on Build GRCh38
rs148352154 Chr.13: 35769827-35769827 on Build GRCh38
rs933828363 Chr.13: 35770066-35770066 on Build GRCh38

View Full Product Details