Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36227015-36227541 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC169
Entrez Gene ID
728591
Gene Name
coiled-coil domain containing 169
Gene Aliases
C13orf38
UniGene
Hs.656745
Gene Chromosome Location
Chr.13: 36227042-36297855 on Build GRCh38
Transcripts
NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001198908.1 NM_001198910.1

Gene Symbol
CCDC169-SOHLH2
Entrez Gene ID
100526761
Gene Name
CCDC169-SOHLH2 readthrough
Gene Aliases
C13orf38-SOHLH2
UniGene
Hs.124519
Gene Chromosome Location
Chr.13: 36168208-36297855 on Build GRCh38
Transcripts
NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001198908.1 NM_001198910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1607042 c.333G>T p.E111D Chr.13: 36227294-36227294 on Build GRCh38
5376709 c.387C>T p.S129S Chr.13: 36227240-36227240 on Build GRCh38
3417564 c.337A>G p.R113G Chr.13: 36227290-36227290 on Build GRCh38
5376710 c.259G>A p.E87K Chr.13: 36227368-36227368 on Build GRCh38
5376711 c.249C>T p.L83L Chr.13: 36227378-36227378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs56183261 Chr.13: 36227491-36227491 on Build GRCh38
rs386769968 Chr.13: 36227511-36227517 on Build GRCh38
rs71869097 Chr.13: 36227510-36227511 on Build GRCh38
rs375041677 Chr.13: 36227263-36227263 on Build GRCh38
rs80133080 Chr.13: 36227516-36227516 on Build GRCh38
rs71084413 Chr.13: 36227490-36227491 on Build GRCh38
rs199540616 Chr.13: 36227500-36227503 on Build GRCh38
rs555419885 Chr.13: 36227444-36227444 on Build GRCh38
rs9546787 Chr.13: 36227450-36227450 on Build GRCh38
rs932045826 Chr.13: 36227481-36227482 on Build GRCh38
rs199823383 Chr.13: 36227425-36227425 on Build GRCh38
rs1051842346 Chr.13: 36227236-36227236 on Build GRCh38
rs184602518 Chr.13: 36227401-36227401 on Build GRCh38
rs386769967 Chr.13: 36227491-36227517 on Build GRCh38
rs376809763 Chr.13: 36227514-36227514 on Build GRCh38
rs758879084 Chr.13: 36227369-36227369 on Build GRCh38
rs769279522 Chr.13: 36227264-36227264 on Build GRCh38
rs55760726 Chr.13: 36227499-36227499 on Build GRCh38
rs66470163 Chr.13: 36227516-36227517 on Build GRCh38
rs1021749636 Chr.13: 36227313-36227313 on Build GRCh38
rs529493448 Chr.13: 36227314-36227314 on Build GRCh38
rs544991684 Chr.13: 36227128-36227128 on Build GRCh38
rs979705575 Chr.13: 36227395-36227395 on Build GRCh38
rs932076996 Chr.13: 36227216-36227216 on Build GRCh38
rs947400037 Chr.13: 36227184-36227185 on Build GRCh38
rs770312541 Chr.13: 36227354-36227354 on Build GRCh38
rs9546784 Chr.13: 36227119-36227119 on Build GRCh38
rs551764567 Chr.13: 36227422-36227422 on Build GRCh38
rs796513469 Chr.13: 36227298-36227298 on Build GRCh38
rs996193225 Chr.13: 36227181-36227181 on Build GRCh38
rs981906258 Chr.13: 36227109-36227109 on Build GRCh38
rs79208479 Chr.13: 36227517-36227517 on Build GRCh38
rs1043145227 Chr.13: 36227191-36227191 on Build GRCh38
rs754071661 Chr.13: 36227300-36227300 on Build GRCh38
rs552702205 Chr.13: 36227507-36227507 on Build GRCh38
rs148368546 Chr.13: 36227517-36227518 on Build GRCh38
rs752864140 Chr.13: 36227212-36227219 on Build GRCh38
rs988479780 Chr.13: 36227151-36227151 on Build GRCh38
rs71084414 Chr.13: 36227506-36227507 on Build GRCh38
rs879188663 Chr.13: 36227306-36227306 on Build GRCh38
rs533415818 Chr.13: 36227421-36227421 on Build GRCh38
rs56017034 Chr.13: 36227503-36227503 on Build GRCh38
rs45549236 Chr.13: 36227459-36227459 on Build GRCh38
rs149476823 Chr.13: 36227173-36227173 on Build GRCh38
rs992646916 Chr.13: 36227407-36227407 on Build GRCh38
rs938527975 Chr.13: 36227099-36227099 on Build GRCh38
rs563096566 Chr.13: 36227144-36227144 on Build GRCh38
rs956778427 Chr.13: 36227441-36227441 on Build GRCh38
rs898828875 Chr.13: 36227281-36227281 on Build GRCh38
rs748845831 Chr.13: 36227257-36227257 on Build GRCh38
rs756852571 Chr.13: 36227510-36227515 on Build GRCh38
rs540956747 Chr.13: 36227044-36227044 on Build GRCh38
rs202009084 Chr.13: 36227515-36227515 on Build GRCh38
rs763019865 Chr.13: 36227243-36227243 on Build GRCh38
rs1037460990 Chr.13: 36227148-36227148 on Build GRCh38
rs9546785 Chr.13: 36227278-36227278 on Build GRCh38
rs138475016 Chr.13: 36227475-36227476 on Build GRCh38
rs181271496 Chr.13: 36227047-36227047 on Build GRCh38
rs755597926 Chr.13: 36227201-36227201 on Build GRCh38
rs9546786 Chr.13: 36227442-36227442 on Build GRCh38
rs759880974 Chr.13: 36227302-36227302 on Build GRCh38
rs753172923 Chr.13: 36227336-36227336 on Build GRCh38
rs896573225 Chr.13: 36227135-36227135 on Build GRCh38
rs968408011 Chr.13: 36227376-36227376 on Build GRCh38
rs370953409 Chr.13: 36227289-36227289 on Build GRCh38
rs927165591 Chr.13: 36227053-36227053 on Build GRCh38
rs368543134 Chr.13: 36227400-36227400 on Build GRCh38
rs1008418268 Chr.13: 36227223-36227223 on Build GRCh38
rs779405960 Chr.13: 36227206-36227206 on Build GRCh38
rs113666985 Chr.13: 36227039-36227039 on Build GRCh38
rs542327259 Chr.13: 36227258-36227258 on Build GRCh38
rs386769969 Chr.13: 36227515-36227517 on Build GRCh38

View Full Product Details