Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 36230932-36231476 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC169
Entrez Gene ID
728591
Gene Name
coiled-coil domain containing 169
Gene Aliases
C13orf38
UniGene
Hs.656745
Gene Chromosome Location
Chr.13: 36227042-36297855 on Build GRCh38
Transcripts
NM_001144981.2 NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001144985.2 NM_001144986.2 NM_001198908.1 NM_001198910.1

Gene Symbol
CCDC169-SOHLH2
Entrez Gene ID
100526761
Gene Name
CCDC169-SOHLH2 readthrough
Gene Aliases
C13orf38-SOHLH2
UniGene
Hs.124519
Gene Chromosome Location
Chr.13: 36168208-36297855 on Build GRCh38
Transcripts
NM_001144981.2 NM_001144982.2 NM_001144983.2 NM_001144984.2 NM_001144985.2 NM_001144986.2 NM_001198908.1 NM_001198910.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1629083 c.598G>A p.V200I Chr.13: 36231240-36231240 on Build GRCh38
5510670 c.553T>A p.Y185N Chr.13: 36231285-36231285 on Build GRCh38
947127 c.604A>C p.K202Q Chr.13: 36231234-36231234 on Build GRCh38
6320345 c.568C>A p.Q190K Chr.13: 36231270-36231270 on Build GRCh38
5569039 c.612A>G p.T204T Chr.13: 36231226-36231226 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs548390968 Chr.13: 36230989-36230989 on Build GRCh38
rs9315397 Chr.13: 36231029-36231029 on Build GRCh38
rs894189161 Chr.13: 36231195-36231198 on Build GRCh38
rs201208352 Chr.13: 36231084-36231084 on Build GRCh38
rs957696446 Chr.13: 36231377-36231377 on Build GRCh38
rs192494063 Chr.13: 36231237-36231237 on Build GRCh38
rs964169828 Chr.13: 36231028-36231028 on Build GRCh38
rs976890035 Chr.13: 36231395-36231395 on Build GRCh38
rs574322268 Chr.13: 36231257-36231257 on Build GRCh38
rs1010829607 Chr.13: 36231385-36231385 on Build GRCh38
rs780039653 Chr.13: 36231233-36231233 on Build GRCh38
rs1032000288 Chr.13: 36231366-36231366 on Build GRCh38
rs760612372 Chr.13: 36231314-36231314 on Build GRCh38
rs534472466 Chr.13: 36231051-36231051 on Build GRCh38
rs983533162 Chr.13: 36231423-36231423 on Build GRCh38
rs183197251 Chr.13: 36231392-36231392 on Build GRCh38
rs867499340 Chr.13: 36231376-36231376 on Build GRCh38
rs909545073 Chr.13: 36231103-36231103 on Build GRCh38
rs774348002 Chr.13: 36231282-36231282 on Build GRCh38
rs372320109 Chr.13: 36231406-36231406 on Build GRCh38
rs556552844 Chr.13: 36231325-36231325 on Build GRCh38
rs911340830 Chr.13: 36231008-36231008 on Build GRCh38
rs1011835523 Chr.13: 36231390-36231390 on Build GRCh38
rs1052338724 Chr.13: 36231250-36231250 on Build GRCh38
rs1032930807 Chr.13: 36231302-36231302 on Build GRCh38
rs367765218 Chr.13: 36231409-36231409 on Build GRCh38
rs376568080 Chr.13: 36231217-36231217 on Build GRCh38
rs772974917 Chr.13: 36231034-36231034 on Build GRCh38
rs1047368365 Chr.13: 36231079-36231079 on Build GRCh38
rs67328439 Chr.13: 36231153-36231153 on Build GRCh38
rs577210414 Chr.13: 36231091-36231091 on Build GRCh38
rs368648363 Chr.13: 36231242-36231242 on Build GRCh38
rs1039306175 Chr.13: 36231200-36231200 on Build GRCh38
rs1018234417 Chr.13: 36231391-36231391 on Build GRCh38
rs745578534 Chr.13: 36231146-36231146 on Build GRCh38
rs2296532 Chr.13: 36231412-36231412 on Build GRCh38
rs764975511 Chr.13: 36231411-36231411 on Build GRCh38
rs866986833 Chr.13: 36231036-36231036 on Build GRCh38
rs999162191 Chr.13: 36231284-36231284 on Build GRCh38
rs999215850 Chr.13: 36231331-36231331 on Build GRCh38
rs2296531 Chr.13: 36231050-36231050 on Build GRCh38
rs910098641 Chr.13: 36231451-36231451 on Build GRCh38
rs909257005 Chr.13: 36231437-36231437 on Build GRCh38

View Full Product Details