Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 37103926-37104459 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSNK1A1L
Entrez Gene ID
122011
Gene Name
casein kinase 1 alpha 1 like
Gene Aliases
CK1
UniGene
Hs.512897
Gene Chromosome Location
Chr.13: 37103260-37105664 on Build GRCh38
Transcripts
NM_145203.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6636711 c.799C>T p.R267C Chr.13: 37104458-37104458 on Build GRCh38
432365 c.874T>C p.Y292H Chr.13: 37104383-37104383 on Build GRCh38
6636712 c.989delA p.N330fs*5 Chr.13: 37104268-37104268 on Build GRCh38
5842474 c.941G>A p.G314E Chr.13: 37104316-37104316 on Build GRCh38
1366720 c.989_990insA p.N330fs*4 Chr.13: 37104267-37104268 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1021710419 Chr.13: 37104144-37104144 on Build GRCh38
rs1038797440 Chr.13: 37104015-37104015 on Build GRCh38
rs376452704 Chr.13: 37104316-37104316 on Build GRCh38
rs769556012 Chr.13: 37104382-37104382 on Build GRCh38
rs559028749 Chr.13: 37104086-37104086 on Build GRCh38
rs781264924 Chr.13: 37104360-37104360 on Build GRCh38
rs547485893 Chr.13: 37104061-37104061 on Build GRCh38
rs754478303 Chr.13: 37104314-37104314 on Build GRCh38
rs766718454 Chr.13: 37104406-37104406 on Build GRCh38
rs769863283 Chr.13: 37104366-37104366 on Build GRCh38
rs754398759 Chr.13: 37104305-37104305 on Build GRCh38
rs553829073 Chr.13: 37104414-37104414 on Build GRCh38
rs777565290 Chr.13: 37104212-37104212 on Build GRCh38
rs746803410 Chr.13: 37104220-37104220 on Build GRCh38
rs752216443 Chr.13: 37104320-37104320 on Build GRCh38
rs774714492 Chr.13: 37104264-37104264 on Build GRCh38
rs201694550 Chr.13: 37104140-37104140 on Build GRCh38
rs761083671 Chr.13: 37104400-37104400 on Build GRCh38
rs766884383 Chr.13: 37104304-37104304 on Build GRCh38
rs759977373 Chr.13: 37104412-37104412 on Build GRCh38
rs767653119 Chr.13: 37104275-37104275 on Build GRCh38
rs908819169 Chr.13: 37103960-37103960 on Build GRCh38
rs1045864859 Chr.13: 37104396-37104396 on Build GRCh38
rs535577422 Chr.13: 37104364-37104364 on Build GRCh38
rs149364672 Chr.13: 37104257-37104257 on Build GRCh38
rs747425670 Chr.13: 37104361-37104361 on Build GRCh38
rs760983718 Chr.13: 37104303-37104303 on Build GRCh38
rs757887483 Chr.13: 37104433-37104433 on Build GRCh38
rs907294599 Chr.13: 37104070-37104070 on Build GRCh38
rs1011914700 Chr.13: 37104432-37104432 on Build GRCh38
rs752163482 Chr.13: 37104413-37104413 on Build GRCh38
rs773638205 Chr.13: 37104434-37104434 on Build GRCh38
rs370679467 Chr.13: 37104163-37104163 on Build GRCh38
rs908688278 Chr.13: 37104181-37104181 on Build GRCh38
rs886139846 Chr.13: 37103952-37103952 on Build GRCh38
rs771716813 Chr.13: 37104207-37104207 on Build GRCh38
rs770861569 Chr.13: 37104223-37104223 on Build GRCh38
rs1856028 Chr.13: 37104012-37104012 on Build GRCh38
rs748729425 Chr.13: 37104178-37104178 on Build GRCh38
rs758021029 Chr.13: 37104331-37104331 on Build GRCh38
rs763105662 Chr.13: 37104245-37104245 on Build GRCh38
rs186355010 Chr.13: 37104280-37104280 on Build GRCh38
rs1011467732 Chr.13: 37104123-37104123 on Build GRCh38
rs372797896 Chr.13: 37104204-37104204 on Build GRCh38
rs957279261 Chr.13: 37104167-37104167 on Build GRCh38
rs539426731 Chr.13: 37104081-37104081 on Build GRCh38
rs754528009 Chr.13: 37104198-37104198 on Build GRCh38
rs770682852 Chr.13: 37104354-37104354 on Build GRCh38
rs776475629 Chr.13: 37104240-37104240 on Build GRCh38
rs762010655 Chr.13: 37104383-37104383 on Build GRCh38
rs970273449 Chr.13: 37104071-37104071 on Build GRCh38
rs748942708 Chr.13: 37103997-37103997 on Build GRCh38
rs1047382806 Chr.13: 37103951-37103951 on Build GRCh38
rs568220564 Chr.13: 37104035-37104035 on Build GRCh38
rs776982240 Chr.13: 37104407-37104407 on Build GRCh38
rs755158301 Chr.13: 37103967-37103970 on Build GRCh38
rs139110306 Chr.13: 37104274-37104274 on Build GRCh38
rs746790307 Chr.13: 37104338-37104338 on Build GRCh38
rs571121591 Chr.13: 37104099-37104099 on Build GRCh38
rs546656835 Chr.13: 37104419-37104421 on Build GRCh38
rs529014050 Chr.13: 37104053-37104053 on Build GRCh38
rs761165501 Chr.13: 37104234-37104235 on Build GRCh38
rs778522002 Chr.13: 37104201-37104201 on Build GRCh38
rs565706213 Chr.13: 37104080-37104080 on Build GRCh38
rs913208486 Chr.13: 37104193-37104193 on Build GRCh38
rs777322085 Chr.13: 37104335-37104335 on Build GRCh38
rs911321683 Chr.13: 37104228-37104228 on Build GRCh38
rs995833221 Chr.13: 37104165-37104165 on Build GRCh38
rs891719164 Chr.13: 37104122-37104122 on Build GRCh38
rs772302382 Chr.13: 37104387-37104387 on Build GRCh38
rs1052943960 Chr.13: 37104113-37104113 on Build GRCh38
rs551077397 Chr.13: 37104267-37104268 on Build GRCh38
rs143083898 Chr.13: 37104388-37104388 on Build GRCh38
rs920132400 Chr.13: 37104313-37104313 on Build GRCh38

View Full Product Details