Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 38687359-38687859 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM2
Entrez Gene ID
341640
Gene Name
FRAS1 related extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.253994
Gene Chromosome Location
Chr.13: 38687036-38887131 on Build GRCh38
Transcripts
NM_207361.5 XM_017020554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
432375 c.311G>A p.R104H Chr.13: 38687655-38687655 on Build GRCh38
4614446 c.96_97delGC p.L33fs*36 Chr.13: 38687440-38687441 on Build GRCh38
5970982 c.376C>T p.R126C Chr.13: 38687720-38687720 on Build GRCh38
4601623 c.301C>G p.P101A Chr.13: 38687645-38687645 on Build GRCh38
3813786 c.388G>A p.D130N Chr.13: 38687732-38687732 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775779253 Chr.13: 38687650-38687650 on Build GRCh38
rs755417969 Chr.13: 38687380-38687380 on Build GRCh38
rs373948549 Chr.13: 38687821-38687821 on Build GRCh38
rs749142677 Chr.13: 38687510-38687510 on Build GRCh38
rs765718616 Chr.13: 38687382-38687382 on Build GRCh38
rs769086979 Chr.13: 38687672-38687672 on Build GRCh38
rs745648044 Chr.13: 38687424-38687424 on Build GRCh38
rs958374714 Chr.13: 38687519-38687519 on Build GRCh38
rs977767139 Chr.13: 38687829-38687829 on Build GRCh38
rs553605556 Chr.13: 38687525-38687525 on Build GRCh38
rs760739026 Chr.13: 38687559-38687559 on Build GRCh38
rs776940394 Chr.13: 38687802-38687802 on Build GRCh38
rs747131398 Chr.13: 38687616-38687616 on Build GRCh38
rs759453910 Chr.13: 38687431-38687433 on Build GRCh38
rs761379011 Chr.13: 38687567-38687567 on Build GRCh38
rs909150998 Chr.13: 38687670-38687670 on Build GRCh38
rs760256111 Chr.13: 38687795-38687795 on Build GRCh38
rs141718695 Chr.13: 38687428-38687428 on Build GRCh38
rs1001538066 Chr.13: 38687750-38687750 on Build GRCh38
rs867371419 Chr.13: 38687798-38687798 on Build GRCh38
rs757084129 Chr.13: 38687412-38687412 on Build GRCh38
rs139988077 Chr.13: 38687534-38687534 on Build GRCh38
rs765770038 Chr.13: 38687455-38687455 on Build GRCh38
rs763373816 Chr.13: 38687663-38687663 on Build GRCh38
rs759269194 Chr.13: 38687811-38687811 on Build GRCh38
rs767098305 Chr.13: 38687679-38687679 on Build GRCh38
rs886050187 Chr.13: 38687519-38687519 on Build GRCh38
rs774903381 Chr.13: 38687674-38687674 on Build GRCh38
rs768628258 Chr.13: 38687430-38687430 on Build GRCh38
rs748310975 Chr.13: 38687431-38687431 on Build GRCh38
rs769979597 Chr.13: 38687744-38687744 on Build GRCh38
rs780164812 Chr.13: 38687507-38687507 on Build GRCh38
rs752680276 Chr.13: 38687431-38687439 on Build GRCh38
rs886050186 Chr.13: 38687479-38687479 on Build GRCh38
rs1020251250 Chr.13: 38687816-38687816 on Build GRCh38
rs994570174 Chr.13: 38687446-38687446 on Build GRCh38
rs778905129 Chr.13: 38687613-38687613 on Build GRCh38
rs764017977 Chr.13: 38687825-38687825 on Build GRCh38
rs1026917863 Chr.13: 38687687-38687687 on Build GRCh38
rs750047827 Chr.13: 38687578-38687578 on Build GRCh38
rs771997262 Chr.13: 38687433-38687433 on Build GRCh38
rs975501544 Chr.13: 38687597-38687597 on Build GRCh38
rs144407623 Chr.13: 38687413-38687414 on Build GRCh38
rs568959201 Chr.13: 38687388-38687388 on Build GRCh38
rs1049931072 Chr.13: 38687609-38687609 on Build GRCh38
rs757356703 Chr.13: 38687721-38687721 on Build GRCh38
rs779648158 Chr.13: 38687582-38687582 on Build GRCh38
rs756201631 Chr.13: 38687836-38687836 on Build GRCh38
rs183560588 Chr.13: 38687453-38687453 on Build GRCh38
rs114864077 Chr.13: 38687727-38687727 on Build GRCh38
rs890055754 Chr.13: 38687427-38687427 on Build GRCh38
rs776324428 Chr.13: 38687806-38687806 on Build GRCh38
rs753120828 Chr.13: 38687465-38687465 on Build GRCh38
rs114929883 Chr.13: 38687712-38687712 on Build GRCh38
rs780296478 Chr.13: 38687745-38687745 on Build GRCh38
rs778761464 Chr.13: 38687714-38687714 on Build GRCh38
rs781080448 Chr.13: 38687415-38687415 on Build GRCh38
rs765038424 Chr.13: 38687814-38687814 on Build GRCh38
rs924908490 Chr.13: 38687830-38687830 on Build GRCh38
rs761430669 Chr.13: 38687676-38687676 on Build GRCh38
rs763682378 Chr.13: 38687753-38687753 on Build GRCh38
rs1011222435 Chr.13: 38687748-38687748 on Build GRCh38
rs751241265 Chr.13: 38687494-38687494 on Build GRCh38
rs765534709 Chr.13: 38687528-38687528 on Build GRCh38
rs777526420 Chr.13: 38687397-38687397 on Build GRCh38
rs777037145 Chr.13: 38687450-38687450 on Build GRCh38
rs368302333 Chr.13: 38687530-38687530 on Build GRCh38
rs936377898 Chr.13: 38687608-38687608 on Build GRCh38
rs773105875 Chr.13: 38687517-38687517 on Build GRCh38
rs746590736 Chr.13: 38687400-38687400 on Build GRCh38
rs749972037 Chr.13: 38687425-38687425 on Build GRCh38
rs750545395 Chr.13: 38687826-38687826 on Build GRCh38
rs750106928 Chr.13: 38687680-38687680 on Build GRCh38
rs115510540 Chr.13: 38687563-38687563 on Build GRCh38
rs370018440 Chr.13: 38687521-38687521 on Build GRCh38
rs771722643 Chr.13: 38687787-38687787 on Build GRCh38
rs994201858 Chr.13: 38687668-38687668 on Build GRCh38
rs201252176 Chr.13: 38687651-38687651 on Build GRCh38
rs754347397 Chr.13: 38687379-38687379 on Build GRCh38
rs773515555 Chr.13: 38687435-38687435 on Build GRCh38
rs1033475256 Chr.13: 38687771-38687771 on Build GRCh38
rs755951157 Chr.13: 38687505-38687505 on Build GRCh38
rs8002488 Chr.13: 38687647-38687647 on Build GRCh38
rs897385516 Chr.13: 38687654-38687654 on Build GRCh38
rs756006207 Chr.13: 38687581-38687581 on Build GRCh38
rs757813863 Chr.13: 38687393-38687393 on Build GRCh38
rs992393914 Chr.13: 38687784-38687784 on Build GRCh38
rs770972612 Chr.13: 38687641-38687641 on Build GRCh38
rs985248083 Chr.13: 38687837-38687837 on Build GRCh38
rs912727515 Chr.13: 38687576-38687576 on Build GRCh38
rs772914923 Chr.13: 38687789-38687789 on Build GRCh38
rs753825876 Chr.13: 38687591-38687591 on Build GRCh38
rs539210404 Chr.13: 38687606-38687606 on Build GRCh38
rs745938776 Chr.13: 38687730-38687730 on Build GRCh38
rs757945177 Chr.13: 38687466-38687466 on Build GRCh38
rs758270973 Chr.13: 38687824-38687824 on Build GRCh38
rs767329186 Chr.13: 38687575-38687575 on Build GRCh38
rs968071971 Chr.13: 38687782-38687782 on Build GRCh38
rs772196216 Chr.13: 38687512-38687512 on Build GRCh38
rs1048397841 Chr.13: 38687653-38687653 on Build GRCh38
rs377464365 Chr.13: 38687503-38687503 on Build GRCh38
rs749667994 Chr.13: 38687753-38687753 on Build GRCh38
rs1055473789 Chr.13: 38687728-38687728 on Build GRCh38
rs752578272 Chr.13: 38687715-38687715 on Build GRCh38
rs188243086 Chr.13: 38687708-38687708 on Build GRCh38
rs760741907 Chr.13: 38687444-38687444 on Build GRCh38
rs776093493 Chr.13: 38687532-38687532 on Build GRCh38
rs576435356 Chr.13: 38687649-38687649 on Build GRCh38
rs757033148 Chr.13: 38687495-38687495 on Build GRCh38
rs141831246 Chr.13: 38687584-38687584 on Build GRCh38
rs766742509 Chr.13: 38687449-38687449 on Build GRCh38
rs115327344 Chr.13: 38687520-38687520 on Build GRCh38
rs564740435 Chr.13: 38687781-38687781 on Build GRCh38
rs540644362 Chr.13: 38687506-38687506 on Build GRCh38
rs775538675 Chr.13: 38687429-38687429 on Build GRCh38
rs568736669 Chr.13: 38687430-38687431 on Build GRCh38
rs952810094 Chr.13: 38687725-38687725 on Build GRCh38
rs745701300 Chr.13: 38687504-38687504 on Build GRCh38
rs766039620 Chr.13: 38687701-38687701 on Build GRCh38
rs763468432 Chr.13: 38687472-38687472 on Build GRCh38
rs906971156 Chr.13: 38687479-38687479 on Build GRCh38
rs894207458 Chr.13: 38687743-38687743 on Build GRCh38
rs764651386 Chr.13: 38687539-38687539 on Build GRCh38
rs760344234 Chr.13: 38687684-38687684 on Build GRCh38
rs1002625446 Chr.13: 38687493-38687493 on Build GRCh38
rs769016740 Chr.13: 38687758-38687758 on Build GRCh38

View Full Product Details