Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 39659189-39659701 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COG6
Entrez Gene ID
57511
Gene Name
component of oligomeric golgi complex 6
Gene Aliases
CDG2L, COD2, SHNS
UniGene
Hs.507805
Gene Chromosome Location
Chr.13: 39655627-39791665 on Build GRCh38
Transcripts
NM_001145079.1 NM_020751.2 NR_026745.1 XM_011535168.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
696582 c.171C>G p.L57L Chr.13: 39659381-39659381 on Build GRCh38
4805021 c.231A>C p.G77G Chr.13: 39659441-39659441 on Build GRCh38
4805022 c.231A>C p.G77G Chr.13: 39659441-39659441 on Build GRCh38
3376549 c.215G>T p.R72L Chr.13: 39659425-39659425 on Build GRCh38
6139223 c.164A>T p.E55V Chr.13: 39659374-39659374 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767519557 Chr.13: 39659439-39659439 on Build GRCh38
rs537691906 Chr.13: 39659228-39659228 on Build GRCh38
rs765535257 Chr.13: 39659379-39659379 on Build GRCh38
rs759903662 Chr.13: 39659557-39659558 on Build GRCh38
rs1041500501 Chr.13: 39659475-39659475 on Build GRCh38
rs773250263 Chr.13: 39659440-39659440 on Build GRCh38
rs375697854 Chr.13: 39659339-39659339 on Build GRCh38
rs911223295 Chr.13: 39659287-39659287 on Build GRCh38
rs192746840 Chr.13: 39659273-39659273 on Build GRCh38
rs755280233 Chr.13: 39659375-39659375 on Build GRCh38
rs368063880 Chr.13: 39659653-39659653 on Build GRCh38
rs371925123 Chr.13: 39659431-39659431 on Build GRCh38
rs759330421 Chr.13: 39659216-39659216 on Build GRCh38
rs769390337 Chr.13: 39659332-39659332 on Build GRCh38
rs536442492 Chr.13: 39659314-39659314 on Build GRCh38
rs371078610 Chr.13: 39659329-39659329 on Build GRCh38
rs947722660 Chr.13: 39659624-39659624 on Build GRCh38
rs781694353 Chr.13: 39659485-39659485 on Build GRCh38
rs370151162 Chr.13: 39659452-39659452 on Build GRCh38
rs756551996 Chr.13: 39659618-39659618 on Build GRCh38
rs201199239 Chr.13: 39659315-39659315 on Build GRCh38
rs201308360 Chr.13: 39659365-39659365 on Build GRCh38
rs747454714 Chr.13: 39659316-39659316 on Build GRCh38
rs138314667 Chr.13: 39659225-39659225 on Build GRCh38
rs748884193 Chr.13: 39659425-39659425 on Build GRCh38
rs779429072 Chr.13: 39659544-39659544 on Build GRCh38
rs763779789 Chr.13: 39659342-39659342 on Build GRCh38
rs755708215 Chr.13: 39659526-39659526 on Build GRCh38
rs545116031 Chr.13: 39659394-39659394 on Build GRCh38
rs146261996 Chr.13: 39659471-39659471 on Build GRCh38
rs751870618 Chr.13: 39659470-39659470 on Build GRCh38
rs7330016 Chr.13: 39659554-39659554 on Build GRCh38
rs1043444824 Chr.13: 39659266-39659266 on Build GRCh38
rs575237298 Chr.13: 39659429-39659429 on Build GRCh38
rs748977488 Chr.13: 39659547-39659547 on Build GRCh38
rs761838797 Chr.13: 39659353-39659353 on Build GRCh38
rs7330515 Chr.13: 39659267-39659267 on Build GRCh38
rs144131145 Chr.13: 39659637-39659637 on Build GRCh38
rs1025505779 Chr.13: 39659572-39659572 on Build GRCh38
rs745344420 Chr.13: 39659418-39659418 on Build GRCh38
rs773954326 Chr.13: 39659436-39659436 on Build GRCh38
rs769263578 Chr.13: 39659419-39659419 on Build GRCh38
rs947721153 Chr.13: 39659243-39659243 on Build GRCh38
rs759735412 Chr.13: 39659446-39659446 on Build GRCh38
rs781415420 Chr.13: 39659386-39659386 on Build GRCh38
rs753029623 Chr.13: 39659451-39659451 on Build GRCh38
rs370792989 Chr.13: 39659333-39659333 on Build GRCh38
rs771300717 Chr.13: 39659548-39659549 on Build GRCh38
rs572987538 Chr.13: 39659324-39659324 on Build GRCh38
rs960609866 Chr.13: 39659574-39659574 on Build GRCh38
rs764976535 Chr.13: 39659232-39659232 on Build GRCh38
rs374915399 Chr.13: 39659499-39659499 on Build GRCh38
rs761857311 Chr.13: 39659437-39659437 on Build GRCh38
rs1000059859 Chr.13: 39659622-39659622 on Build GRCh38
rs752836835 Chr.13: 39659381-39659381 on Build GRCh38
rs61754106 Chr.13: 39659384-39659384 on Build GRCh38
rs992043776 Chr.13: 39659589-39659589 on Build GRCh38
rs765213953 Chr.13: 39659447-39659447 on Build GRCh38
rs775183788 Chr.13: 39659334-39659334 on Build GRCh38
rs766573924 Chr.13: 39659357-39659357 on Build GRCh38
rs768138297 Chr.13: 39659553-39659553 on Build GRCh38
rs762565644 Chr.13: 39659361-39659362 on Build GRCh38
rs141356976 Chr.13: 39659455-39659455 on Build GRCh38
rs368678409 Chr.13: 39659352-39659352 on Build GRCh38
rs780705604 Chr.13: 39659413-39659413 on Build GRCh38
rs776050050 Chr.13: 39659328-39659328 on Build GRCh38
rs916505109 Chr.13: 39659600-39659600 on Build GRCh38
rs61753158 Chr.13: 39659424-39659424 on Build GRCh38
rs558503643 Chr.13: 39659395-39659395 on Build GRCh38
rs985899875 Chr.13: 39659276-39659276 on Build GRCh38
rs774768041 Chr.13: 39659552-39659553 on Build GRCh38

View Full Product Details