Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42887450-42887948 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPSTI1
Entrez Gene ID
94240
Gene Name
epithelial stromal interaction 1 (breast)
Gene Aliases
BRESI1
UniGene
Hs.546467
Gene Chromosome Location
Chr.13: 42886388-42992271 on Build GRCh38
Transcripts
NM_001002264.2 NM_033255.3 XM_005266596.1 XM_005266597.1 XM_011535312.1 XM_011535313.1 XM_011535314.1 XM_017020853.1 XM_017020854.1 XM_017020855.1

SNP Detail

SNP ID Chromosome Location
rs2281810 Chr.13: 42887507-42887507 on Build GRCh38
rs560019160 Chr.13: 42887567-42887567 on Build GRCh38
rs961856258 Chr.13: 42887681-42887681 on Build GRCh38
rs370861286 Chr.13: 42887901-42887901 on Build GRCh38
rs913058071 Chr.13: 42887562-42887562 on Build GRCh38
rs528761770 Chr.13: 42887601-42887601 on Build GRCh38
rs912719284 Chr.13: 42887492-42887492 on Build GRCh38
rs1020560045 Chr.13: 42887765-42887765 on Build GRCh38
rs888414004 Chr.13: 42887647-42887647 on Build GRCh38
rs985156555 Chr.13: 42887742-42887742 on Build GRCh38
rs956985631 Chr.13: 42887778-42887778 on Build GRCh38
rs964527295 Chr.13: 42887788-42887788 on Build GRCh38
rs149083816 Chr.13: 42887800-42887800 on Build GRCh38
rs1046091626 Chr.13: 42887581-42887581 on Build GRCh38
rs975194090 Chr.13: 42887793-42887793 on Build GRCh38
rs1046239855 Chr.13: 42887565-42887565 on Build GRCh38
rs949291865 Chr.13: 42887564-42887564 on Build GRCh38
rs1011560111 Chr.13: 42887735-42887735 on Build GRCh38
rs935725581 Chr.13: 42887545-42887545 on Build GRCh38
rs140241335 Chr.13: 42887894-42887894 on Build GRCh38
rs535999559 Chr.13: 42887779-42887779 on Build GRCh38
rs919612310 Chr.13: 42887534-42887534 on Build GRCh38
rs967648786 Chr.13: 42887766-42887766 on Build GRCh38
rs905763678 Chr.13: 42887587-42887587 on Build GRCh38
rs1005595749 Chr.13: 42887656-42887656 on Build GRCh38
rs774214647 Chr.13: 42887579-42887579 on Build GRCh38
rs140369921 Chr.13: 42887516-42887516 on Build GRCh38
rs764004063 Chr.13: 42887566-42887566 on Build GRCh38
rs138557442 Chr.13: 42887855-42887855 on Build GRCh38
rs531075034 Chr.13: 42887746-42887746 on Build GRCh38
rs908199782 Chr.13: 42887902-42887902 on Build GRCh38
rs953872258 Chr.13: 42887517-42887517 on Build GRCh38
rs1030351128 Chr.13: 42887718-42887720 on Build GRCh38
rs554023966 Chr.13: 42887665-42887667 on Build GRCh38
rs1046988382 Chr.13: 42887558-42887558 on Build GRCh38
rs117661149 Chr.13: 42887559-42887559 on Build GRCh38
rs71202240 Chr.13: 42887675-42887675 on Build GRCh38
rs925731874 Chr.13: 42887541-42887541 on Build GRCh38
rs538540183 Chr.13: 42887868-42887868 on Build GRCh38
rs565272597 Chr.13: 42887743-42887743 on Build GRCh38
rs79684361 Chr.13: 42887664-42887664 on Build GRCh38
rs1002657499 Chr.13: 42887602-42887602 on Build GRCh38
rs998351325 Chr.13: 42887701-42887701 on Build GRCh38
rs574177880 Chr.13: 42887537-42887537 on Build GRCh38
rs145020588 Chr.13: 42887781-42887781 on Build GRCh38
rs144724358 Chr.13: 42887801-42887801 on Build GRCh38
rs761741126 Chr.13: 42887622-42887622 on Build GRCh38
rs919648750 Chr.13: 42887794-42887794 on Build GRCh38

View Full Product Details