Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42889072-42889570 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPSTI1
Entrez Gene ID
94240
Gene Name
epithelial stromal interaction 1 (breast)
Gene Aliases
BRESI1
UniGene
Hs.546467
Gene Chromosome Location
Chr.13: 42886388-42992271 on Build GRCh38
Transcripts
NM_001002264.2 NM_033255.3 XM_005266596.1 XM_005266597.1 XM_011535312.1 XM_011535313.1 XM_011535314.1 XM_017020853.1 XM_017020854.1 XM_017020855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232261 c.968C>T p.S323L Chr.13: 42889223-42889223 on Build GRCh38
3987413 c.970A>T p.R324W Chr.13: 42889221-42889221 on Build GRCh38
3987412 c.971G>A p.R324K Chr.13: 42889220-42889220 on Build GRCh38
7207909 c.958T>C p.L320L Chr.13: 42889233-42889233 on Build GRCh38
1167816 c.952_953insGGGTT p.S318fs*5 Chr.13: 42889238-42889239 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764792026 Chr.13: 42889422-42889422 on Build GRCh38
rs928935169 Chr.13: 42889219-42889219 on Build GRCh38
rs773075052 Chr.13: 42889275-42889276 on Build GRCh38
rs1018835676 Chr.13: 42889236-42889236 on Build GRCh38
rs984167358 Chr.13: 42889375-42889375 on Build GRCh38
rs906700952 Chr.13: 42889133-42889133 on Build GRCh38
rs760411510 Chr.13: 42889192-42889192 on Build GRCh38
rs972960943 Chr.13: 42889446-42889446 on Build GRCh38
rs762249608 Chr.13: 42889171-42889171 on Build GRCh38
rs897457134 Chr.13: 42889542-42889542 on Build GRCh38
rs576896781 Chr.13: 42889215-42889215 on Build GRCh38
rs954781322 Chr.13: 42889107-42889107 on Build GRCh38
rs375973689 Chr.13: 42889223-42889224 on Build GRCh38
rs943015966 Chr.13: 42889319-42889319 on Build GRCh38
rs928212606 Chr.13: 42889450-42889450 on Build GRCh38
rs756313179 Chr.13: 42889225-42889225 on Build GRCh38
rs538528842 Chr.13: 42889162-42889162 on Build GRCh38
rs747437711 Chr.13: 42889261-42889261 on Build GRCh38
rs765931339 Chr.13: 42889193-42889193 on Build GRCh38
rs768826582 Chr.13: 42889235-42889235 on Build GRCh38
rs59267170 Chr.13: 42889223-42889223 on Build GRCh38
rs777547704 Chr.13: 42889222-42889222 on Build GRCh38
rs776387264 Chr.13: 42889272-42889272 on Build GRCh38
rs537728981 Chr.13: 42889510-42889510 on Build GRCh38
rs374404304 Chr.13: 42889232-42889232 on Build GRCh38
rs1037325510 Chr.13: 42889516-42889516 on Build GRCh38
rs964070475 Chr.13: 42889322-42889322 on Build GRCh38
rs991714876 Chr.13: 42889457-42889457 on Build GRCh38
rs78219530 Chr.13: 42889166-42889166 on Build GRCh38
rs772962171 Chr.13: 42889180-42889180 on Build GRCh38
rs993380704 Chr.13: 42889256-42889256 on Build GRCh38
rs990132801 Chr.13: 42889353-42889353 on Build GRCh38
rs909917151 Chr.13: 42889410-42889410 on Build GRCh38
rs941670766 Chr.13: 42889234-42889234 on Build GRCh38
rs200098794 Chr.13: 42889230-42889230 on Build GRCh38
rs928796475 Chr.13: 42889431-42889431 on Build GRCh38
rs916734738 Chr.13: 42889495-42889495 on Build GRCh38
rs764833741 Chr.13: 42889276-42889276 on Build GRCh38
rs939587194 Chr.13: 42889455-42889455 on Build GRCh38
rs1043453900 Chr.13: 42889206-42889206 on Build GRCh38
rs372229110 Chr.13: 42889212-42889212 on Build GRCh38
rs762515761 Chr.13: 42889277-42889278 on Build GRCh38
rs79111014 Chr.13: 42889242-42889242 on Build GRCh38
rs544789853 Chr.13: 42889357-42889357 on Build GRCh38
rs753276401 Chr.13: 42889197-42889197 on Build GRCh38
rs939494586 Chr.13: 42889139-42889139 on Build GRCh38
rs568559573 Chr.13: 42889263-42889264 on Build GRCh38
rs779242577 Chr.13: 42889237-42889237 on Build GRCh38
rs916149905 Chr.13: 42889506-42889506 on Build GRCh38
rs772591715 Chr.13: 42889178-42889178 on Build GRCh38
rs752305015 Chr.13: 42889473-42889473 on Build GRCh38
rs563086477 Chr.13: 42889262-42889262 on Build GRCh38
rs776521078 Chr.13: 42889252-42889253 on Build GRCh38
rs986032620 Chr.13: 42889109-42889109 on Build GRCh38
rs765054633 Chr.13: 42889208-42889208 on Build GRCh38
rs557667468 Chr.13: 42889511-42889511 on Build GRCh38
rs770485045 Chr.13: 42889263-42889263 on Build GRCh38
rs572863853 Chr.13: 42889214-42889214 on Build GRCh38
rs531548641 Chr.13: 42889233-42889233 on Build GRCh38
rs779263323 Chr.13: 42889217-42889217 on Build GRCh38
rs527305553 Chr.13: 42889260-42889260 on Build GRCh38
rs552844267 Chr.13: 42889545-42889545 on Build GRCh38
rs1042275409 Chr.13: 42889216-42889216 on Build GRCh38
rs772436897 Chr.13: 42889239-42889239 on Build GRCh38
rs765875561 Chr.13: 42889283-42889285 on Build GRCh38
rs1323864 Chr.13: 42889163-42889163 on Build GRCh38
rs147274859 Chr.13: 42889111-42889111 on Build GRCh38
rs540578279 Chr.13: 42889207-42889207 on Build GRCh38
rs561977519 Chr.13: 42889258-42889258 on Build GRCh38
rs759295083 Chr.13: 42889182-42889182 on Build GRCh38
rs1017506294 Chr.13: 42889301-42889301 on Build GRCh38
rs977497433 Chr.13: 42889423-42889423 on Build GRCh38
rs375982143 Chr.13: 42889274-42889274 on Build GRCh38
rs1029229008 Chr.13: 42889265-42889265 on Build GRCh38

View Full Product Details