Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 42900232-42900746 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPSTI1
Entrez Gene ID
94240
Gene Name
epithelial stromal interaction 1 (breast)
Gene Aliases
BRESI1
UniGene
Hs.546467
Gene Chromosome Location
Chr.13: 42886388-42992271 on Build GRCh38
Transcripts
NM_001002264.2 NM_033255.3 XM_005266596.1 XM_005266597.1 XM_006719896.3 XM_011535312.1 XM_011535313.1 XM_011535314.1 XM_017020853.1 XM_017020854.1 XM_017020855.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5889229 c.846G>A p.R282R Chr.13: 42900312-42900312 on Build GRCh38
4575941 c.810G>A p.E270E Chr.13: 42900348-42900348 on Build GRCh38
4891283 c.796C>T p.R266W Chr.13: 42900362-42900362 on Build GRCh38
3468994 c.845G>A p.R282K Chr.13: 42900313-42900313 on Build GRCh38
6139172 c.848G>T p.R283M Chr.13: 42900310-42900310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748118768 Chr.13: 42900543-42900543 on Build GRCh38
rs940699491 Chr.13: 42900499-42900499 on Build GRCh38
rs964029637 Chr.13: 42900617-42900617 on Build GRCh38
rs768173803 Chr.13: 42900280-42900280 on Build GRCh38
rs771711127 Chr.13: 42900307-42900307 on Build GRCh38
rs756474171 Chr.13: 42900344-42900344 on Build GRCh38
rs1041422355 Chr.13: 42900556-42900556 on Build GRCh38
rs533957128 Chr.13: 42900385-42900385 on Build GRCh38
rs746030726 Chr.13: 42900341-42900341 on Build GRCh38
rs756480395 Chr.13: 42900413-42900413 on Build GRCh38
rs774235178 Chr.13: 42900282-42900282 on Build GRCh38
rs966996990 Chr.13: 42900374-42900374 on Build GRCh38
rs756376524 Chr.13: 42900445-42900445 on Build GRCh38
rs780075707 Chr.13: 42900466-42900466 on Build GRCh38
rs138836186 Chr.13: 42900582-42900582 on Build GRCh38
rs997126645 Chr.13: 42900687-42900687 on Build GRCh38
rs369430222 Chr.13: 42900343-42900343 on Build GRCh38
rs143039845 Chr.13: 42900427-42900437 on Build GRCh38
rs975313348 Chr.13: 42900451-42900451 on Build GRCh38
rs907888416 Chr.13: 42900479-42900479 on Build GRCh38
rs546234745 Chr.13: 42900476-42900476 on Build GRCh38
rs748771561 Chr.13: 42900269-42900269 on Build GRCh38
rs1011302313 Chr.13: 42900555-42900555 on Build GRCh38
rs767177454 Chr.13: 42900282-42900282 on Build GRCh38
rs191616831 Chr.13: 42900714-42900714 on Build GRCh38
rs902927841 Chr.13: 42900563-42900563 on Build GRCh38
rs919747137 Chr.13: 42900454-42900454 on Build GRCh38
rs757769505 Chr.13: 42900388-42900388 on Build GRCh38
rs1045741107 Chr.13: 42900527-42900527 on Build GRCh38
rs773049270 Chr.13: 42900314-42900314 on Build GRCh38
rs551999508 Chr.13: 42900580-42900580 on Build GRCh38
rs896897529 Chr.13: 42900643-42900643 on Build GRCh38
rs17543853 Chr.13: 42900706-42900706 on Build GRCh38
rs753018513 Chr.13: 42900361-42900361 on Build GRCh38
rs765525873 Chr.13: 42900352-42900352 on Build GRCh38
rs753017662 Chr.13: 42900343-42900344 on Build GRCh38
rs1019557773 Chr.13: 42900395-42900395 on Build GRCh38
rs143282676 Chr.13: 42900535-42900535 on Build GRCh38
rs9594829 Chr.13: 42900692-42900692 on Build GRCh38
rs1005213761 Chr.13: 42900616-42900616 on Build GRCh38
rs937580232 Chr.13: 42900542-42900542 on Build GRCh38
rs907432924 Chr.13: 42900281-42900281 on Build GRCh38
rs940939885 Chr.13: 42900276-42900276 on Build GRCh38
rs529931808 Chr.13: 42900410-42900410 on Build GRCh38
rs185926890 Chr.13: 42900673-42900673 on Build GRCh38
rs910583095 Chr.13: 42900359-42900359 on Build GRCh38
rs764272606 Chr.13: 42900371-42900371 on Build GRCh38
rs560884402 Chr.13: 42900362-42900362 on Build GRCh38
rs12867175 Chr.13: 42900497-42900497 on Build GRCh38
rs559542680 Chr.13: 42900551-42900551 on Build GRCh38
rs768566682 Chr.13: 42900501-42900501 on Build GRCh38
rs1056363635 Chr.13: 42900284-42900284 on Build GRCh38
rs774095389 Chr.13: 42900539-42900539 on Build GRCh38
rs749436392 Chr.13: 42900467-42900467 on Build GRCh38
rs781645842 Chr.13: 42900337-42900337 on Build GRCh38
rs112763870 Chr.13: 42900574-42900574 on Build GRCh38
rs751951757 Chr.13: 42900375-42900375 on Build GRCh38
rs149173137 Chr.13: 42900283-42900283 on Build GRCh38
rs938291274 Chr.13: 42900564-42900564 on Build GRCh38
rs182894006 Chr.13: 42900590-42900590 on Build GRCh38
rs781615402 Chr.13: 42900393-42900393 on Build GRCh38
rs1015232769 Chr.13: 42900633-42900633 on Build GRCh38
rs907834638 Chr.13: 42900272-42900272 on Build GRCh38

View Full Product Details