Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43085652-43086157 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
UniGene
Hs.438830
Gene Chromosome Location
Chr.13: 43023226-43109170 on Build GRCh38
Transcripts
NM_013238.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4678720 c.356T>C p.V119A Chr.13: 43085812-43085812 on Build GRCh38
4047714 c.319G>A p.A107T Chr.13: 43085775-43085775 on Build GRCh38
1366981 c.312-1G>A p.? Chr.13: 43085767-43085767 on Build GRCh38
947570 c.369T>G p.N123K Chr.13: 43085825-43085825 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs994428585 Chr.13: 43085733-43085733 on Build GRCh38
rs756740765 Chr.13: 43085774-43085774 on Build GRCh38
rs900460925 Chr.13: 43085683-43085683 on Build GRCh38
rs756655319 Chr.13: 43085868-43085868 on Build GRCh38
rs930558998 Chr.13: 43085959-43085959 on Build GRCh38
rs761278008 Chr.13: 43085720-43085720 on Build GRCh38
rs539356057 Chr.13: 43086132-43086133 on Build GRCh38
rs183745861 Chr.13: 43085886-43085886 on Build GRCh38
rs537282387 Chr.13: 43085771-43085771 on Build GRCh38
rs763160185 Chr.13: 43085748-43085748 on Build GRCh38
rs545335726 Chr.13: 43085946-43085946 on Build GRCh38
rs989474449 Chr.13: 43085921-43085921 on Build GRCh38
rs116995864 Chr.13: 43086011-43086011 on Build GRCh38
rs867307560 Chr.13: 43085768-43085768 on Build GRCh38
rs773076307 Chr.13: 43085809-43085809 on Build GRCh38
rs768331848 Chr.13: 43085793-43085793 on Build GRCh38
rs930561928 Chr.13: 43085705-43085705 on Build GRCh38
rs71813178 Chr.13: 43085909-43085910 on Build GRCh38
rs944534798 Chr.13: 43086002-43086002 on Build GRCh38
rs904543929 Chr.13: 43086110-43086110 on Build GRCh38
rs753692528 Chr.13: 43085744-43085744 on Build GRCh38
rs907701494 Chr.13: 43085985-43085985 on Build GRCh38
rs761890863 Chr.13: 43085835-43085835 on Build GRCh38
rs374055433 Chr.13: 43085870-43085870 on Build GRCh38
rs367964575 Chr.13: 43085737-43085737 on Build GRCh38
rs1028746595 Chr.13: 43085828-43085828 on Build GRCh38
rs778452626 Chr.13: 43085802-43085802 on Build GRCh38
rs950472198 Chr.13: 43085772-43085772 on Build GRCh38
rs771996026 Chr.13: 43085724-43085724 on Build GRCh38
rs758264142 Chr.13: 43085861-43085862 on Build GRCh38
rs1044471164 Chr.13: 43085700-43085700 on Build GRCh38
rs12428117 Chr.13: 43085984-43085984 on Build GRCh38
rs766053495 Chr.13: 43085942-43085942 on Build GRCh38
rs746937885 Chr.13: 43085812-43085812 on Build GRCh38
rs754205846 Chr.13: 43085881-43085881 on Build GRCh38
rs886310745 Chr.13: 43085722-43085722 on Build GRCh38
rs930573159 Chr.13: 43086047-43086047 on Build GRCh38
rs60311912 Chr.13: 43085910-43085911 on Build GRCh38
rs750595613 Chr.13: 43085861-43085861 on Build GRCh38
rs181060890 Chr.13: 43085827-43085827 on Build GRCh38
rs749708619 Chr.13: 43085789-43085789 on Build GRCh38
rs764735605 Chr.13: 43086043-43086043 on Build GRCh38
rs1024470118 Chr.13: 43086100-43086100 on Build GRCh38
rs979765187 Chr.13: 43085950-43085950 on Build GRCh38
rs757310008 Chr.13: 43085757-43085757 on Build GRCh38
rs750188466 Chr.13: 43085855-43085855 on Build GRCh38
rs886089913 Chr.13: 43086080-43086080 on Build GRCh38
rs763777316 Chr.13: 43085968-43085968 on Build GRCh38
rs139689863 Chr.13: 43085969-43085969 on Build GRCh38
rs773054604 Chr.13: 43085732-43085732 on Build GRCh38
rs772110439 Chr.13: 43085840-43085840 on Build GRCh38
rs570685870 Chr.13: 43086119-43086119 on Build GRCh38
rs764267263 Chr.13: 43085750-43085750 on Build GRCh38
rs920285452 Chr.13: 43085958-43085959 on Build GRCh38
rs375767738 Chr.13: 43085831-43085831 on Build GRCh38
rs781672288 Chr.13: 43085758-43085758 on Build GRCh38
rs1004573648 Chr.13: 43086084-43086084 on Build GRCh38
rs773879400 Chr.13: 43085719-43085719 on Build GRCh38
rs369956960 Chr.13: 43085746-43085746 on Build GRCh38
rs771488218 Chr.13: 43085808-43085808 on Build GRCh38
rs1049736440 Chr.13: 43086099-43086099 on Build GRCh38
rs141024500 Chr.13: 43085908-43085909 on Build GRCh38
rs906962033 Chr.13: 43086101-43086101 on Build GRCh38
rs532658813 Chr.13: 43085919-43085919 on Build GRCh38
rs370069729 Chr.13: 43085854-43085854 on Build GRCh38
rs545446622 Chr.13: 43086118-43086118 on Build GRCh38
rs1000447563 Chr.13: 43086111-43086111 on Build GRCh38
rs959583700 Chr.13: 43086127-43086127 on Build GRCh38
rs544464692 Chr.13: 43086026-43086026 on Build GRCh38
rs73476111 Chr.13: 43085943-43085943 on Build GRCh38
rs552118338 Chr.13: 43086068-43086068 on Build GRCh38
rs531818240 Chr.13: 43085781-43085781 on Build GRCh38
rs1005051943 Chr.13: 43085786-43085786 on Build GRCh38
rs1000882013 Chr.13: 43085872-43085872 on Build GRCh38
rs751640255 Chr.13: 43085756-43085756 on Build GRCh38
rs776382656 Chr.13: 43085826-43085826 on Build GRCh38
rs747654771 Chr.13: 43085803-43085803 on Build GRCh38
rs770923944 Chr.13: 43085822-43085822 on Build GRCh38
rs920482854 Chr.13: 43085701-43085701 on Build GRCh38
rs553343620 Chr.13: 43085929-43085929 on Build GRCh38
rs75881215 Chr.13: 43085913-43085913 on Build GRCh38
rs959615783 Chr.13: 43085900-43085900 on Build GRCh38
rs760396231 Chr.13: 43085735-43085735 on Build GRCh38

View Full Product Details