Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43107802-43108347 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC15
Entrez Gene ID
29103
Gene Name
DnaJ heat shock protein family (Hsp40) member C15
Gene Aliases
DNAJD1, HSD18, MCJ
UniGene
Hs.438830
Gene Chromosome Location
Chr.13: 43023226-43109170 on Build GRCh38
Transcripts
NM_013238.2

SNP Detail

SNP ID Chromosome Location
rs73476126 Chr.13: 43108324-43108324 on Build GRCh38
rs751918957 Chr.13: 43108071-43108071 on Build GRCh38
rs890600798 Chr.13: 43108305-43108305 on Build GRCh38
rs891462562 Chr.13: 43108274-43108274 on Build GRCh38
rs974353334 Chr.13: 43108276-43108279 on Build GRCh38
rs780221230 Chr.13: 43108318-43108318 on Build GRCh38
rs983560028 Chr.13: 43108315-43108315 on Build GRCh38
rs554468242 Chr.13: 43108120-43108120 on Build GRCh38
rs980198757 Chr.13: 43107949-43107949 on Build GRCh38
rs765824841 Chr.13: 43108013-43108013 on Build GRCh38
rs1029843803 Chr.13: 43108295-43108295 on Build GRCh38
rs138599487 Chr.13: 43108007-43108007 on Build GRCh38
rs949875852 Chr.13: 43108188-43108188 on Build GRCh38
rs1032699881 Chr.13: 43107999-43107999 on Build GRCh38
rs529848022 Chr.13: 43108207-43108207 on Build GRCh38
rs903632161 Chr.13: 43108158-43108159 on Build GRCh38
rs111250117 Chr.13: 43108287-43108287 on Build GRCh38
rs920700632 Chr.13: 43107963-43107963 on Build GRCh38
rs930660176 Chr.13: 43107966-43107966 on Build GRCh38
rs999848116 Chr.13: 43107969-43107969 on Build GRCh38
rs539316881 Chr.13: 43108219-43108219 on Build GRCh38
rs370760509 Chr.13: 43107902-43107902 on Build GRCh38
rs774052252 Chr.13: 43107900-43107900 on Build GRCh38
rs969176249 Chr.13: 43108266-43108266 on Build GRCh38
rs895675369 Chr.13: 43107845-43107845 on Build GRCh38
rs1014148013 Chr.13: 43107865-43107865 on Build GRCh38
rs969206847 Chr.13: 43107941-43107941 on Build GRCh38
rs766193175 Chr.13: 43107977-43107977 on Build GRCh38
rs1017825217 Chr.13: 43108027-43108027 on Build GRCh38
rs17064184 Chr.13: 43107827-43107827 on Build GRCh38
rs74062352 Chr.13: 43108074-43108074 on Build GRCh38
rs368663313 Chr.13: 43107842-43107842 on Build GRCh38
rs987659542 Chr.13: 43108025-43108025 on Build GRCh38
rs750969992 Chr.13: 43108081-43108089 on Build GRCh38
rs149259050 Chr.13: 43108010-43108010 on Build GRCh38
rs181034620 Chr.13: 43108149-43108149 on Build GRCh38
rs79830825 Chr.13: 43107891-43107891 on Build GRCh38
rs781597566 Chr.13: 43108094-43108094 on Build GRCh38
rs916391846 Chr.13: 43107909-43107909 on Build GRCh38
rs1054118810 Chr.13: 43108260-43108260 on Build GRCh38
rs750628255 Chr.13: 43108218-43108218 on Build GRCh38
rs777814192 Chr.13: 43108123-43108123 on Build GRCh38
rs1027732377 Chr.13: 43108284-43108284 on Build GRCh38
rs917052749 Chr.13: 43108128-43108128 on Build GRCh38
rs764649373 Chr.13: 43108065-43108065 on Build GRCh38
rs552083798 Chr.13: 43107946-43107946 on Build GRCh38
rs879440666 Chr.13: 43107952-43107952 on Build GRCh38
rs114341170 Chr.13: 43108063-43108063 on Build GRCh38
rs753216039 Chr.13: 43108050-43108050 on Build GRCh38
rs905758526 Chr.13: 43108216-43108216 on Build GRCh38
rs567024782 Chr.13: 43108184-43108184 on Build GRCh38
rs2491100 Chr.13: 43108189-43108189 on Build GRCh38
rs1010434191 Chr.13: 43108011-43108011 on Build GRCh38
rs955047718 Chr.13: 43108019-43108019 on Build GRCh38
rs190624422 Chr.13: 43107884-43107884 on Build GRCh38
rs944902655 Chr.13: 43108040-43108040 on Build GRCh38

View Full Product Details