Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43836054-43836572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC122
Entrez Gene ID
160857
Gene Name
coiled-coil domain containing 122
Gene Aliases
-
UniGene
Hs.170849
Gene Chromosome Location
Chr.13: 43821959-43879690 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020399.1

SNP Detail

SNP ID Chromosome Location
rs769296996 Chr.13: 43836209-43836209 on Build GRCh38
rs959892722 Chr.13: 43836286-43836286 on Build GRCh38
rs996050165 Chr.13: 43836360-43836360 on Build GRCh38
rs911120235 Chr.13: 43836415-43836417 on Build GRCh38
rs537468209 Chr.13: 43836472-43836472 on Build GRCh38
rs192341917 Chr.13: 43836516-43836516 on Build GRCh38
rs951426870 Chr.13: 43836154-43836154 on Build GRCh38
rs1051863561 Chr.13: 43836324-43836324 on Build GRCh38
rs564666367 Chr.13: 43836191-43836191 on Build GRCh38
rs555622884 Chr.13: 43836287-43836290 on Build GRCh38
rs936483315 Chr.13: 43836274-43836274 on Build GRCh38
rs200026745 Chr.13: 43836417-43836417 on Build GRCh38
rs962890119 Chr.13: 43836346-43836346 on Build GRCh38
rs559551266 Chr.13: 43836332-43836332 on Build GRCh38
rs187054485 Chr.13: 43836458-43836458 on Build GRCh38
rs1018500249 Chr.13: 43836345-43836345 on Build GRCh38
rs561098523 Chr.13: 43836385-43836385 on Build GRCh38
rs1041438949 Chr.13: 43836527-43836527 on Build GRCh38
rs759053159 Chr.13: 43836208-43836208 on Build GRCh38
rs936535585 Chr.13: 43836481-43836481 on Build GRCh38
rs1012392217 Chr.13: 43836313-43836313 on Build GRCh38
rs180929840 Chr.13: 43836276-43836276 on Build GRCh38
rs543099246 Chr.13: 43836157-43836157 on Build GRCh38
rs9533646 Chr.13: 43836533-43836533 on Build GRCh38
rs1051489072 Chr.13: 43836536-43836536 on Build GRCh38
rs1052677035 Chr.13: 43836517-43836517 on Build GRCh38
rs560719855 Chr.13: 43836109-43836109 on Build GRCh38
rs141419168 Chr.13: 43836140-43836140 on Build GRCh38
rs1007155483 Chr.13: 43836333-43836333 on Build GRCh38
rs925170885 Chr.13: 43836476-43836476 on Build GRCh38
rs925118553 Chr.13: 43836263-43836263 on Build GRCh38
rs951529999 Chr.13: 43836384-43836384 on Build GRCh38
rs892194343 Chr.13: 43836283-43836283 on Build GRCh38
rs1025756447 Chr.13: 43836376-43836376 on Build GRCh38
rs12872943 Chr.13: 43836417-43836417 on Build GRCh38
rs189629794 Chr.13: 43836155-43836155 on Build GRCh38
rs185609655 Chr.13: 43836102-43836102 on Build GRCh38
rs1051015399 Chr.13: 43836101-43836101 on Build GRCh38
rs549174993 Chr.13: 43836310-43836310 on Build GRCh38
rs530877479 Chr.13: 43836118-43836118 on Build GRCh38
rs934020661 Chr.13: 43836099-43836099 on Build GRCh38
rs962894963 Chr.13: 43836354-43836354 on Build GRCh38
rs933056800 Chr.13: 43836534-43836534 on Build GRCh38
rs1002527635 Chr.13: 43836267-43836268 on Build GRCh38

View Full Product Details