Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43858665-43859168 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC122
Entrez Gene ID
160857
Gene Name
coiled-coil domain containing 122
Gene Aliases
-
UniGene
Hs.170849
Gene Chromosome Location
Chr.13: 43821959-43879690 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020398.1 XM_017020399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6621390 c.664G>T p.E222* Chr.13: 43858789-43858789 on Build GRCh38
3733840 c.599A>G p.E200G Chr.13: 43858854-43858854 on Build GRCh38
6621389 c.664G>T p.E222* Chr.13: 43858789-43858789 on Build GRCh38
4867365 c.581A>C p.K194T Chr.13: 43858872-43858872 on Build GRCh38
5642484 c.641A>G p.K214R Chr.13: 43858812-43858812 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs905338244 Chr.13: 43859004-43859005 on Build GRCh38
rs1027500664 Chr.13: 43858699-43858699 on Build GRCh38
rs1014568305 Chr.13: 43859039-43859039 on Build GRCh38
rs542025353 Chr.13: 43859088-43859088 on Build GRCh38
rs772594271 Chr.13: 43858810-43858810 on Build GRCh38
rs376428525 Chr.13: 43858870-43858870 on Build GRCh38
rs781181080 Chr.13: 43858778-43858778 on Build GRCh38
rs931453453 Chr.13: 43859016-43859016 on Build GRCh38
rs775432374 Chr.13: 43858766-43858767 on Build GRCh38
rs1004201279 Chr.13: 43858725-43858725 on Build GRCh38
rs946717675 Chr.13: 43858707-43858707 on Build GRCh38
rs765411178 Chr.13: 43858947-43858947 on Build GRCh38
rs751519947 Chr.13: 43858869-43858869 on Build GRCh38
rs991351614 Chr.13: 43859133-43859133 on Build GRCh38
rs916617127 Chr.13: 43858704-43858704 on Build GRCh38
rs752776038 Chr.13: 43858948-43858948 on Build GRCh38
rs778994996 Chr.13: 43858789-43858789 on Build GRCh38
rs199699870 Chr.13: 43858837-43858837 on Build GRCh38
rs771257193 Chr.13: 43858824-43858824 on Build GRCh38
rs77930776 Chr.13: 43858747-43858747 on Build GRCh38
rs780676800 Chr.13: 43858942-43858942 on Build GRCh38
rs1048584278 Chr.13: 43859017-43859017 on Build GRCh38
rs975329970 Chr.13: 43858767-43858767 on Build GRCh38
rs992450964 Chr.13: 43858944-43858944 on Build GRCh38
rs750258517 Chr.13: 43858901-43858901 on Build GRCh38
rs949606533 Chr.13: 43858979-43858979 on Build GRCh38
rs774995481 Chr.13: 43858853-43858853 on Build GRCh38
rs960899431 Chr.13: 43859086-43859086 on Build GRCh38
rs557372368 Chr.13: 43859074-43859074 on Build GRCh38
rs151127816 Chr.13: 43858821-43858823 on Build GRCh38
rs972992990 Chr.13: 43859067-43859067 on Build GRCh38
rs1035628946 Chr.13: 43859081-43859081 on Build GRCh38
rs916919768 Chr.13: 43858963-43858964 on Build GRCh38
rs377395606 Chr.13: 43858746-43858746 on Build GRCh38
rs771530767 Chr.13: 43858892-43858892 on Build GRCh38
rs755185979 Chr.13: 43858923-43858923 on Build GRCh38
rs760634194 Chr.13: 43858777-43858777 on Build GRCh38
rs1028675979 Chr.13: 43859130-43859130 on Build GRCh38
rs777043512 Chr.13: 43858825-43858825 on Build GRCh38
rs916666507 Chr.13: 43859141-43859141 on Build GRCh38
rs59946000 Chr.13: 43859101-43859101 on Build GRCh38
rs750108276 Chr.13: 43858908-43858910 on Build GRCh38
rs374286286 Chr.13: 43858783-43858783 on Build GRCh38
rs764150094 Chr.13: 43858786-43858790 on Build GRCh38
rs778168505 Chr.13: 43858818-43858818 on Build GRCh38
rs764929517 Chr.13: 43858837-43858837 on Build GRCh38
rs887221260 Chr.13: 43858720-43858720 on Build GRCh38
rs961643284 Chr.13: 43859042-43859042 on Build GRCh38
rs753348617 Chr.13: 43858812-43858812 on Build GRCh38
rs553318500 Chr.13: 43858781-43858781 on Build GRCh38
rs751326970 Chr.13: 43858765-43858765 on Build GRCh38
rs764794974 Chr.13: 43858742-43858742 on Build GRCh38
rs996736722 Chr.13: 43859104-43859104 on Build GRCh38
rs1004337217 Chr.13: 43859076-43859076 on Build GRCh38
rs755508859 Chr.13: 43858760-43858760 on Build GRCh38
rs887484492 Chr.13: 43858903-43858903 on Build GRCh38
rs764043222 Chr.13: 43858864-43858864 on Build GRCh38
rs996362189 Chr.13: 43858879-43858879 on Build GRCh38
rs369885255 Chr.13: 43858755-43858755 on Build GRCh38
rs113494820 Chr.13: 43858846-43858846 on Build GRCh38
rs1019657986 Chr.13: 43858729-43858729 on Build GRCh38
rs762400183 Chr.13: 43858863-43858863 on Build GRCh38
rs1019587473 Chr.13: 43859077-43859077 on Build GRCh38
rs767407128 Chr.13: 43858891-43858891 on Build GRCh38
rs1000429943 Chr.13: 43859143-43859143 on Build GRCh38
rs968694367 Chr.13: 43859139-43859139 on Build GRCh38
rs377732767 Chr.13: 43858809-43858809 on Build GRCh38
rs529686937 Chr.13: 43858766-43858766 on Build GRCh38
rs1045305160 Chr.13: 43858988-43858988 on Build GRCh38
rs370161144 Chr.13: 43858845-43858845 on Build GRCh38

View Full Product Details