Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43868564-43869092 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC122
Entrez Gene ID
160857
Gene Name
coiled-coil domain containing 122
Gene Aliases
-
UniGene
Hs.170849
Gene Chromosome Location
Chr.13: 43821959-43879690 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020398.1 XM_017020399.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947603 c.111A>C p.Q37H Chr.13: 43868739-43868739 on Build GRCh38
1238439 c.124G>A p.E42K Chr.13: 43868726-43868726 on Build GRCh38
6315681 c.52C>A p.Q18K Chr.13: 43868798-43868798 on Build GRCh38
6315680 c.52C>A p.Q18K Chr.13: 43868798-43868798 on Build GRCh38
4865504 c.111A>C p.Q37H Chr.13: 43868739-43868739 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054209109 Chr.13: 43868882-43868882 on Build GRCh38
rs760832855 Chr.13: 43868751-43868751 on Build GRCh38
rs746530568 Chr.13: 43868703-43868703 on Build GRCh38
rs973892249 Chr.13: 43868683-43868683 on Build GRCh38
rs750981629 Chr.13: 43868811-43868811 on Build GRCh38
rs993769318 Chr.13: 43868883-43868883 on Build GRCh38
rs1017233199 Chr.13: 43868915-43868915 on Build GRCh38
rs890684952 Chr.13: 43868617-43868617 on Build GRCh38
rs914832206 Chr.13: 43868942-43868942 on Build GRCh38
rs1018173831 Chr.13: 43868647-43868647 on Build GRCh38
rs573520690 Chr.13: 43868879-43868879 on Build GRCh38
rs540491832 Chr.13: 43869021-43869021 on Build GRCh38
rs4942252 Chr.13: 43868903-43868903 on Build GRCh38
rs1017808599 Chr.13: 43869005-43869005 on Build GRCh38
rs539987713 Chr.13: 43868842-43868842 on Build GRCh38
rs1024246670 Chr.13: 43869062-43869062 on Build GRCh38
rs1030516440 Chr.13: 43868590-43868590 on Build GRCh38
rs561884859 Chr.13: 43868859-43868859 on Build GRCh38
rs990381672 Chr.13: 43868930-43868930 on Build GRCh38
rs954392023 Chr.13: 43868602-43868602 on Build GRCh38
rs983439378 Chr.13: 43869048-43869048 on Build GRCh38
rs769533052 Chr.13: 43868752-43868752 on Build GRCh38
rs945709211 Chr.13: 43868905-43868905 on Build GRCh38
rs946227385 Chr.13: 43868945-43868945 on Build GRCh38
rs964235537 Chr.13: 43868923-43868923 on Build GRCh38
rs79697600 Chr.13: 43868738-43868738 on Build GRCh38
rs370703884 Chr.13: 43868788-43868788 on Build GRCh38
rs950726984 Chr.13: 43868713-43868713 on Build GRCh38
rs749137476 Chr.13: 43868867-43868868 on Build GRCh38
rs1009552877 Chr.13: 43869001-43869001 on Build GRCh38
rs950778693 Chr.13: 43869030-43869030 on Build GRCh38
rs935805840 Chr.13: 43868849-43868849 on Build GRCh38
rs979989232 Chr.13: 43868825-43868825 on Build GRCh38
rs556900131 Chr.13: 43868615-43868615 on Build GRCh38
rs780604886 Chr.13: 43868832-43868832 on Build GRCh38
rs900668935 Chr.13: 43869020-43869020 on Build GRCh38
rs201257755 Chr.13: 43868712-43868712 on Build GRCh38
rs753128961 Chr.13: 43868800-43868800 on Build GRCh38
rs368020834 Chr.13: 43868733-43868733 on Build GRCh38
rs78953797 Chr.13: 43868758-43868758 on Build GRCh38
rs769545650 Chr.13: 43868725-43868725 on Build GRCh38
rs1009701996 Chr.13: 43868795-43868795 on Build GRCh38
rs765686693 Chr.13: 43868789-43868789 on Build GRCh38
rs551733594 Chr.13: 43869028-43869028 on Build GRCh38
rs985795538 Chr.13: 43868605-43868605 on Build GRCh38
rs374803113 Chr.13: 43868728-43868728 on Build GRCh38
rs756766956 Chr.13: 43868826-43868826 on Build GRCh38
rs942031882 Chr.13: 43869055-43869055 on Build GRCh38
rs763834802 Chr.13: 43868810-43868810 on Build GRCh38
rs758187366 Chr.13: 43868682-43868682 on Build GRCh38
rs562449899 Chr.13: 43868988-43868988 on Build GRCh38
rs773126226 Chr.13: 43868745-43868745 on Build GRCh38
rs1009088751 Chr.13: 43868619-43868619 on Build GRCh38
rs745839539 Chr.13: 43868716-43868716 on Build GRCh38
rs777167536 Chr.13: 43868778-43868778 on Build GRCh38
rs752612786 Chr.13: 43868968-43868968 on Build GRCh38
rs1027708113 Chr.13: 43869024-43869024 on Build GRCh38
rs202087360 Chr.13: 43868828-43868828 on Build GRCh38
rs752397020 Chr.13: 43868680-43868680 on Build GRCh38
rs758052892 Chr.13: 43868807-43868807 on Build GRCh38
rs769043880 Chr.13: 43869004-43869004 on Build GRCh38
rs533105442 Chr.13: 43869026-43869026 on Build GRCh38
rs200310684 Chr.13: 43868766-43868766 on Build GRCh38
rs371242790 Chr.13: 43868721-43868721 on Build GRCh38
rs964855966 Chr.13: 43868660-43868660 on Build GRCh38
rs1006153810 Chr.13: 43868865-43868865 on Build GRCh38
rs756100870 Chr.13: 43868841-43868841 on Build GRCh38
rs369819010 Chr.13: 43868691-43868691 on Build GRCh38
rs777085087 Chr.13: 43868699-43868699 on Build GRCh38
rs1030075330 Chr.13: 43868900-43868900 on Build GRCh38
rs927112321 Chr.13: 43868848-43868848 on Build GRCh38
rs907329975 Chr.13: 43868907-43868907 on Build GRCh38
rs890740250 Chr.13: 43868986-43868986 on Build GRCh38
rs894360697 Chr.13: 43868853-43868853 on Build GRCh38

View Full Product Details