Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 43879182-43879721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC122
Entrez Gene ID
160857
Gene Name
coiled-coil domain containing 122
Gene Aliases
-
UniGene
Hs.170849
Gene Chromosome Location
Chr.13: 43821959-43879690 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020398.1 XM_017020399.1 NM_001128303.1 XM_005266261.3 XM_011534933.2

Gene Symbol
LACC1
Entrez Gene ID
144811
Gene Name
laccase domain containing 1
Gene Aliases
C13orf31
UniGene
Hs.210586
Gene Chromosome Location
Chr.13: 43878834-43893932 on Build GRCh38
Transcripts
NM_144974.3 XM_017020397.1 XM_017020398.1 XM_017020399.1 NM_001128303.1 XM_005266261.3 XM_011534933.2

SNP Detail

SNP ID Chromosome Location
rs1045215103 Chr.13: 43879225-43879225 on Build GRCh38
rs1024453252 Chr.13: 43879448-43879448 on Build GRCh38
rs917816149 Chr.13: 43879691-43879691 on Build GRCh38
rs950134022 Chr.13: 43879652-43879652 on Build GRCh38
rs541205817 Chr.13: 43879411-43879411 on Build GRCh38
rs940524242 Chr.13: 43879552-43879552 on Build GRCh38
rs957652381 Chr.13: 43879484-43879484 on Build GRCh38
rs12868462 Chr.13: 43879344-43879344 on Build GRCh38
rs997074490 Chr.13: 43879700-43879700 on Build GRCh38
rs994834766 Chr.13: 43879349-43879349 on Build GRCh38
rs1005516915 Chr.13: 43879326-43879326 on Build GRCh38
rs921678690 Chr.13: 43879246-43879246 on Build GRCh38
rs1037019413 Chr.13: 43879557-43879557 on Build GRCh38
rs953470115 Chr.13: 43879427-43879427 on Build GRCh38
rs904987053 Chr.13: 43879252-43879252 on Build GRCh38
rs979408081 Chr.13: 43879443-43879443 on Build GRCh38
rs987641571 Chr.13: 43879545-43879545 on Build GRCh38
rs3816311 Chr.13: 43879647-43879647 on Build GRCh38
rs537103164 Chr.13: 43879212-43879212 on Build GRCh38
rs953110196 Chr.13: 43879537-43879537 on Build GRCh38
rs560678652 Chr.13: 43879689-43879689 on Build GRCh38
rs1055835385 Chr.13: 43879362-43879362 on Build GRCh38
rs976315832 Chr.13: 43879405-43879405 on Build GRCh38
rs999425691 Chr.13: 43879672-43879672 on Build GRCh38
rs989093306 Chr.13: 43879487-43879487 on Build GRCh38
rs971963382 Chr.13: 43879563-43879563 on Build GRCh38
rs902125794 Chr.13: 43879579-43879579 on Build GRCh38
rs1036699175 Chr.13: 43879329-43879329 on Build GRCh38
rs1014776500 Chr.13: 43879446-43879446 on Build GRCh38
rs867687625 Chr.13: 43879431-43879431 on Build GRCh38
rs949253761 Chr.13: 43879223-43879223 on Build GRCh38
rs111606059 Chr.13: 43879406-43879406 on Build GRCh38
rs967962030 Chr.13: 43879472-43879472 on Build GRCh38
rs989150530 Chr.13: 43879201-43879201 on Build GRCh38
rs752449833 Chr.13: 43879482-43879482 on Build GRCh38
rs762617600 Chr.13: 43879626-43879626 on Build GRCh38
rs559018571 Chr.13: 43879324-43879324 on Build GRCh38
rs1052379511 Chr.13: 43879204-43879204 on Build GRCh38
rs574758397 Chr.13: 43879551-43879551 on Build GRCh38
rs1026632643 Chr.13: 43879304-43879304 on Build GRCh38
rs920500490 Chr.13: 43879590-43879590 on Build GRCh38
rs940591891 Chr.13: 43879325-43879325 on Build GRCh38
rs999082705 Chr.13: 43879481-43879481 on Build GRCh38
rs896277076 Chr.13: 43879206-43879206 on Build GRCh38
rs935330939 Chr.13: 43879467-43879467 on Build GRCh38
rs931042958 Chr.13: 43879532-43879532 on Build GRCh38
rs904934637 Chr.13: 43879491-43879491 on Build GRCh38
rs1051588008 Chr.13: 43879294-43879294 on Build GRCh38
rs1029483615 Chr.13: 43879412-43879412 on Build GRCh38
rs890552410 Chr.13: 43879207-43879207 on Build GRCh38
rs1046238975 Chr.13: 43879659-43879659 on Build GRCh38
rs911695992 Chr.13: 43879548-43879548 on Build GRCh38
rs921627652 Chr.13: 43879533-43879533 on Build GRCh38
rs1033170020 Chr.13: 43879674-43879674 on Build GRCh38
rs553186419 Chr.13: 43879417-43879417 on Build GRCh38
rs886741544 Chr.13: 43879539-43879539 on Build GRCh38
rs998079228 Chr.13: 43879580-43879580 on Build GRCh38
rs903311407 Chr.13: 43879668-43879668 on Build GRCh38
rs931762876 Chr.13: 43879253-43879253 on Build GRCh38
rs930573920 Chr.13: 43879597-43879597 on Build GRCh38
rs893372258 Chr.13: 43879695-43879695 on Build GRCh38

View Full Product Details