Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49331861-49332358 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAB39L
Entrez Gene ID
81617
Gene Name
calcium binding protein 39 like
Gene Aliases
MO25-BETA, MO2L, bA103J18.3
UniGene
Hs.742971
Gene Chromosome Location
Chr.13: 49308628-49444346 on Build GRCh38
Transcripts
NM_001079670.2 NM_001287337.1 NM_001287338.1 NM_001287339.1 NM_030925.3 XM_011535254.2 XM_011535255.2 XM_011535256.2 XM_017020785.1 XM_017020786.1 XM_017020787.1 XM_017020788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4047960 c.807C>T p.I269I Chr.13: 49331974-49331974 on Build GRCh38
3957245 c.806T>A p.I269N Chr.13: 49331975-49331975 on Build GRCh38
6042751 c.805A>G p.I269V Chr.13: 49331976-49331976 on Build GRCh38
6617160 c.759G>T p.E253D Chr.13: 49332022-49332022 on Build GRCh38
6617158 c.834+7delA p.? Chr.13: 49331940-49331940 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766940665 Chr.13: 49331900-49331901 on Build GRCh38
rs1046843959 Chr.13: 49332265-49332265 on Build GRCh38
rs766379793 Chr.13: 49331937-49331937 on Build GRCh38
rs758847465 Chr.13: 49332072-49332072 on Build GRCh38
rs752051091 Chr.13: 49332138-49332139 on Build GRCh38
rs1017885417 Chr.13: 49332110-49332110 on Build GRCh38
rs745843719 Chr.13: 49332026-49332026 on Build GRCh38
rs188632766 Chr.13: 49332325-49332325 on Build GRCh38
rs997810492 Chr.13: 49332161-49332161 on Build GRCh38
rs963968156 Chr.13: 49332148-49332148 on Build GRCh38
rs537156196 Chr.13: 49332227-49332227 on Build GRCh38
rs769331386 Chr.13: 49332096-49332096 on Build GRCh38
rs937569873 Chr.13: 49332258-49332258 on Build GRCh38
rs139674284 Chr.13: 49332034-49332034 on Build GRCh38
rs77990369 Chr.13: 49331885-49331885 on Build GRCh38
rs113858270 Chr.13: 49332213-49332214 on Build GRCh38
rs937607982 Chr.13: 49331997-49331997 on Build GRCh38
rs566058470 Chr.13: 49332115-49332115 on Build GRCh38
rs780878836 Chr.13: 49332089-49332089 on Build GRCh38
rs748457475 Chr.13: 49331894-49331894 on Build GRCh38
rs758831530 Chr.13: 49331993-49331993 on Build GRCh38
rs773423921 Chr.13: 49332045-49332045 on Build GRCh38
rs149844947 Chr.13: 49332065-49332065 on Build GRCh38
rs866160457 Chr.13: 49332292-49332292 on Build GRCh38
rs750627307 Chr.13: 49331915-49331915 on Build GRCh38
rs779866530 Chr.13: 49332097-49332097 on Build GRCh38
rs907945620 Chr.13: 49332188-49332188 on Build GRCh38
rs772809655 Chr.13: 49332183-49332186 on Build GRCh38
rs771760756 Chr.13: 49331991-49331991 on Build GRCh38
rs755389893 Chr.13: 49331955-49331955 on Build GRCh38
rs554687697 Chr.13: 49332153-49332153 on Build GRCh38
rs895183260 Chr.13: 49332012-49332012 on Build GRCh38
rs930072601 Chr.13: 49332290-49332290 on Build GRCh38
rs975740747 Chr.13: 49332151-49332151 on Build GRCh38
rs191491092 Chr.13: 49332264-49332264 on Build GRCh38
rs769671617 Chr.13: 49332027-49332027 on Build GRCh38
rs955187611 Chr.13: 49332154-49332154 on Build GRCh38
rs773433800 Chr.13: 49331911-49331911 on Build GRCh38
rs761571626 Chr.13: 49332122-49332122 on Build GRCh38
rs977269875 Chr.13: 49332185-49332185 on Build GRCh38
rs372617834 Chr.13: 49331994-49331994 on Build GRCh38
rs957266166 Chr.13: 49332216-49332216 on Build GRCh38
rs748504090 Chr.13: 49331985-49331985 on Build GRCh38
rs1053785170 Chr.13: 49331990-49331990 on Build GRCh38
rs779450983 Chr.13: 49331957-49331957 on Build GRCh38
rs201100340 Chr.13: 49332078-49332078 on Build GRCh38
rs760846081 Chr.13: 49332051-49332051 on Build GRCh38
rs745433220 Chr.13: 49332091-49332091 on Build GRCh38
rs749152680 Chr.13: 49332108-49332108 on Build GRCh38
rs559275027 Chr.13: 49332241-49332241 on Build GRCh38
rs536614293 Chr.13: 49332128-49332128 on Build GRCh38
rs774081978 Chr.13: 49332118-49332118 on Build GRCh38
rs368594431 Chr.13: 49332217-49332217 on Build GRCh38
rs762712310 Chr.13: 49332041-49332041 on Build GRCh38
rs74074569 Chr.13: 49332318-49332318 on Build GRCh38
rs183754650 Chr.13: 49332287-49332287 on Build GRCh38
rs75421260 Chr.13: 49332069-49332069 on Build GRCh38
rs770573828 Chr.13: 49332010-49332010 on Build GRCh38
rs748404099 Chr.13: 49331932-49331932 on Build GRCh38
rs775967382 Chr.13: 49332025-49332025 on Build GRCh38
rs116043436 Chr.13: 49332180-49332180 on Build GRCh38
rs530543348 Chr.13: 49332058-49332058 on Build GRCh38
rs940663675 Chr.13: 49331921-49331921 on Build GRCh38
rs776909962 Chr.13: 49332141-49332141 on Build GRCh38
rs186705879 Chr.13: 49332042-49332042 on Build GRCh38
rs933384219 Chr.13: 49331886-49331886 on Build GRCh38
rs576671158 Chr.13: 49332254-49332254 on Build GRCh38
rs754399182 Chr.13: 49331948-49331948 on Build GRCh38
rs539332301 Chr.13: 49332076-49332076 on Build GRCh38
rs770295695 Chr.13: 49331904-49331904 on Build GRCh38
rs199498743 Chr.13: 49332066-49332066 on Build GRCh38
rs559259463 Chr.13: 49332282-49332282 on Build GRCh38
rs746603549 Chr.13: 49332009-49332009 on Build GRCh38
rs756924758 Chr.13: 49332085-49332085 on Build GRCh38
rs368994626 Chr.13: 49331935-49331935 on Build GRCh38

View Full Product Details