Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 49660988-49661497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EBPL
Entrez Gene ID
84650
Gene Name
emopamil binding protein like
Gene Aliases
EBRP
UniGene
Hs.433278
Gene Chromosome Location
Chr.13: 49660676-49691487 on Build GRCh38
Transcripts
NM_001278636.1 NM_032565.4 NR_103802.1 NR_103803.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
947900 c.587A>C p.Q196P Chr.13: 49661002-49661002 on Build GRCh38
3885406 c.537C>T p.I179I Chr.13: 49661052-49661052 on Build GRCh38
1322855 c.528G>A p.W176* Chr.13: 49661061-49661061 on Build GRCh38
4391448 c.589A>G p.K197E Chr.13: 49661000-49661000 on Build GRCh38
1367385 c.516_517insT p.N173fs*1 Chr.13: 49661072-49661073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572015420 Chr.13: 49661363-49661363 on Build GRCh38
rs1042680789 Chr.13: 49661304-49661304 on Build GRCh38
rs768948494 Chr.13: 49661121-49661121 on Build GRCh38
rs200901347 Chr.13: 49661069-49661069 on Build GRCh38
rs1009141462 Chr.13: 49661354-49661354 on Build GRCh38
rs555716863 Chr.13: 49661391-49661391 on Build GRCh38
rs760934732 Chr.13: 49661072-49661073 on Build GRCh38
rs185971851 Chr.13: 49661070-49661070 on Build GRCh38
rs757958154 Chr.13: 49661183-49661183 on Build GRCh38
rs751630731 Chr.13: 49661022-49661022 on Build GRCh38
rs750911543 Chr.13: 49661037-49661037 on Build GRCh38
rs11840284 Chr.13: 49661244-49661244 on Build GRCh38
rs758261182 Chr.13: 49661177-49661182 on Build GRCh38
rs773277059 Chr.13: 49661243-49661243 on Build GRCh38
rs527503332 Chr.13: 49661051-49661051 on Build GRCh38
rs1029257207 Chr.13: 49661060-49661060 on Build GRCh38
rs987679242 Chr.13: 49661091-49661091 on Build GRCh38
rs776417561 Chr.13: 49661094-49661094 on Build GRCh38
rs754650479 Chr.13: 49661177-49661177 on Build GRCh38
rs764834168 Chr.13: 49661178-49661178 on Build GRCh38
rs1028996626 Chr.13: 49661320-49661320 on Build GRCh38
rs769680385 Chr.13: 49661211-49661211 on Build GRCh38
rs191584297 Chr.13: 49661284-49661284 on Build GRCh38
rs902650937 Chr.13: 49661308-49661308 on Build GRCh38
rs940465419 Chr.13: 49661129-49661129 on Build GRCh38
rs923320192 Chr.13: 49661279-49661279 on Build GRCh38
rs767684833 Chr.13: 49661128-49661128 on Build GRCh38
rs753117191 Chr.13: 49661254-49661254 on Build GRCh38
rs754176340 Chr.13: 49661165-49661165 on Build GRCh38
rs762022914 Chr.13: 49661125-49661125 on Build GRCh38
rs369293935 Chr.13: 49661081-49661082 on Build GRCh38
rs370834479 Chr.13: 49661118-49661118 on Build GRCh38
rs576788202 Chr.13: 49661407-49661407 on Build GRCh38
rs747612403 Chr.13: 49661063-49661063 on Build GRCh38
rs780870144 Chr.13: 49661199-49661199 on Build GRCh38
rs772111930 Chr.13: 49661239-49661239 on Build GRCh38
rs770695528 Chr.13: 49661076-49661076 on Build GRCh38
rs765134934 Chr.13: 49661073-49661073 on Build GRCh38
rs752898216 Chr.13: 49661070-49661071 on Build GRCh38
rs920358352 Chr.13: 49661408-49661408 on Build GRCh38
rs975136736 Chr.13: 49661264-49661265 on Build GRCh38
rs954903886 Chr.13: 49661074-49661074 on Build GRCh38
rs991148780 Chr.13: 49661448-49661448 on Build GRCh38
rs567442382 Chr.13: 49661103-49661103 on Build GRCh38
rs936827271 Chr.13: 49661319-49661319 on Build GRCh38
rs766658976 Chr.13: 49661154-49661154 on Build GRCh38
rs746947774 Chr.13: 49661075-49661075 on Build GRCh38
rs762431643 Chr.13: 49661024-49661024 on Build GRCh38
rs777329394 Chr.13: 49661184-49661184 on Build GRCh38
rs752318902 Chr.13: 49661181-49661181 on Build GRCh38
rs746922050 Chr.13: 49661193-49661193 on Build GRCh38
rs753503039 Chr.13: 49661045-49661045 on Build GRCh38
rs996182200 Chr.13: 49661312-49661312 on Build GRCh38
rs750949510 Chr.13: 49661133-49661133 on Build GRCh38
rs536322531 Chr.13: 49661369-49661369 on Build GRCh38
rs544508082 Chr.13: 49661416-49661416 on Build GRCh38
rs958695139 Chr.13: 49661446-49661446 on Build GRCh38
rs763229610 Chr.13: 49661018-49661018 on Build GRCh38
rs768096497 Chr.13: 49661028-49661028 on Build GRCh38
rs762821460 Chr.13: 49661265-49661265 on Build GRCh38
rs764272651 Chr.13: 49661021-49661021 on Build GRCh38
rs937028777 Chr.13: 49661170-49661170 on Build GRCh38
rs757159795 Chr.13: 49661196-49661196 on Build GRCh38
rs750172538 Chr.13: 49661109-49661150 on Build GRCh38
rs780455715 Chr.13: 49661041-49661041 on Build GRCh38
rs774763562 Chr.13: 49661230-49661230 on Build GRCh38
rs550099444 Chr.13: 49661252-49661252 on Build GRCh38
rs1015126465 Chr.13: 49661390-49661390 on Build GRCh38
rs768864195 Chr.13: 49661122-49661122 on Build GRCh38
rs562802393 Chr.13: 49661442-49661442 on Build GRCh38
rs61747169 Chr.13: 49661050-49661050 on Build GRCh38
rs766389499 Chr.13: 49661352-49661352 on Build GRCh38
rs1053888067 Chr.13: 49661292-49661292 on Build GRCh38
rs984800611 Chr.13: 49661200-49661200 on Build GRCh38
rs760924603 Chr.13: 49661082-49661082 on Build GRCh38
rs893850163 Chr.13: 49661293-49661293 on Build GRCh38
rs139407673 Chr.13: 49661263-49661263 on Build GRCh38
rs374216796 Chr.13: 49661150-49661150 on Build GRCh38
rs553845704 Chr.13: 49661350-49661350 on Build GRCh38
rs745617704 Chr.13: 49661208-49661208 on Build GRCh38
rs954977255 Chr.13: 49661326-49661326 on Build GRCh38

View Full Product Details