Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51760157-51760661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS12
Entrez Gene ID
79758
Gene Name
dehydrogenase/reductase 12
Gene Aliases
SDR40C1
UniGene
Hs.266728
Gene Chromosome Location
Chr.13: 51754856-51804206 on Build GRCh38
Transcripts
XM_017020750.1 XM_017020751.1 NM_052950.3 XM_011534914.1 XM_011534915.2 XM_017020386.1

Gene Symbol
WDFY2
Entrez Gene ID
115825
Gene Name
WD repeat and FYVE domain containing 2
Gene Aliases
PROF, WDF2, ZFYVE22
UniGene
Hs.208550
Gene Chromosome Location
Chr.13: 51584194-51766799 on Build GRCh38
Transcripts
XM_017020750.1 XM_017020751.1 NM_052950.3 XM_011534914.1 XM_011534915.2 XM_017020386.1

SNP Detail

SNP ID Chromosome Location
rs980375811 Chr.13: 51760273-51760273 on Build GRCh38
rs919451423 Chr.13: 51760364-51760364 on Build GRCh38
rs973169055 Chr.13: 51760416-51760417 on Build GRCh38
rs547048264 Chr.13: 51760509-51760509 on Build GRCh38
rs527292741 Chr.13: 51760505-51760505 on Build GRCh38
rs904520676 Chr.13: 51760404-51760404 on Build GRCh38
rs374038026 Chr.13: 51760272-51760272 on Build GRCh38
rs973568074 Chr.13: 51760566-51760566 on Build GRCh38
rs542389096 Chr.13: 51760330-51760330 on Build GRCh38
rs1048903018 Chr.13: 51760454-51760454 on Build GRCh38
rs757716225 Chr.13: 51760189-51760189 on Build GRCh38
rs1007500703 Chr.13: 51760183-51760183 on Build GRCh38
rs963478955 Chr.13: 51760311-51760311 on Build GRCh38
rs1027418819 Chr.13: 51760627-51760627 on Build GRCh38
rs1053176415 Chr.13: 51760479-51760479 on Build GRCh38
rs1009331373 Chr.13: 51760485-51760485 on Build GRCh38
rs536390194 Chr.13: 51760635-51760636 on Build GRCh38
rs1040024860 Chr.13: 51760188-51760188 on Build GRCh38
rs12856726 Chr.13: 51760483-51760483 on Build GRCh38
rs539696174 Chr.13: 51760254-51760254 on Build GRCh38
rs1039080081 Chr.13: 51760400-51760400 on Build GRCh38
rs920011786 Chr.13: 51760417-51760417 on Build GRCh38
rs530598272 Chr.13: 51760616-51760616 on Build GRCh38
rs188142837 Chr.13: 51760533-51760533 on Build GRCh38
rs963067335 Chr.13: 51760542-51760542 on Build GRCh38
rs544502500 Chr.13: 51760474-51760474 on Build GRCh38
rs903577366 Chr.13: 51760565-51760565 on Build GRCh38
rs1033825317 Chr.13: 51760373-51760373 on Build GRCh38
rs879545151 Chr.13: 51760453-51760454 on Build GRCh38
rs973172884 Chr.13: 51760357-51760357 on Build GRCh38
rs778317890 Chr.13: 51760291-51760291 on Build GRCh38
rs889008406 Chr.13: 51760182-51760182 on Build GRCh38
rs970385195 Chr.13: 51760194-51760194 on Build GRCh38
rs555900217 Chr.13: 51760387-51760387 on Build GRCh38
rs535716126 Chr.13: 51760623-51760623 on Build GRCh38
rs975014870 Chr.13: 51760620-51760620 on Build GRCh38
rs533491739 Chr.13: 51760532-51760532 on Build GRCh38
rs9596568 Chr.13: 51760275-51760275 on Build GRCh38
rs575659073 Chr.13: 51760389-51760389 on Build GRCh38
rs746865331 Chr.13: 51760561-51760561 on Build GRCh38
rs528081679 Chr.13: 51760599-51760599 on Build GRCh38
rs959522599 Chr.13: 51760595-51760595 on Build GRCh38
rs193132669 Chr.13: 51760600-51760600 on Build GRCh38
rs912121270 Chr.13: 51760393-51760393 on Build GRCh38
rs964998962 Chr.13: 51760617-51760617 on Build GRCh38
rs79593484 Chr.13: 51760464-51760464 on Build GRCh38
rs943591202 Chr.13: 51760396-51760396 on Build GRCh38
rs1026041323 Chr.13: 51760582-51760582 on Build GRCh38
rs35118122 Chr.13: 51760396-51760397 on Build GRCh38
rs1001012306 Chr.13: 51760335-51760335 on Build GRCh38
rs931402691 Chr.13: 51760430-51760430 on Build GRCh38
rs1033800381 Chr.13: 51760540-51760540 on Build GRCh38
rs1040311185 Chr.13: 51760492-51760492 on Build GRCh38
rs918201295 Chr.13: 51760394-51760394 on Build GRCh38
rs112591719 Chr.13: 51760447-51760447 on Build GRCh38
rs567827859 Chr.13: 51760604-51760604 on Build GRCh38

View Full Product Details