Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51771014-51771553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHRS12
Entrez Gene ID
79758
Gene Name
dehydrogenase/reductase 12
Gene Aliases
SDR40C1
UniGene
Hs.266728
Gene Chromosome Location
Chr.13: 51754856-51804206 on Build GRCh38
Transcripts
NM_001031719.2 NM_001270424.1 NM_024705.2 XM_005266527.3 XM_011535235.2 XM_011535236.2 XM_011535237.1 XM_011535238.1 XM_017020750.1 XM_017020751.1 XM_017020753.1 XM_017020754.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5362091 c.678G>A p.S226S Chr.13: 51771377-51771377 on Build GRCh38
6871437 c.657C>T p.C219C Chr.13: 51771398-51771398 on Build GRCh38
6236633 c.693G>A p.A231A Chr.13: 51771362-51771362 on Build GRCh38
144097 c.593G>A p.G198E Chr.13: 51771462-51771462 on Build GRCh38
1477279 c.768C>T p.L256L Chr.13: 51771287-51771287 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747500421 Chr.13: 51771511-51771511 on Build GRCh38
rs1043962694 Chr.13: 51771159-51771159 on Build GRCh38
rs756911088 Chr.13: 51771124-51771124 on Build GRCh38
rs747491357 Chr.13: 51771443-51771443 on Build GRCh38
rs759286740 Chr.13: 51771467-51771467 on Build GRCh38
rs767897725 Chr.13: 51771235-51771235 on Build GRCh38
rs576935051 Chr.13: 51771177-51771177 on Build GRCh38
rs746973880 Chr.13: 51771303-51771303 on Build GRCh38
rs140377210 Chr.13: 51771378-51771378 on Build GRCh38
rs144695820 Chr.13: 51771260-51771260 on Build GRCh38
rs1001868981 Chr.13: 51771200-51771200 on Build GRCh38
rs1025725894 Chr.13: 51771243-51771243 on Build GRCh38
rs142737331 Chr.13: 51771417-51771417 on Build GRCh38
rs775272903 Chr.13: 51771530-51771530 on Build GRCh38
rs757736656 Chr.13: 51771521-51771521 on Build GRCh38
rs571257952 Chr.13: 51771158-51771158 on Build GRCh38
rs748675271 Chr.13: 51771332-51771332 on Build GRCh38
rs995025696 Chr.13: 51771220-51771220 on Build GRCh38
rs373088287 Chr.13: 51771195-51771195 on Build GRCh38
rs115939467 Chr.13: 51771216-51771216 on Build GRCh38
rs769549582 Chr.13: 51771468-51771468 on Build GRCh38
rs758952161 Chr.13: 51771271-51771271 on Build GRCh38
rs368614345 Chr.13: 51771494-51771494 on Build GRCh38
rs748783226 Chr.13: 51771097-51771097 on Build GRCh38
rs770682735 Chr.13: 51771508-51771509 on Build GRCh38
rs756990621 Chr.13: 51771384-51771384 on Build GRCh38
rs895514966 Chr.13: 51771204-51771204 on Build GRCh38
rs760938494 Chr.13: 51771356-51771356 on Build GRCh38
rs1045144634 Chr.13: 51771354-51771354 on Build GRCh38
rs764873546 Chr.13: 51771363-51771363 on Build GRCh38
rs953346092 Chr.13: 51771108-51771108 on Build GRCh38
rs775358557 Chr.13: 51771250-51771250 on Build GRCh38
rs993885375 Chr.13: 51771238-51771238 on Build GRCh38
rs536081671 Chr.13: 51771196-51771196 on Build GRCh38
rs760151168 Chr.13: 51771417-51771419 on Build GRCh38
rs530187843 Chr.13: 51771471-51771471 on Build GRCh38
rs369787711 Chr.13: 51771262-51771262 on Build GRCh38
rs780983539 Chr.13: 51771399-51771399 on Build GRCh38
rs1013995893 Chr.13: 51771211-51771211 on Build GRCh38
rs147409219 Chr.13: 51771440-51771440 on Build GRCh38
rs763054031 Chr.13: 51771391-51771391 on Build GRCh38
rs375721036 Chr.13: 51771492-51771492 on Build GRCh38
rs750194424 Chr.13: 51771484-51771484 on Build GRCh38
rs368509464 Chr.13: 51771408-51771408 on Build GRCh38
rs776157636 Chr.13: 51771360-51771360 on Build GRCh38
rs570390316 Chr.13: 51771049-51771049 on Build GRCh38
rs373329685 Chr.13: 51771298-51771298 on Build GRCh38
rs1036155608 Chr.13: 51771213-51771213 on Build GRCh38
rs866666686 Chr.13: 51771325-51771325 on Build GRCh38
rs761745872 Chr.13: 51771481-51771481 on Build GRCh38
rs752208846 Chr.13: 51771366-51771366 on Build GRCh38
rs898445043 Chr.13: 51771485-51771485 on Build GRCh38
rs74086276 Chr.13: 51771500-51771500 on Build GRCh38
rs561246895 Chr.13: 51771412-51771412 on Build GRCh38
rs994458968 Chr.13: 51771489-51771489 on Build GRCh38
rs541376533 Chr.13: 51771402-51771402 on Build GRCh38
rs777012313 Chr.13: 51771464-51771464 on Build GRCh38
rs746205071 Chr.13: 51771524-51771524 on Build GRCh38
rs144077287 Chr.13: 51771194-51771194 on Build GRCh38
rs867234864 Chr.13: 51771435-51771435 on Build GRCh38
rs532985197 Chr.13: 51771323-51771323 on Build GRCh38
rs911954556 Chr.13: 51771138-51771138 on Build GRCh38
rs753450704 Chr.13: 51771270-51771270 on Build GRCh38
rs77682467 Chr.13: 51771324-51771324 on Build GRCh38
rs764154476 Chr.13: 51771480-51771480 on Build GRCh38
rs1028283204 Chr.13: 51771226-51771228 on Build GRCh38
rs955120980 Chr.13: 51771086-51771086 on Build GRCh38
rs918365351 Chr.13: 51771060-51771060 on Build GRCh38
rs1055746850 Chr.13: 51771426-51771426 on Build GRCh38
rs371437952 Chr.13: 51771436-51771436 on Build GRCh38
rs755222453 Chr.13: 51771225-51771226 on Build GRCh38
rs553594163 Chr.13: 51771189-51771189 on Build GRCh38
rs955134187 Chr.13: 51771255-51771255 on Build GRCh38
rs770671299 Chr.13: 51771358-51771358 on Build GRCh38
rs925582332 Chr.13: 51771305-51771305 on Build GRCh38
rs1015496022 Chr.13: 51771056-51771056 on Build GRCh38
rs542705422 Chr.13: 51771362-51771362 on Build GRCh38
rs532899610 Chr.13: 51771522-51771522 on Build GRCh38
rs745713743 Chr.13: 51771198-51771198 on Build GRCh38
rs1045593328 Chr.13: 51771197-51771197 on Build GRCh38
rs925289129 Chr.13: 51771166-51771166 on Build GRCh38
rs995391349 Chr.13: 51771495-51771495 on Build GRCh38
rs773398594 Chr.13: 51771242-51771242 on Build GRCh38
rs74086275 Chr.13: 51771091-51771091 on Build GRCh38
rs988214197 Chr.13: 51771175-51771175 on Build GRCh38
rs771333568 Chr.13: 51771247-51771247 on Build GRCh38
rs370679809 Chr.13: 51771380-51771380 on Build GRCh38
rs760891506 Chr.13: 51771228-51771228 on Build GRCh38
rs778899902 Chr.13: 51771328-51771328 on Build GRCh38
rs756404043 Chr.13: 51771310-51771310 on Build GRCh38
rs751294498 Chr.13: 51771304-51771304 on Build GRCh38
rs551436722 Chr.13: 51771126-51771126 on Build GRCh38
rs752683299 Chr.13: 51771294-51771294 on Build GRCh38
rs771202456 Chr.13: 51771451-51771451 on Build GRCh38
rs1001396531 Chr.13: 51771421-51771421 on Build GRCh38
rs769637625 Chr.13: 51771528-51771528 on Build GRCh38
rs772004504 Chr.13: 51771248-51771248 on Build GRCh38
rs572112078 Chr.13: 51771349-51771349 on Build GRCh38
rs141623866 Chr.13: 51771289-51771289 on Build GRCh38
rs1036488314 Chr.13: 51771470-51771470 on Build GRCh38
rs936949327 Chr.13: 51771199-51771199 on Build GRCh38
rs773665028 Chr.13: 51771350-51771350 on Build GRCh38
rs139624454 Chr.13: 51771469-51771469 on Build GRCh38
rs781463237 Chr.13: 51771257-51771259 on Build GRCh38
rs2296028 Chr.13: 51771501-51771501 on Build GRCh38
rs911357325 Chr.13: 51771113-51771113 on Build GRCh38

View Full Product Details