Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 51942291-51942829 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP7B
Entrez Gene ID
540
Gene Name
ATPase copper transporting beta
Gene Aliases
PWD, WC1, WD, WND
UniGene
Hs.492280
Gene Chromosome Location
Chr.13: 51932669-52012130 on Build GRCh38
Transcripts
NM_000053.3 NM_001005918.2 NM_001243182.1 XM_005266423.2 XM_005266424.4 XM_005266427.2 XM_005266428.1 XM_005266430.4 XM_005266431.3 XM_005266432.2 XM_006719837.3 XM_006719838.1 XM_006719839.1 XM_011535117.2 XM_011535118.1 XM_011535119.1 XM_011535122.2 XM_017020627.1 XM_017020628.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1367424 c.3325G>T p.V1109L Chr.13: 51942473-51942473 on Build GRCh38
5788866 c.3320G>C p.S1107T Chr.13: 51942478-51942478 on Build GRCh38
4664513 c.3317T>C p.V1106A Chr.13: 51942481-51942481 on Build GRCh38
6608205 c.3357T>C p.P1119P Chr.13: 51942441-51942441 on Build GRCh38
5654549 c.3370G>T p.A1124S Chr.13: 51942428-51942428 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs376492632 Chr.13: 51942339-51942339 on Build GRCh38
rs79742277 Chr.13: 51942622-51942622 on Build GRCh38
rs1015167774 Chr.13: 51942740-51942740 on Build GRCh38
rs182659444 Chr.13: 51942466-51942466 on Build GRCh38
rs748614005 Chr.13: 51942456-51942456 on Build GRCh38
rs749815334 Chr.13: 51942364-51942364 on Build GRCh38
rs772880934 Chr.13: 51942389-51942389 on Build GRCh38
rs868506510 Chr.13: 51942660-51942660 on Build GRCh38
rs1014509023 Chr.13: 51942723-51942723 on Build GRCh38
rs756389208 Chr.13: 51942436-51942436 on Build GRCh38
rs778086706 Chr.13: 51942462-51942462 on Build GRCh38
rs779495893 Chr.13: 51942454-51942454 on Build GRCh38
rs373081328 Chr.13: 51942393-51942393 on Build GRCh38
rs772372910 Chr.13: 51942387-51942387 on Build GRCh38
rs1034544448 Chr.13: 51942691-51942691 on Build GRCh38
rs74744480 Chr.13: 51942807-51942807 on Build GRCh38
rs566132264 Chr.13: 51942662-51942662 on Build GRCh38
rs552425082 Chr.13: 51942706-51942706 on Build GRCh38
rs786204584 Chr.13: 51942556-51942556 on Build GRCh38
rs763995218 Chr.13: 51942383-51942383 on Build GRCh38
rs764108297 Chr.13: 51942515-51942515 on Build GRCh38
rs761632029 Chr.13: 51942503-51942503 on Build GRCh38
rs770624100 Chr.13: 51942464-51942464 on Build GRCh38
rs1056865978 Chr.13: 51942692-51942692 on Build GRCh38
rs545284208 Chr.13: 51942446-51942446 on Build GRCh38
rs746051031 Chr.13: 51942340-51942340 on Build GRCh38
rs772464343 Chr.13: 51942458-51942458 on Build GRCh38
rs962259937 Chr.13: 51942768-51942768 on Build GRCh38
rs370947152 Chr.13: 51942402-51942402 on Build GRCh38
rs370823890 Chr.13: 51942357-51942357 on Build GRCh38
rs766632973 Chr.13: 51942507-51942507 on Build GRCh38
rs560952220 Chr.13: 51942493-51942493 on Build GRCh38
rs372456815 Chr.13: 51942474-51942474 on Build GRCh38
rs895442917 Chr.13: 51942475-51942475 on Build GRCh38
rs932774907 Chr.13: 51942614-51942614 on Build GRCh38
rs746731660 Chr.13: 51942463-51942463 on Build GRCh38
rs747387270 Chr.13: 51942559-51942559 on Build GRCh38
rs563659275 Chr.13: 51942670-51942670 on Build GRCh38
rs786204483 Chr.13: 51942497-51942497 on Build GRCh38
rs898334248 Chr.13: 51942322-51942322 on Build GRCh38
rs587783311 Chr.13: 51942417-51942417 on Build GRCh38
rs202233544 Chr.13: 51942445-51942445 on Build GRCh38
rs775541743 Chr.13: 51942481-51942481 on Build GRCh38
rs532404806 Chr.13: 51942673-51942673 on Build GRCh38
rs377020436 Chr.13: 51942522-51942522 on Build GRCh38
rs60579905 Chr.13: 51942773-51942773 on Build GRCh38
rs145887771 Chr.13: 51942396-51942396 on Build GRCh38
rs182659444 Chr.13: 51942466-51942466 on Build GRCh38
rs767078371 Chr.13: 51942325-51942325 on Build GRCh38
rs753250853 Chr.13: 51942535-51942535 on Build GRCh38
rs764041557 Chr.13: 51942487-51942487 on Build GRCh38
rs774368997 Chr.13: 51942601-51942601 on Build GRCh38
rs59120265 Chr.13: 51942432-51942432 on Build GRCh38
rs778825095 Chr.13: 51942526-51942526 on Build GRCh38
rs772615378 Chr.13: 51942376-51942377 on Build GRCh38
rs541208827 Chr.13: 51942482-51942482 on Build GRCh38
rs768717223 Chr.13: 51942598-51942598 on Build GRCh38
rs762876976 Chr.13: 51942431-51942431 on Build GRCh38
rs375820067 Chr.13: 51942537-51942537 on Build GRCh38
rs137853286 Chr.13: 51942398-51942398 on Build GRCh38
rs767154748 Chr.13: 51942569-51942570 on Build GRCh38
rs947281149 Chr.13: 51942666-51942666 on Build GRCh38
rs146623472 Chr.13: 51942430-51942430 on Build GRCh38
rs1012710284 Chr.13: 51942721-51942721 on Build GRCh38
rs760772190 Chr.13: 51942476-51942478 on Build GRCh38
rs1037713645 Chr.13: 51942319-51942319 on Build GRCh38
rs759109027 Chr.13: 51942473-51942473 on Build GRCh38
rs761686671 Chr.13: 51942603-51942603 on Build GRCh38
rs756305626 Chr.13: 51942343-51942343 on Build GRCh38
rs113218782 Chr.13: 51942318-51942318 on Build GRCh38
rs902990886 Chr.13: 51942672-51942672 on Build GRCh38
rs1008542300 Chr.13: 51942358-51942358 on Build GRCh38
rs769198765 Chr.13: 51942368-51942368 on Build GRCh38
rs368545738 Chr.13: 51942523-51942523 on Build GRCh38
rs780062799 Chr.13: 51942440-51942440 on Build GRCh38
rs760666435 Chr.13: 51942355-51942355 on Build GRCh38
rs149218548 Chr.13: 51942663-51942663 on Build GRCh38
rs1022618305 Chr.13: 51942756-51942756 on Build GRCh38
rs750248763 Chr.13: 51942593-51942593 on Build GRCh38
rs587783312 Chr.13: 51942399-51942399 on Build GRCh38
rs775129474 Chr.13: 51942587-51942587 on Build GRCh38
rs748522019 Chr.13: 51942379-51942379 on Build GRCh38
rs137853281 Chr.13: 51942396-51942396 on Build GRCh38
rs780932352 Chr.13: 51942582-51942582 on Build GRCh38
rs182086845 Chr.13: 51942331-51942331 on Build GRCh38
rs771153027 Chr.13: 51942572-51942572 on Build GRCh38
rs61733679 Chr.13: 51942429-51942429 on Build GRCh38
rs565888819 Chr.13: 51942371-51942371 on Build GRCh38
rs781332476 Chr.13: 51942584-51942584 on Build GRCh38
rs891438556 Chr.13: 51942394-51942394 on Build GRCh38
rs1042990266 Chr.13: 51942669-51942669 on Build GRCh38
rs539308473 Chr.13: 51942327-51942327 on Build GRCh38
rs187200982 Chr.13: 51942395-51942395 on Build GRCh38
rs953506898 Chr.13: 51942781-51942781 on Build GRCh38
rs1022670769 Chr.13: 51942520-51942520 on Build GRCh38
rs373698024 Chr.13: 51942422-51942422 on Build GRCh38
rs1006194197 Chr.13: 51942778-51942778 on Build GRCh38
rs1015850448 Chr.13: 51942789-51942789 on Build GRCh38

View Full Product Details