Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 59774638-59775143 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DIAPH3
Entrez Gene ID
81624
Gene Name
diaphanous related formin 3
Gene Aliases
AN, AUNA1, DIA2, DRF3, NSDAN, diap3, mDia2
UniGene
Hs.283127
Gene Chromosome Location
Chr.13: 59665583-60163985 on Build GRCh38
Transcripts
NM_001042517.1 NM_001258366.1 NM_001258367.1 NM_001258368.1 NM_001258369.1 NM_030932.3 XM_006719876.1 XM_011535258.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1477290 c.3187G>A p.D1063N Chr.13: 59774800-59774800 on Build GRCh38
6521104 c.3216G>T p.G1072G Chr.13: 59774771-59774771 on Build GRCh38
6521105 c.3179G>T p.G1060V Chr.13: 59774808-59774808 on Build GRCh38
6531774 c.3254C>G p.P1085R Chr.13: 59774733-59774733 on Build GRCh38
948191 c.3249G>A p.P1083P Chr.13: 59774738-59774738 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765864080 Chr.13: 59774748-59774751 on Build GRCh38
rs544328549 Chr.13: 59774748-59774748 on Build GRCh38
rs769704130 Chr.13: 59774708-59774708 on Build GRCh38
rs572778814 Chr.13: 59774693-59774693 on Build GRCh38
rs182834238 Chr.13: 59774700-59774700 on Build GRCh38
rs185048465 Chr.13: 59775050-59775050 on Build GRCh38
rs751595565 Chr.13: 59774777-59774777 on Build GRCh38
rs1020720170 Chr.13: 59774976-59774979 on Build GRCh38
rs527601529 Chr.13: 59774870-59774870 on Build GRCh38
rs201791921 Chr.13: 59774802-59774802 on Build GRCh38
rs139589054 Chr.13: 59775060-59775060 on Build GRCh38
rs187951023 Chr.13: 59774705-59774705 on Build GRCh38
rs1001872930 Chr.13: 59774916-59774916 on Build GRCh38
rs1053423199 Chr.13: 59774866-59774866 on Build GRCh38
rs904827577 Chr.13: 59774896-59774897 on Build GRCh38
rs758840929 Chr.13: 59775070-59775070 on Build GRCh38
rs761465883 Chr.13: 59774741-59774741 on Build GRCh38
rs766622481 Chr.13: 59774951-59774951 on Build GRCh38
rs767349693 Chr.13: 59774745-59774745 on Build GRCh38
rs1004742376 Chr.13: 59774936-59774936 on Build GRCh38
rs923688289 Chr.13: 59775068-59775068 on Build GRCh38
rs983375275 Chr.13: 59775094-59775094 on Build GRCh38
rs192229691 Chr.13: 59774965-59774965 on Build GRCh38
rs755540715 Chr.13: 59774685-59774685 on Build GRCh38
rs768487677 Chr.13: 59774825-59774825 on Build GRCh38
rs1032706883 Chr.13: 59774926-59774926 on Build GRCh38
rs779852860 Chr.13: 59774811-59774811 on Build GRCh38
rs886882703 Chr.13: 59774876-59774876 on Build GRCh38
rs1015674618 Chr.13: 59774947-59774947 on Build GRCh38
rs758546977 Chr.13: 59774695-59774695 on Build GRCh38
rs554551041 Chr.13: 59774681-59774681 on Build GRCh38
rs376432274 Chr.13: 59774739-59774739 on Build GRCh38
rs1027361991 Chr.13: 59774980-59774980 on Build GRCh38
rs757495408 Chr.13: 59774779-59774779 on Build GRCh38
rs765868029 Chr.13: 59775004-59775004 on Build GRCh38
rs372983167 Chr.13: 59774893-59774893 on Build GRCh38
rs1012544062 Chr.13: 59774878-59774878 on Build GRCh38
rs986088760 Chr.13: 59775022-59775022 on Build GRCh38
rs773484884 Chr.13: 59774853-59774853 on Build GRCh38
rs771830933 Chr.13: 59774843-59774843 on Build GRCh38
rs1053474519 Chr.13: 59775086-59775086 on Build GRCh38
rs920183360 Chr.13: 59774690-59774690 on Build GRCh38
rs920200218 Chr.13: 59775074-59775074 on Build GRCh38
rs762335116 Chr.13: 59774737-59774737 on Build GRCh38
rs749196642 Chr.13: 59774822-59774822 on Build GRCh38
rs531791483 Chr.13: 59774725-59774725 on Build GRCh38
rs371902789 Chr.13: 59774738-59774738 on Build GRCh38
rs368829900 Chr.13: 59774807-59774807 on Build GRCh38
rs931507510 Chr.13: 59774833-59774833 on Build GRCh38
rs760866906 Chr.13: 59774887-59774887 on Build GRCh38
rs373358939 Chr.13: 59774835-59774835 on Build GRCh38
rs1020352099 Chr.13: 59774669-59774669 on Build GRCh38
rs777746844 Chr.13: 59774698-59774698 on Build GRCh38
rs775453234 Chr.13: 59774718-59774718 on Build GRCh38
rs777126146 Chr.13: 59774752-59774752 on Build GRCh38
rs552161227 Chr.13: 59774910-59774910 on Build GRCh38
rs936383539 Chr.13: 59775069-59775069 on Build GRCh38
rs374884917 Chr.13: 59774818-59774818 on Build GRCh38
rs201517886 Chr.13: 59774761-59774761 on Build GRCh38
rs754015712 Chr.13: 59774962-59774962 on Build GRCh38
rs369429654 Chr.13: 59774867-59774867 on Build GRCh38
rs566092450 Chr.13: 59774834-59774834 on Build GRCh38
rs35877913 Chr.13: 59774919-59774919 on Build GRCh38
rs200654315 Chr.13: 59774760-59774760 on Build GRCh38
rs779624116 Chr.13: 59774692-59774692 on Build GRCh38
rs777943433 Chr.13: 59775101-59775101 on Build GRCh38
rs751075566 Chr.13: 59774779-59774779 on Build GRCh38
rs369935944 Chr.13: 59774759-59774759 on Build GRCh38
rs201971399 Chr.13: 59774735-59774735 on Build GRCh38
rs537625986 Chr.13: 59774985-59774985 on Build GRCh38
rs771071527 Chr.13: 59774704-59774704 on Build GRCh38
rs369382898 Chr.13: 59774689-59774689 on Build GRCh38
rs932937205 Chr.13: 59774719-59774719 on Build GRCh38
rs760029457 Chr.13: 59774757-59774757 on Build GRCh38
rs964431933 Chr.13: 59775053-59775053 on Build GRCh38
rs765809400 Chr.13: 59774758-59774758 on Build GRCh38
rs73208932 Chr.13: 59775040-59775040 on Build GRCh38
rs867747061 Chr.13: 59775051-59775051 on Build GRCh38
rs554215951 Chr.13: 59775079-59775079 on Build GRCh38

View Full Product Details