Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 70107486-70107992 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN8OS
Entrez Gene ID
6315
Gene Name
ATXN8 opposite strand (non-protein coding)
Gene Aliases
KLHL1AS, NCRNA00003, SCA8
UniGene
Hs.676453
Gene Chromosome Location
Chr.13: 70107213-70139753 on Build GRCh38
Transcripts
NR_002717.2 NM_001286725.1 NM_020866.2

Gene Symbol
KLHL1
Entrez Gene ID
57626
Gene Name
kelch like family member 1
Gene Aliases
MRP2
UniGene
Hs.508201
Gene Chromosome Location
Chr.13: 69700593-70108493 on Build GRCh38
Transcripts
NR_002717.2 NM_001286725.1 NM_020866.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6286859 c.185G>T p.R62I Chr.13: 70107515-70107515 on Build GRCh38
6696437 c.164G>T p.R55L Chr.13: 70107536-70107536 on Build GRCh38
6358962 c.72C>G p.H24Q Chr.13: 70107628-70107628 on Build GRCh38
118191 c.171C>T p.L57L Chr.13: 70107529-70107529 on Build GRCh38
4534670 c.210G>A p.K70K Chr.13: 70107490-70107490 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989550053 Chr.13: 70107792-70107792 on Build GRCh38
rs140989246 Chr.13: 70107626-70107626 on Build GRCh38
rs1005673658 Chr.13: 70107544-70107544 on Build GRCh38
rs746738440 Chr.13: 70107706-70107706 on Build GRCh38
rs41283978 Chr.13: 70107600-70107600 on Build GRCh38
rs141447437 Chr.13: 70107577-70107577 on Build GRCh38
rs188602966 Chr.13: 70107873-70107873 on Build GRCh38
rs757237410 Chr.13: 70107682-70107683 on Build GRCh38
rs767770132 Chr.13: 70107661-70107661 on Build GRCh38
rs111324959 Chr.13: 70107517-70107517 on Build GRCh38
rs773097859 Chr.13: 70107645-70107645 on Build GRCh38
rs184946008 Chr.13: 70107704-70107704 on Build GRCh38
rs754998896 Chr.13: 70107683-70107683 on Build GRCh38
rs757814028 Chr.13: 70107777-70107777 on Build GRCh38
rs547047442 Chr.13: 70107697-70107697 on Build GRCh38
rs1012496803 Chr.13: 70107533-70107533 on Build GRCh38
rs777661082 Chr.13: 70107595-70107596 on Build GRCh38
rs781658816 Chr.13: 70107614-70107614 on Build GRCh38
rs762708051 Chr.13: 70107660-70107660 on Build GRCh38
rs757649187 Chr.13: 70107590-70107590 on Build GRCh38
rs773746540 Chr.13: 70107529-70107529 on Build GRCh38
rs764920556 Chr.13: 70107610-70107610 on Build GRCh38
rs749307923 Chr.13: 70107627-70107627 on Build GRCh38
rs993877697 Chr.13: 70107802-70107802 on Build GRCh38
rs769741754 Chr.13: 70107596-70107596 on Build GRCh38
rs3751429 Chr.13: 70107824-70107824 on Build GRCh38
rs764863092 Chr.13: 70107653-70107653 on Build GRCh38
rs191673952 Chr.13: 70107613-70107613 on Build GRCh38
rs371093873 Chr.13: 70107721-70107721 on Build GRCh38
rs148367063 Chr.13: 70107715-70107715 on Build GRCh38
rs781511350 Chr.13: 70107593-70107593 on Build GRCh38
rs776718823 Chr.13: 70107650-70107650 on Build GRCh38
rs754317021 Chr.13: 70107749-70107749 on Build GRCh38
rs145049894 Chr.13: 70107649-70107649 on Build GRCh38
rs746410016 Chr.13: 70107594-70107594 on Build GRCh38
rs941154473 Chr.13: 70107854-70107854 on Build GRCh38
rs534215165 Chr.13: 70107841-70107841 on Build GRCh38
rs749011008 Chr.13: 70107596-70107596 on Build GRCh38
rs985795185 Chr.13: 70107832-70107832 on Build GRCh38
rs746592989 Chr.13: 70107926-70107926 on Build GRCh38
rs192961688 Chr.13: 70107946-70107946 on Build GRCh38
rs768406656 Chr.13: 70107542-70107542 on Build GRCh38
rs576342466 Chr.13: 70107949-70107949 on Build GRCh38
rs762498239 Chr.13: 70107734-70107734 on Build GRCh38
rs773878923 Chr.13: 70107740-70107740 on Build GRCh38
rs749505853 Chr.13: 70107598-70107598 on Build GRCh38
rs978058798 Chr.13: 70107805-70107805 on Build GRCh38
rs929672585 Chr.13: 70107902-70107902 on Build GRCh38
rs1055767822 Chr.13: 70107941-70107941 on Build GRCh38
rs781731142 Chr.13: 70107519-70107519 on Build GRCh38
rs144867124 Chr.13: 70107689-70107689 on Build GRCh38
rs3751428 Chr.13: 70107741-70107741 on Build GRCh38
rs1016613270 Chr.13: 70107573-70107573 on Build GRCh38
rs906835091 Chr.13: 70107923-70107923 on Build GRCh38
rs761796623 Chr.13: 70107565-70107565 on Build GRCh38
rs935425832 Chr.13: 70107879-70107879 on Build GRCh38
rs751816506 Chr.13: 70107588-70107588 on Build GRCh38
rs868002168 Chr.13: 70107900-70107900 on Build GRCh38
rs556389912 Chr.13: 70107873-70107873 on Build GRCh38
rs756483048 Chr.13: 70107669-70107669 on Build GRCh38
rs1005270099 Chr.13: 70107569-70107569 on Build GRCh38
rs376267562 Chr.13: 70107574-70107574 on Build GRCh38
rs201323322 Chr.13: 70107617-70107617 on Build GRCh38
rs577420693 Chr.13: 70107889-70107889 on Build GRCh38
rs1018168298 Chr.13: 70107833-70107833 on Build GRCh38
rs1004169851 Chr.13: 70107784-70107784 on Build GRCh38
rs1045410859 Chr.13: 70107916-70107916 on Build GRCh38
rs768924039 Chr.13: 70107599-70107599 on Build GRCh38
rs758706872 Chr.13: 70107692-70107692 on Build GRCh38
rs763743600 Chr.13: 70107736-70107736 on Build GRCh38
rs923934409 Chr.13: 70107874-70107874 on Build GRCh38
rs966717808 Chr.13: 70107804-70107804 on Build GRCh38
rs1049974608 Chr.13: 70107534-70107534 on Build GRCh38
rs555272767 Chr.13: 70107787-70107787 on Build GRCh38
rs959783050 Chr.13: 70107790-70107790 on Build GRCh38
rs111575544 Chr.13: 70107931-70107931 on Build GRCh38
rs147322205 Chr.13: 70107612-70107612 on Build GRCh38
rs1038043140 Chr.13: 70107948-70107948 on Build GRCh38
rs565048506 Chr.13: 70107510-70107510 on Build GRCh38
rs1033866933 Chr.13: 70107786-70107786 on Build GRCh38
rs896846795 Chr.13: 70107735-70107735 on Build GRCh38
rs778272080 Chr.13: 70107582-70107582 on Build GRCh38
rs766748389 Chr.13: 70107743-70107743 on Build GRCh38
rs942281442 Chr.13: 70107938-70107938 on Build GRCh38
rs139358653 Chr.13: 70107601-70107601 on Build GRCh38
rs544464374 Chr.13: 70107848-70107848 on Build GRCh38
rs770884636 Chr.13: 70107707-70107707 on Build GRCh38
rs755054363 Chr.13: 70107631-70107631 on Build GRCh38
rs530631519 Chr.13: 70107723-70107723 on Build GRCh38
rs753459470 Chr.13: 70107605-70107605 on Build GRCh38
rs985751021 Chr.13: 70107826-70107826 on Build GRCh38
rs754747379 Chr.13: 70107580-70107580 on Build GRCh38
rs922553554 Chr.13: 70107806-70107806 on Build GRCh38
rs780150653 Chr.13: 70107597-70107597 on Build GRCh38
rs111444961 Chr.13: 70107583-70107583 on Build GRCh38
rs774985458 Chr.13: 70107654-70107654 on Build GRCh38
rs375724042 Chr.13: 70107521-70107521 on Build GRCh38
rs992229238 Chr.13: 70107887-70107887 on Build GRCh38
rs748421357 Chr.13: 70107639-70107639 on Build GRCh38
rs1016616139 Chr.13: 70107575-70107575 on Build GRCh38
rs746909985 Chr.13: 70107647-70107647 on Build GRCh38
rs749846665 Chr.13: 70107767-70107767 on Build GRCh38
rs1038153795 Chr.13: 70107885-70107885 on Build GRCh38
rs778959588 Chr.13: 70107636-70107636 on Build GRCh38
rs1022325661 Chr.13: 70107798-70107798 on Build GRCh38
rs753112360 Chr.13: 70107672-70107672 on Build GRCh38
rs754369885 Chr.13: 70107677-70107677 on Build GRCh38
rs370644626 Chr.13: 70107576-70107576 on Build GRCh38
rs145694020 Chr.13: 70107609-70107609 on Build GRCh38
rs763213239 Chr.13: 70107538-70107538 on Build GRCh38
rs775643172 Chr.13: 70107536-70107536 on Build GRCh38
rs998017866 Chr.13: 70107530-70107530 on Build GRCh38
rs968043119 Chr.13: 70107862-70107862 on Build GRCh38
rs750856218 Chr.13: 70107662-70107662 on Build GRCh38
rs774196463 Chr.13: 70107552-70107552 on Build GRCh38
rs368924182 Chr.13: 70107708-70107708 on Build GRCh38
rs769949629 Chr.13: 70107531-70107531 on Build GRCh38

View Full Product Details