Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77895998-77896498 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDNRB
Entrez Gene ID
1910
Gene Name
endothelin receptor type B
Gene Aliases
ABCDS, ET-B, ET-BR, ETB, ETB1, ETBR, ETRB, HSCR, HSCR2, WS4A
UniGene
Hs.732046
Gene Chromosome Location
Chr.13: 77895481-77975529 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Gene Symbol
EDNRB-AS1
Entrez Gene ID
100505518
Gene Name
EDNRB antisense RNA 1
Gene Aliases
-
UniGene
Hs.742215
Gene Chromosome Location
Chr.13: 77818937-77908442 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1367888 c.1306A>G p.N436D Chr.13: 77896447-77896447 on Build GRCh38
3700424 c.1292G>T p.R431I Chr.13: 77896461-77896461 on Build GRCh38
4638177 c.1310A>C p.*437S Chr.13: 77896443-77896443 on Build GRCh38
4945142 c.1258A>G p.T420A Chr.13: 77896495-77896495 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs201086847 Chr.13: 77896035-77896035 on Build GRCh38
rs779137623 Chr.13: 77896396-77896396 on Build GRCh38
rs200248884 Chr.13: 77896462-77896462 on Build GRCh38
rs921424917 Chr.13: 77896362-77896362 on Build GRCh38
rs998358843 Chr.13: 77896094-77896094 on Build GRCh38
rs200943218 Chr.13: 77896428-77896428 on Build GRCh38
rs191136728 Chr.13: 77896033-77896033 on Build GRCh38
rs201311811 Chr.13: 77896356-77896356 on Build GRCh38
rs3027092 Chr.13: 77896433-77896433 on Build GRCh38
rs201189051 Chr.13: 77896181-77896181 on Build GRCh38
rs200768146 Chr.13: 77896286-77896286 on Build GRCh38
rs886050320 Chr.13: 77896294-77896294 on Build GRCh38
rs201629890 Chr.13: 77896245-77896245 on Build GRCh38
rs201936958 Chr.13: 77896032-77896032 on Build GRCh38
rs957614996 Chr.13: 77896179-77896179 on Build GRCh38
rs201150464 Chr.13: 77896470-77896470 on Build GRCh38
rs368094735 Chr.13: 77896248-77896248 on Build GRCh38
rs74529986 Chr.13: 77896390-77896390 on Build GRCh38
rs780478968 Chr.13: 77896285-77896285 on Build GRCh38
rs1051262746 Chr.13: 77896473-77896473 on Build GRCh38
rs199574691 Chr.13: 77896270-77896270 on Build GRCh38
rs886050321 Chr.13: 77896340-77896340 on Build GRCh38
rs12720204 Chr.13: 77896145-77896145 on Build GRCh38
rs758238086 Chr.13: 77896425-77896425 on Build GRCh38
rs201072047 Chr.13: 77896103-77896103 on Build GRCh38
rs771397644 Chr.13: 77896037-77896037 on Build GRCh38
rs896939028 Chr.13: 77896090-77896090 on Build GRCh38
rs201850106 Chr.13: 77896345-77896345 on Build GRCh38
rs74878537 Chr.13: 77896392-77896392 on Build GRCh38
rs184732068 Chr.13: 77896435-77896435 on Build GRCh38
rs1020620475 Chr.13: 77896187-77896187 on Build GRCh38
rs200321220 Chr.13: 77896096-77896096 on Build GRCh38
rs199537579 Chr.13: 77896153-77896153 on Build GRCh38
rs200276436 Chr.13: 77896159-77896159 on Build GRCh38
rs202187389 Chr.13: 77896108-77896108 on Build GRCh38
rs912231842 Chr.13: 77896475-77896475 on Build GRCh38
rs528216732 Chr.13: 77896280-77896280 on Build GRCh38
rs1013493746 Chr.13: 77896254-77896254 on Build GRCh38
rs982838025 Chr.13: 77896053-77896053 on Build GRCh38
rs1005430643 Chr.13: 77896047-77896047 on Build GRCh38
rs962820782 Chr.13: 77896328-77896328 on Build GRCh38
rs200394131 Chr.13: 77896427-77896427 on Build GRCh38
rs564303926 Chr.13: 77896261-77896261 on Build GRCh38
rs950216760 Chr.13: 77896234-77896234 on Build GRCh38
rs982869085 Chr.13: 77896318-77896318 on Build GRCh38
rs200985166 Chr.13: 77896337-77896337 on Build GRCh38
rs202170085 Chr.13: 77896063-77896063 on Build GRCh38
rs376095550 Chr.13: 77896430-77896430 on Build GRCh38
rs900118914 Chr.13: 77896106-77896106 on Build GRCh38
rs770758645 Chr.13: 77896432-77896432 on Build GRCh38
rs917233638 Chr.13: 77896039-77896039 on Build GRCh38
rs1009289013 Chr.13: 77896154-77896154 on Build GRCh38
rs200431243 Chr.13: 77896136-77896136 on Build GRCh38
rs765413187 Chr.13: 77896078-77896078 on Build GRCh38
rs201956974 Chr.13: 77896406-77896406 on Build GRCh38
rs76025653 Chr.13: 77896394-77896394 on Build GRCh38
rs4885491 Chr.13: 77896215-77896215 on Build GRCh38
rs1038625378 Chr.13: 77896102-77896102 on Build GRCh38
rs574887769 Chr.13: 77896117-77896117 on Build GRCh38
rs41288258 Chr.13: 77896308-77896308 on Build GRCh38
rs199887964 Chr.13: 77896323-77896323 on Build GRCh38
rs200008387 Chr.13: 77896354-77896354 on Build GRCh38
rs199877664 Chr.13: 77896195-77896195 on Build GRCh38
rs769383742 Chr.13: 77896458-77896458 on Build GRCh38
rs941488411 Chr.13: 77896095-77896096 on Build GRCh38
rs77041871 Chr.13: 77896391-77896391 on Build GRCh38

View Full Product Details