Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 77900442-77900955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EDNRB
Entrez Gene ID
1910
Gene Name
endothelin receptor type B
Gene Aliases
ABCDS, ET-B, ET-BR, ETB, ETB1, ETBR, ETRB, HSCR, HSCR2, WS4A
UniGene
Hs.732046
Gene Chromosome Location
Chr.13: 77895481-77975529 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Gene Symbol
EDNRB-AS1
Entrez Gene ID
100505518
Gene Name
EDNRB antisense RNA 1
Gene Aliases
-
UniGene
Hs.742215
Gene Chromosome Location
Chr.13: 77818937-77908442 on Build GRCh38
Transcripts
NM_000115.3 NM_001122659.2 NM_001201397.1 NM_003991.3 NR_103853.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7301662 c.1079T>C p.L360P Chr.13: 77900527-77900527 on Build GRCh38
4525364 c.1326G>C p.Q442H Chr.13: 77900550-77900550 on Build GRCh38
6651680 c.1285C>A p.L429I Chr.13: 77900591-77900591 on Build GRCh38
6651684 c.1056G>T p.Q352H Chr.13: 77900550-77900550 on Build GRCh38
4774804 c.1039C>T p.L347F Chr.13: 77900567-77900567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771999239 Chr.13: 77900498-77900498 on Build GRCh38
rs1006232128 Chr.13: 77900738-77900738 on Build GRCh38
rs779498069 Chr.13: 77900687-77900687 on Build GRCh38
rs947108047 Chr.13: 77900482-77900482 on Build GRCh38
rs531691342 Chr.13: 77900709-77900709 on Build GRCh38
rs930503672 Chr.13: 77900707-77900707 on Build GRCh38
rs201988235 Chr.13: 77900704-77900704 on Build GRCh38
rs747204572 Chr.13: 77900519-77900519 on Build GRCh38
rs201437745 Chr.13: 77900633-77900633 on Build GRCh38
rs538853995 Chr.13: 77900797-77900797 on Build GRCh38
rs201503461 Chr.13: 77900481-77900481 on Build GRCh38
rs199798249 Chr.13: 77900668-77900668 on Build GRCh38
rs868348559 Chr.13: 77900642-77900642 on Build GRCh38
rs1026731943 Chr.13: 77900706-77900706 on Build GRCh38
rs1008663884 Chr.13: 77900783-77900783 on Build GRCh38
rs113966737 Chr.13: 77900833-77900833 on Build GRCh38
rs200303663 Chr.13: 77900496-77900496 on Build GRCh38
rs905668367 Chr.13: 77900509-77900509 on Build GRCh38
rs1031156335 Chr.13: 77900910-77900910 on Build GRCh38
rs369090616 Chr.13: 77900522-77900522 on Build GRCh38
rs200746009 Chr.13: 77900701-77900701 on Build GRCh38
rs866808412 Chr.13: 77900597-77900597 on Build GRCh38
rs964810060 Chr.13: 77900770-77900770 on Build GRCh38
rs116962929 Chr.13: 77900723-77900723 on Build GRCh38
rs904108774 Chr.13: 77900865-77900865 on Build GRCh38
rs1040505584 Chr.13: 77900796-77900796 on Build GRCh38
rs759543922 Chr.13: 77900650-77900650 on Build GRCh38
rs369891478 Chr.13: 77900693-77900693 on Build GRCh38
rs747292537 Chr.13: 77900700-77900700 on Build GRCh38
rs200428001 Chr.13: 77900479-77900479 on Build GRCh38
rs201952865 Chr.13: 77900491-77900491 on Build GRCh38
rs773618614 Chr.13: 77900506-77900506 on Build GRCh38
rs759192823 Chr.13: 77900565-77900565 on Build GRCh38
rs916597014 Chr.13: 77900493-77900493 on Build GRCh38
rs754237530 Chr.13: 77900669-77900669 on Build GRCh38
rs755259146 Chr.13: 77900686-77900686 on Build GRCh38
rs765224396 Chr.13: 77900601-77900601 on Build GRCh38
rs555515280 Chr.13: 77900887-77900887 on Build GRCh38
rs976310328 Chr.13: 77900802-77900802 on Build GRCh38
rs57369841 Chr.13: 77900634-77900634 on Build GRCh38
rs200363611 Chr.13: 77900651-77900651 on Build GRCh38
rs956485720 Chr.13: 77900886-77900886 on Build GRCh38
rs770915321 Chr.13: 77900529-77900529 on Build GRCh38
rs761061467 Chr.13: 77900815-77900815 on Build GRCh38
rs914210884 Chr.13: 77900909-77900909 on Build GRCh38
rs776554067 Chr.13: 77900564-77900564 on Build GRCh38
rs571492508 Chr.13: 77900755-77900755 on Build GRCh38
rs373012448 Chr.13: 77900665-77900665 on Build GRCh38
rs370856353 Chr.13: 77900489-77900489 on Build GRCh38

View Full Product Details