Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95433521-95434031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN10
Entrez Gene ID
9071
Gene Name
claudin 10
Gene Aliases
CPETRL3, OSP-L
UniGene
Hs.534377
Gene Chromosome Location
Chr.13: 95433588-95579756 on Build GRCh38
Transcripts
NM_001160100.1 NM_182848.3 XM_017020843.1 XM_017020844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6139472 c.183C>A p.C61* Chr.13: 95434016-95434016 on Build GRCh38
3814158 c.43G>A p.G15R Chr.13: 95433876-95433876 on Build GRCh38
6139473 c.95C>T p.T32M Chr.13: 95433928-95433928 on Build GRCh38
368581 c.127G>A p.V43I Chr.13: 95433960-95433960 on Build GRCh38
336031 c.179A>G p.H60R Chr.13: 95434012-95434012 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs555827487 Chr.13: 95433995-95433995 on Build GRCh38
rs771610353 Chr.13: 95433921-95433921 on Build GRCh38
rs1024760103 Chr.13: 95433910-95433910 on Build GRCh38
rs747687660 Chr.13: 95433909-95433909 on Build GRCh38
rs568647578 Chr.13: 95433749-95433749 on Build GRCh38
rs374417711 Chr.13: 95433890-95433890 on Build GRCh38
rs763024030 Chr.13: 95433966-95433966 on Build GRCh38
rs149372773 Chr.13: 95433941-95433941 on Build GRCh38
rs376321996 Chr.13: 95433828-95433828 on Build GRCh38
rs539415703 Chr.13: 95433848-95433848 on Build GRCh38
rs759786238 Chr.13: 95433844-95433844 on Build GRCh38
rs773857197 Chr.13: 95433832-95433832 on Build GRCh38
rs899451165 Chr.13: 95433818-95433818 on Build GRCh38
rs779035142 Chr.13: 95433793-95433793 on Build GRCh38
rs747059689 Chr.13: 95433969-95433972 on Build GRCh38
rs960733536 Chr.13: 95433676-95433676 on Build GRCh38
rs3751334 Chr.13: 95433756-95433756 on Build GRCh38
rs757224911 Chr.13: 95433878-95433878 on Build GRCh38
rs1022171884 Chr.13: 95433608-95433608 on Build GRCh38
rs866806839 Chr.13: 95433986-95433986 on Build GRCh38
rs768312342 Chr.13: 95433904-95433904 on Build GRCh38
rs764429956 Chr.13: 95433979-95433979 on Build GRCh38
rs779968361 Chr.13: 95433900-95433900 on Build GRCh38
rs780076642 Chr.13: 95433835-95433835 on Build GRCh38
rs1034715214 Chr.13: 95433664-95433664 on Build GRCh38
rs780229172 Chr.13: 95433795-95433796 on Build GRCh38
rs900379191 Chr.13: 95433581-95433581 on Build GRCh38
rs748827025 Chr.13: 95433902-95433902 on Build GRCh38
rs115306504 Chr.13: 95433626-95433626 on Build GRCh38
rs917735330 Chr.13: 95433585-95433585 on Build GRCh38
rs774985955 Chr.13: 95433824-95433824 on Build GRCh38
rs760145374 Chr.13: 95433934-95433934 on Build GRCh38
rs767766614 Chr.13: 95433998-95433998 on Build GRCh38
rs369938854 Chr.13: 95433886-95433886 on Build GRCh38
rs201997413 Chr.13: 95434010-95434010 on Build GRCh38
rs777992637 Chr.13: 95433771-95433771 on Build GRCh38
rs751698463 Chr.13: 95433872-95433872 on Build GRCh38
rs150191933 Chr.13: 95433861-95433861 on Build GRCh38
rs1053758566 Chr.13: 95433726-95433726 on Build GRCh38
rs753002912 Chr.13: 95433794-95433794 on Build GRCh38
rs755850012 Chr.13: 95433895-95433895 on Build GRCh38
rs752800223 Chr.13: 95433705-95433705 on Build GRCh38
rs777098814 Chr.13: 95433929-95433929 on Build GRCh38
rs373210781 Chr.13: 95433803-95433803 on Build GRCh38
rs1005029558 Chr.13: 95433804-95433804 on Build GRCh38
rs753689971 Chr.13: 95434009-95434009 on Build GRCh38
rs949213509 Chr.13: 95433590-95433590 on Build GRCh38
rs751873803 Chr.13: 95433987-95433987 on Build GRCh38
rs900503821 Chr.13: 95433738-95433738 on Build GRCh38
rs570623351 Chr.13: 95433807-95433807 on Build GRCh38
rs61736194 Chr.13: 95433769-95433769 on Build GRCh38
rs549033030 Chr.13: 95433614-95433614 on Build GRCh38
rs554524498 Chr.13: 95433780-95433780 on Build GRCh38
rs199739747 Chr.13: 95433948-95433948 on Build GRCh38
rs746935105 Chr.13: 95433801-95433801 on Build GRCh38
rs555298278 Chr.13: 95433905-95433905 on Build GRCh38
rs756083702 Chr.13: 95434001-95434001 on Build GRCh38
rs918845225 Chr.13: 95433761-95433761 on Build GRCh38
rs1017391022 Chr.13: 95433565-95433565 on Build GRCh38
rs769674763 Chr.13: 95433808-95433808 on Build GRCh38
rs779656350 Chr.13: 95434002-95434002 on Build GRCh38
rs990572476 Chr.13: 95433574-95433574 on Build GRCh38
rs776939421 Chr.13: 95433843-95433843 on Build GRCh38
rs765511486 Chr.13: 95433940-95433940 on Build GRCh38
rs923905182 Chr.13: 95433696-95433696 on Build GRCh38
rs758384695 Chr.13: 95433795-95433795 on Build GRCh38

View Full Product Details