Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95560303-95560806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN10
Entrez Gene ID
9071
Gene Name
claudin 10
Gene Aliases
CPETRL3, OSP-L
UniGene
Hs.534377
Gene Chromosome Location
Chr.13: 95433588-95579756 on Build GRCh38
Transcripts
NM_001160100.1 NM_006984.4 NM_182848.3 XM_011521134.2 XM_017020843.1 XM_017020844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
168918 c.432C>A p.F144L Chr.13: 95560437-95560437 on Build GRCh38
6629949 c.442G>T p.D148Y Chr.13: 95560441-95560441 on Build GRCh38
4607586 c.383G>T p.C128F Chr.13: 95560388-95560388 on Build GRCh38
5815508 c.440C>T p.P147L Chr.13: 95560445-95560445 on Build GRCh38
948985 c.388A>G p.M130V Chr.13: 95560393-95560393 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971742258 Chr.13: 95560643-95560643 on Build GRCh38
rs201430734 Chr.13: 95560479-95560479 on Build GRCh38
rs758369755 Chr.13: 95560476-95560476 on Build GRCh38
rs550013383 Chr.13: 95560717-95560717 on Build GRCh38
rs374187735 Chr.13: 95560430-95560430 on Build GRCh38
rs766827012 Chr.13: 95560453-95560453 on Build GRCh38
rs768046859 Chr.13: 95560441-95560441 on Build GRCh38
rs769511383 Chr.13: 95560511-95560511 on Build GRCh38
rs752846602 Chr.13: 95560474-95560474 on Build GRCh38
rs189097534 Chr.13: 95560714-95560714 on Build GRCh38
rs985777928 Chr.13: 95560704-95560704 on Build GRCh38
rs760273082 Chr.13: 95560767-95560767 on Build GRCh38
rs545889393 Chr.13: 95560679-95560679 on Build GRCh38
rs781105813 Chr.13: 95560501-95560501 on Build GRCh38
rs775158736 Chr.13: 95560764-95560764 on Build GRCh38
rs373345015 Chr.13: 95560356-95560356 on Build GRCh38
rs745770511 Chr.13: 95560460-95560460 on Build GRCh38
rs1049064936 Chr.13: 95560414-95560414 on Build GRCh38
rs888875713 Chr.13: 95560689-95560689 on Build GRCh38
rs368714235 Chr.13: 95560447-95560447 on Build GRCh38
rs991260264 Chr.13: 95560742-95560742 on Build GRCh38
rs1044483008 Chr.13: 95560490-95560490 on Build GRCh38
rs755231416 Chr.13: 95560464-95560464 on Build GRCh38
rs963255561 Chr.13: 95560758-95560758 on Build GRCh38
rs540660973 Chr.13: 95560700-95560700 on Build GRCh38
rs929251225 Chr.13: 95560750-95560750 on Build GRCh38
rs770886300 Chr.13: 95560421-95560421 on Build GRCh38
rs1012209557 Chr.13: 95560676-95560676 on Build GRCh38
rs387907413 Chr.13: 95560419-95560419 on Build GRCh38
rs747968404 Chr.13: 95560377-95560377 on Build GRCh38
rs766479345 Chr.13: 95560342-95560342 on Build GRCh38
rs1003078169 Chr.13: 95560537-95560537 on Build GRCh38
rs149697610 Chr.13: 95560461-95560461 on Build GRCh38
rs116133291 Chr.13: 95560777-95560777 on Build GRCh38
rs370703602 Chr.13: 95560393-95560393 on Build GRCh38
rs761005313 Chr.13: 95560451-95560451 on Build GRCh38
rs959462709 Chr.13: 95560739-95560740 on Build GRCh38
rs4773917 Chr.13: 95560720-95560720 on Build GRCh38
rs577207328 Chr.13: 95560628-95560628 on Build GRCh38
rs113065528 Chr.13: 95560328-95560328 on Build GRCh38
rs756292784 Chr.13: 95560335-95560335 on Build GRCh38
rs745456011 Chr.13: 95560509-95560509 on Build GRCh38
rs745423311 Chr.13: 95560672-95560672 on Build GRCh38
rs909106083 Chr.13: 95560624-95560624 on Build GRCh38
rs553863173 Chr.13: 95560568-95560568 on Build GRCh38
rs542466700 Chr.13: 95560358-95560358 on Build GRCh38
rs762262906 Chr.13: 95560434-95560434 on Build GRCh38
rs116119435 Chr.13: 95560431-95560431 on Build GRCh38
rs376283811 Chr.13: 95560359-95560359 on Build GRCh38
rs756413282 Chr.13: 95560739-95560739 on Build GRCh38
rs754139363 Chr.13: 95560346-95560346 on Build GRCh38
rs746988094 Chr.13: 95560491-95560491 on Build GRCh38
rs564156653 Chr.13: 95560710-95560710 on Build GRCh38
rs918949863 Chr.13: 95560651-95560651 on Build GRCh38
rs527392614 Chr.13: 95560695-95560695 on Build GRCh38
rs991291165 Chr.13: 95560506-95560506 on Build GRCh38
rs141876655 Chr.13: 95560713-95560715 on Build GRCh38
rs967684538 Chr.13: 95560701-95560701 on Build GRCh38
rs765170859 Chr.13: 95560466-95560466 on Build GRCh38
rs776122725 Chr.13: 95560429-95560429 on Build GRCh38
rs79906565 Chr.13: 95560401-95560401 on Build GRCh38
rs185670386 Chr.13: 95560774-95560774 on Build GRCh38
rs543974972 Chr.13: 95560775-95560775 on Build GRCh38
rs181761058 Chr.13: 95560729-95560729 on Build GRCh38
rs982641603 Chr.13: 95560470-95560470 on Build GRCh38
rs767815108 Chr.13: 95560332-95560332 on Build GRCh38

View Full Product Details