Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 95576975-95577478 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLDN10
Entrez Gene ID
9071
Gene Name
claudin 10
Gene Aliases
CPETRL3, OSP-L
UniGene
Hs.534377
Gene Chromosome Location
Chr.13: 95433588-95579756 on Build GRCh38
Transcripts
NM_001160100.1 NM_006984.4 NM_182848.3 XM_011521134.2 XM_017020843.1 XM_017020844.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5528727 c.465-1G>A p.? Chr.13: 95577230-95577230 on Build GRCh38
5679462 c.545C>A p.S182* Chr.13: 95577311-95577311 on Build GRCh38
5899082 c.520G>A p.G174S Chr.13: 95577292-95577292 on Build GRCh38
4672674 c.504A>G p.S168S Chr.13: 95577276-95577276 on Build GRCh38
4048821 c.552T>C p.S184S Chr.13: 95577318-95577318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs940237034 Chr.13: 95577093-95577093 on Build GRCh38
rs199796470 Chr.13: 95577246-95577246 on Build GRCh38
rs773696060 Chr.13: 95577204-95577204 on Build GRCh38
rs775021598 Chr.13: 95577382-95577384 on Build GRCh38
rs765526989 Chr.13: 95577221-95577221 on Build GRCh38
rs768204475 Chr.13: 95577197-95577197 on Build GRCh38
rs201348784 Chr.13: 95577271-95577271 on Build GRCh38
rs377203980 Chr.13: 95577399-95577399 on Build GRCh38
rs756242071 Chr.13: 95577400-95577400 on Build GRCh38
rs562952757 Chr.13: 95577170-95577170 on Build GRCh38
rs748769353 Chr.13: 95577186-95577186 on Build GRCh38
rs759626987 Chr.13: 95577218-95577218 on Build GRCh38
rs904908913 Chr.13: 95577003-95577003 on Build GRCh38
rs199672474 Chr.13: 95577405-95577405 on Build GRCh38
rs998743795 Chr.13: 95577325-95577325 on Build GRCh38
rs540645477 Chr.13: 95577444-95577444 on Build GRCh38
rs1055588331 Chr.13: 95577245-95577245 on Build GRCh38
rs371375503 Chr.13: 95577226-95577226 on Build GRCh38
rs1027605362 Chr.13: 95577042-95577042 on Build GRCh38
rs936497509 Chr.13: 95577327-95577327 on Build GRCh38
rs925138976 Chr.13: 95577323-95577323 on Build GRCh38
rs202080749 Chr.13: 95577191-95577191 on Build GRCh38
rs965364594 Chr.13: 95577169-95577169 on Build GRCh38
rs1038602578 Chr.13: 95577122-95577122 on Build GRCh38
rs764323936 Chr.13: 95577257-95577257 on Build GRCh38
rs755905660 Chr.13: 95577285-95577285 on Build GRCh38
rs1045954410 Chr.13: 95577281-95577281 on Build GRCh38
rs569728685 Chr.13: 95577095-95577095 on Build GRCh38
rs778401094 Chr.13: 95577338-95577338 on Build GRCh38
rs750267230 Chr.13: 95577283-95577283 on Build GRCh38
rs1032858832 Chr.13: 95577334-95577334 on Build GRCh38
rs934213809 Chr.13: 95577041-95577041 on Build GRCh38
rs975474857 Chr.13: 95577032-95577032 on Build GRCh38
rs779127315 Chr.13: 95577196-95577196 on Build GRCh38
rs774518408 Chr.13: 95577451-95577451 on Build GRCh38
rs902998167 Chr.13: 95577287-95577287 on Build GRCh38
rs771490748 Chr.13: 95577374-95577374 on Build GRCh38
rs920267147 Chr.13: 95577123-95577123 on Build GRCh38
rs1015824216 Chr.13: 95577149-95577149 on Build GRCh38
rs766469952 Chr.13: 95577273-95577273 on Build GRCh38
rs747414760 Chr.13: 95577342-95577342 on Build GRCh38
rs921307081 Chr.13: 95577040-95577040 on Build GRCh38
rs957372107 Chr.13: 95577352-95577352 on Build GRCh38
rs776820587 Chr.13: 95577385-95577385 on Build GRCh38
rs904856324 Chr.13: 95577450-95577450 on Build GRCh38
rs957337600 Chr.13: 95577015-95577015 on Build GRCh38
rs987365751 Chr.13: 95577050-95577050 on Build GRCh38
rs774826522 Chr.13: 95577184-95577184 on Build GRCh38
rs371833389 Chr.13: 95577264-95577264 on Build GRCh38
rs748998305 Chr.13: 95577316-95577316 on Build GRCh38
rs535236680 Chr.13: 95577159-95577159 on Build GRCh38
rs59299736 Chr.13: 95577134-95577134 on Build GRCh38
rs577293648 Chr.13: 95577213-95577213 on Build GRCh38
rs949038787 Chr.13: 95577422-95577422 on Build GRCh38
rs556508397 Chr.13: 95577070-95577070 on Build GRCh38
rs761240138 Chr.13: 95577205-95577205 on Build GRCh38
rs1032990717 Chr.13: 95577008-95577008 on Build GRCh38
rs779658942 Chr.13: 95577293-95577293 on Build GRCh38
rs771135066 Chr.13: 95577208-95577208 on Build GRCh38
rs768152902 Chr.13: 95577324-95577324 on Build GRCh38
rs771974638 Chr.13: 95577366-95577366 on Build GRCh38

View Full Product Details