Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99864740-99865249 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLYBL
Entrez Gene ID
171425
Gene Name
citrate lyase beta like
Gene Aliases
CLB
UniGene
Hs.655642
Gene Chromosome Location
Chr.13: 99606664-99909459 on Build GRCh38
Transcripts
NM_206808.3 NR_104592.1 XM_005254030.2 XM_005254031.2 XM_006719915.3 XM_011521051.2 XM_011521052.2 XM_017020402.1 XM_017020403.1 XM_017020404.1 NR_120421.1

Gene Symbol
LOC101927437
Entrez Gene ID
101927437
Gene Name
uncharacterized LOC101927437
Gene Aliases
-
UniGene
Hs.619936
Gene Chromosome Location
Chr.13: 99740870-99957165 on Build GRCh38
Transcripts
NM_206808.3 NR_104592.1 XM_005254030.2 XM_005254031.2 XM_006719915.3 XM_011521051.2 XM_011521052.2 XM_017020402.1 XM_017020403.1 XM_017020404.1 NR_120421.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1586809 c.452C>A p.S151Y Chr.13: 99864978-99864978 on Build GRCh38
6331732 c.441A>G p.V147V Chr.13: 99864967-99864967 on Build GRCh38
249270 c.608G>C p.G203A Chr.13: 99864885-99864885 on Build GRCh38
5948611 c.416_417delTG p.M145fs*16 Chr.13: 99864942-99864943 on Build GRCh38
944974 c.634+4T>G p.? Chr.13: 99864915-99864915 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765450751 Chr.13: 99865004-99865004 on Build GRCh38
rs745639540 Chr.13: 99865072-99865072 on Build GRCh38
rs776298489 Chr.13: 99864884-99864886 on Build GRCh38
rs746533473 Chr.13: 99864765-99864765 on Build GRCh38
rs748394532 Chr.13: 99865146-99865146 on Build GRCh38
rs747308343 Chr.13: 99864899-99864899 on Build GRCh38
rs763427582 Chr.13: 99864950-99864959 on Build GRCh38
rs1001428114 Chr.13: 99865198-99865198 on Build GRCh38
rs766437511 Chr.13: 99864820-99864821 on Build GRCh38
rs780856333 Chr.13: 99865050-99865050 on Build GRCh38
rs1043351812 Chr.13: 99864989-99864989 on Build GRCh38
rs34078213 Chr.13: 99864848-99864848 on Build GRCh38
rs933161798 Chr.13: 99864940-99864941 on Build GRCh38
rs781703716 Chr.13: 99864785-99864785 on Build GRCh38
rs760953060 Chr.13: 99865103-99865106 on Build GRCh38
rs7317032 Chr.13: 99865155-99865155 on Build GRCh38
rs761262046 Chr.13: 99864979-99864979 on Build GRCh38
rs201111226 Chr.13: 99864941-99864941 on Build GRCh38
rs746286628 Chr.13: 99864943-99864944 on Build GRCh38
rs781556496 Chr.13: 99865117-99865117 on Build GRCh38
rs769425435 Chr.13: 99864967-99864967 on Build GRCh38
rs748048221 Chr.13: 99864816-99864816 on Build GRCh38
rs780662456 Chr.13: 99864850-99864850 on Build GRCh38
rs923211956 Chr.13: 99865219-99865219 on Build GRCh38
rs757071703 Chr.13: 99864889-99864891 on Build GRCh38
rs1038680500 Chr.13: 99865023-99865023 on Build GRCh38
rs770776696 Chr.13: 99864945-99864945 on Build GRCh38
rs749506057 Chr.13: 99864799-99864799 on Build GRCh38
rs12430038 Chr.13: 99864774-99864774 on Build GRCh38
rs745837149 Chr.13: 99864923-99864924 on Build GRCh38
rs764770853 Chr.13: 99864778-99864778 on Build GRCh38
rs769358515 Chr.13: 99865093-99865093 on Build GRCh38
rs181692023 Chr.13: 99865005-99865005 on Build GRCh38
rs780375278 Chr.13: 99864796-99864796 on Build GRCh38
rs118114285 Chr.13: 99864780-99864780 on Build GRCh38
rs753020570 Chr.13: 99865009-99865009 on Build GRCh38
rs141341905 Chr.13: 99864877-99864877 on Build GRCh38
rs974849216 Chr.13: 99865202-99865202 on Build GRCh38
rs561836717 Chr.13: 99865211-99865212 on Build GRCh38
rs767877772 Chr.13: 99864972-99864972 on Build GRCh38
rs57999250 Chr.13: 99864956-99864957 on Build GRCh38
rs1016025010 Chr.13: 99865169-99865169 on Build GRCh38
rs776476663 Chr.13: 99864957-99864957 on Build GRCh38
rs950577766 Chr.13: 99864810-99864810 on Build GRCh38
rs774301953 Chr.13: 99864956-99864959 on Build GRCh38
rs779439271 Chr.13: 99864934-99864937 on Build GRCh38
rs778751698 Chr.13: 99864924-99864925 on Build GRCh38
rs766853539 Chr.13: 99864951-99864952 on Build GRCh38
rs763677082 Chr.13: 99864973-99864973 on Build GRCh38
rs768325050 Chr.13: 99864942-99864945 on Build GRCh38
rs563497370 Chr.13: 99864969-99864969 on Build GRCh38
rs578252753 Chr.13: 99864941-99864942 on Build GRCh38
rs200030197 Chr.13: 99864955-99864955 on Build GRCh38
rs942934227 Chr.13: 99865014-99865014 on Build GRCh38
rs796986882 Chr.13: 99864955-99864956 on Build GRCh38
rs762504808 Chr.13: 99864972-99864972 on Build GRCh38
rs57126726 Chr.13: 99864953-99864954 on Build GRCh38
rs140503030 Chr.13: 99864844-99864844 on Build GRCh38
rs899079141 Chr.13: 99865174-99865174 on Build GRCh38
rs151284228 Chr.13: 99864841-99864841 on Build GRCh38
rs755265961 Chr.13: 99864838-99864838 on Build GRCh38
rs3831038 Chr.13: 99864942-99864943 on Build GRCh38
rs545942459 Chr.13: 99865089-99865089 on Build GRCh38
rs751646196 Chr.13: 99864821-99864822 on Build GRCh38
rs768102404 Chr.13: 99864866-99864866 on Build GRCh38
rs747004830 Chr.13: 99865025-99865025 on Build GRCh38
rs759015217 Chr.13: 99864777-99864777 on Build GRCh38
rs772015279 Chr.13: 99864937-99864937 on Build GRCh38
rs147737718 Chr.13: 99864838-99864838 on Build GRCh38
rs754163807 Chr.13: 99864996-99864996 on Build GRCh38
rs750840828 Chr.13: 99864871-99864871 on Build GRCh38
rs779035752 Chr.13: 99864813-99864813 on Build GRCh38
rs377279428 Chr.13: 99864895-99864895 on Build GRCh38
rs879524978 Chr.13: 99865180-99865180 on Build GRCh38
rs1016217359 Chr.13: 99865170-99865170 on Build GRCh38
rs758510440 Chr.13: 99865015-99865015 on Build GRCh38
rs773621769 Chr.13: 99864948-99864959 on Build GRCh38
rs767783162 Chr.13: 99864881-99864882 on Build GRCh38
rs758339775 Chr.13: 99864924-99864927 on Build GRCh38
rs372957844 Chr.13: 99864773-99864773 on Build GRCh38
rs980105908 Chr.13: 99864870-99864870 on Build GRCh38
rs982049612 Chr.13: 99864781-99864781 on Build GRCh38
rs764714516 Chr.13: 99864847-99864847 on Build GRCh38
rs752074769 Chr.13: 99864779-99864779 on Build GRCh38
rs200974910 Chr.13: 99864959-99864959 on Build GRCh38
rs768659762 Chr.13: 99864811-99864811 on Build GRCh38
rs960129275 Chr.13: 99864878-99864878 on Build GRCh38
rs79423181 Chr.13: 99864786-99864786 on Build GRCh38
rs748218823 Chr.13: 99864936-99864936 on Build GRCh38
rs766664974 Chr.13: 99864983-99864983 on Build GRCh38
rs933086832 Chr.13: 99865104-99865104 on Build GRCh38
rs933258077 Chr.13: 99865222-99865222 on Build GRCh38
rs201825793 Chr.13: 99864943-99864943 on Build GRCh38
rs755185509 Chr.13: 99864998-99864998 on Build GRCh38
rs777733786 Chr.13: 99864938-99864938 on Build GRCh38
rs777823645 Chr.13: 99865024-99865024 on Build GRCh38
rs755441815 Chr.13: 99865083-99865083 on Build GRCh38
rs773896956 Chr.13: 99864975-99864975 on Build GRCh38
rs756436411 Chr.13: 99864793-99864793 on Build GRCh38
rs757163916 Chr.13: 99865033-99865033 on Build GRCh38
rs778861747 Chr.13: 99864916-99864916 on Build GRCh38
rs145666717 Chr.13: 99864845-99864845 on Build GRCh38

View Full Product Details