Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 99892745-99893246 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLYBL
Entrez Gene ID
171425
Gene Name
citrate lyase beta like
Gene Aliases
CLB
UniGene
Hs.655642
Gene Chromosome Location
Chr.13: 99606664-99909459 on Build GRCh38
Transcripts
NM_206808.3 NR_104592.1 XM_006719915.3 XM_011521051.2 XM_017020402.1 XM_017020404.1 NR_120421.1

Gene Symbol
LOC101927437
Entrez Gene ID
101927437
Gene Name
uncharacterized LOC101927437
Gene Aliases
-
UniGene
Hs.619936
Gene Chromosome Location
Chr.13: 99740870-99957165 on Build GRCh38
Transcripts
NM_206808.3 NR_104592.1 XM_006719915.3 XM_011521051.2 XM_017020402.1 XM_017020404.1 NR_120421.1

SNP Detail

SNP ID Chromosome Location
rs530623200 Chr.13: 99892815-99892815 on Build GRCh38
rs544395206 Chr.13: 99892907-99892907 on Build GRCh38
rs758529031 Chr.13: 99892840-99892840 on Build GRCh38
rs1003208906 Chr.13: 99893106-99893106 on Build GRCh38
rs1040435904 Chr.13: 99892786-99892786 on Build GRCh38
rs745435714 Chr.13: 99893083-99893083 on Build GRCh38
rs918823047 Chr.13: 99893157-99893157 on Build GRCh38
rs767528419 Chr.13: 99892788-99892788 on Build GRCh38
rs879684062 Chr.13: 99892970-99892970 on Build GRCh38
rs756028608 Chr.13: 99892852-99892852 on Build GRCh38
rs753541199 Chr.13: 99893024-99893024 on Build GRCh38
rs1036101408 Chr.13: 99892911-99892911 on Build GRCh38
rs920344482 Chr.13: 99892904-99892904 on Build GRCh38
rs941873188 Chr.13: 99893058-99893058 on Build GRCh38
rs1047532431 Chr.13: 99893044-99893044 on Build GRCh38
rs574491963 Chr.13: 99892988-99892988 on Build GRCh38
rs943076866 Chr.13: 99892784-99892784 on Build GRCh38
rs747529390 Chr.13: 99892845-99892846 on Build GRCh38
rs763779269 Chr.13: 99892910-99892910 on Build GRCh38
rs892335212 Chr.13: 99892960-99892960 on Build GRCh38
rs1057016478 Chr.13: 99893120-99893120 on Build GRCh38
rs143231237 Chr.13: 99893082-99893082 on Build GRCh38
rs529413527 Chr.13: 99893144-99893144 on Build GRCh38
rs905764124 Chr.13: 99892880-99892880 on Build GRCh38
rs750637728 Chr.13: 99892838-99892838 on Build GRCh38
rs929051425 Chr.13: 99892801-99892801 on Build GRCh38
rs985579521 Chr.13: 99892962-99892962 on Build GRCh38
rs913823048 Chr.13: 99893158-99893158 on Build GRCh38
rs972235767 Chr.13: 99893087-99893087 on Build GRCh38
rs992971311 Chr.13: 99892827-99892827 on Build GRCh38
rs971839567 Chr.13: 99892863-99892863 on Build GRCh38
rs922937661 Chr.13: 99892987-99892987 on Build GRCh38
rs976993011 Chr.13: 99892977-99892977 on Build GRCh38
rs528490742 Chr.13: 99893132-99893132 on Build GRCh38
rs558912168 Chr.13: 99893211-99893211 on Build GRCh38
rs2281757 Chr.13: 99892814-99892814 on Build GRCh38
rs561008150 Chr.13: 99892924-99892924 on Build GRCh38
rs375975329 Chr.13: 99892905-99892905 on Build GRCh38
rs2281755 Chr.13: 99893125-99893125 on Build GRCh38
rs574010507 Chr.13: 99893135-99893135 on Build GRCh38
rs2281756 Chr.13: 99893054-99893054 on Build GRCh38
rs143109201 Chr.13: 99893143-99893143 on Build GRCh38
rs138144536 Chr.13: 99892984-99892985 on Build GRCh38
rs530996229 Chr.13: 99893119-99893119 on Build GRCh38
rs908997412 Chr.13: 99893212-99893212 on Build GRCh38
rs780085408 Chr.13: 99892923-99892923 on Build GRCh38
rs1000171209 Chr.13: 99892992-99892992 on Build GRCh38
rs752100042 Chr.13: 99893019-99893019 on Build GRCh38
rs138147792 Chr.13: 99892890-99892890 on Build GRCh38
rs1021640701 Chr.13: 99892948-99892948 on Build GRCh38
rs1034780064 Chr.13: 99893011-99893011 on Build GRCh38
rs1014541937 Chr.13: 99893053-99893053 on Build GRCh38
rs544991199 Chr.13: 99892783-99892783 on Build GRCh38
rs966012928 Chr.13: 99892966-99892966 on Build GRCh38
rs190105127 Chr.13: 99892781-99892781 on Build GRCh38
rs961775387 Chr.13: 99892851-99892851 on Build GRCh38
rs537207374 Chr.13: 99893171-99893171 on Build GRCh38
rs148333055 Chr.13: 99893126-99893126 on Build GRCh38
rs149130281 Chr.13: 99892934-99892934 on Build GRCh38
rs968881478 Chr.13: 99892961-99892961 on Build GRCh38
rs965541734 Chr.13: 99893149-99893149 on Build GRCh38
rs950388584 Chr.13: 99893190-99893190 on Build GRCh38
rs569899389 Chr.13: 99893164-99893164 on Build GRCh38
rs911511103 Chr.13: 99892963-99892963 on Build GRCh38
rs1043962493 Chr.13: 99893209-99893209 on Build GRCh38
rs147203845 Chr.13: 99893152-99893152 on Build GRCh38
rs563570480 Chr.13: 99893070-99893070 on Build GRCh38
rs887623634 Chr.13: 99892824-99892824 on Build GRCh38
rs986214912 Chr.13: 99892918-99892918 on Build GRCh38
rs944650824 Chr.13: 99892933-99892933 on Build GRCh38

View Full Product Details