Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102799387-102799876 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIVM
Entrez Gene ID
54841
Gene Name
basic, immunoglobulin-like variable motif containing
Gene Aliases
-
UniGene
Hs.288809
Gene Chromosome Location
Chr.13: 102799049-102841538 on Build GRCh38
Transcripts
NM_001159596.1 NM_017693.3

SNP Detail

SNP ID Chromosome Location
rs372784385 Chr.13: 102799803-102799803 on Build GRCh38
rs967560480 Chr.13: 102799556-102799556 on Build GRCh38
rs1018697104 Chr.13: 102799504-102799504 on Build GRCh38
rs978908949 Chr.13: 102799573-102799573 on Build GRCh38
rs926112285 Chr.13: 102799794-102799794 on Build GRCh38
rs951364337 Chr.13: 102799791-102799791 on Build GRCh38
rs937016639 Chr.13: 102799411-102799411 on Build GRCh38
rs1007442124 Chr.13: 102799684-102799684 on Build GRCh38
rs577079972 Chr.13: 102799609-102799609 on Build GRCh38
rs995726936 Chr.13: 102799756-102799756 on Build GRCh38
rs1020815686 Chr.13: 102799757-102799757 on Build GRCh38
rs926224817 Chr.13: 102799575-102799575 on Build GRCh38
rs921377772 Chr.13: 102799567-102799567 on Build GRCh38
rs916908126 Chr.13: 102799611-102799611 on Build GRCh38
rs936806826 Chr.13: 102799568-102799568 on Build GRCh38
rs1026856404 Chr.13: 102799773-102799773 on Build GRCh38
rs982742196 Chr.13: 102799492-102799492 on Build GRCh38
rs1027301888 Chr.13: 102799489-102799489 on Build GRCh38
rs944051883 Chr.13: 102799498-102799498 on Build GRCh38
rs888911947 Chr.13: 102799671-102799671 on Build GRCh38
rs949717682 Chr.13: 102799620-102799620 on Build GRCh38
rs530292067 Chr.13: 102799718-102799718 on Build GRCh38
rs112067426 Chr.13: 102799712-102799712 on Build GRCh38
rs139347644 Chr.13: 102799626-102799626 on Build GRCh38
rs529772891 Chr.13: 102799792-102799792 on Build GRCh38
rs12872442 Chr.13: 102799493-102799493 on Build GRCh38
rs967444705 Chr.13: 102799759-102799759 on Build GRCh38
rs1048879642 Chr.13: 102799663-102799663 on Build GRCh38
rs1002556554 Chr.13: 102799634-102799634 on Build GRCh38
rs977256463 Chr.13: 102799534-102799534 on Build GRCh38
rs958759819 Chr.13: 102799467-102799467 on Build GRCh38
rs1006887658 Chr.13: 102799503-102799503 on Build GRCh38
rs1034069965 Chr.13: 102799699-102799699 on Build GRCh38
rs760349168 Chr.13: 102799738-102799738 on Build GRCh38
rs1053986449 Chr.13: 102799608-102799608 on Build GRCh38
rs906827850 Chr.13: 102799632-102799632 on Build GRCh38
rs1010304940 Chr.13: 102799408-102799408 on Build GRCh38
rs545737021 Chr.13: 102799677-102799677 on Build GRCh38
rs937573010 Chr.13: 102799597-102799597 on Build GRCh38
rs975432122 Chr.13: 102799527-102799527 on Build GRCh38
rs367554035 Chr.13: 102799748-102799748 on Build GRCh38
rs867880343 Chr.13: 102799623-102799623 on Build GRCh38
rs906943962 Chr.13: 102799415-102799415 on Build GRCh38
rs1034102505 Chr.13: 102799442-102799442 on Build GRCh38
rs965328611 Chr.13: 102799825-102799825 on Build GRCh38
rs1002589266 Chr.13: 102799429-102799429 on Build GRCh38
rs1019495416 Chr.13: 102799822-102799822 on Build GRCh38
rs113327700 Chr.13: 102799561-102799561 on Build GRCh38
rs554469510 Chr.13: 102799480-102799480 on Build GRCh38
rs543728558 Chr.13: 102799543-102799543 on Build GRCh38
rs913290574 Chr.13: 102799546-102799546 on Build GRCh38
rs542774885 Chr.13: 102799414-102799414 on Build GRCh38
rs965374994 Chr.13: 102799532-102799532 on Build GRCh38
rs945687739 Chr.13: 102799621-102799621 on Build GRCh38
rs951351971 Chr.13: 102799490-102799490 on Build GRCh38

View Full Product Details