Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 102821940-102822437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BIVM
Entrez Gene ID
54841
Gene Name
basic, immunoglobulin-like variable motif containing
Gene Aliases
-
UniGene
Hs.288809
Gene Chromosome Location
Chr.13: 102799049-102841538 on Build GRCh38
Transcripts
NM_001159596.1 NM_017693.3 NM_001204425.1

Gene Symbol
BIVM-ERCC5
Entrez Gene ID
100533467
Gene Name
BIVM-ERCC5 readthrough
Gene Aliases
ERCC5-202
UniGene
Hs.258429
Gene Chromosome Location
Chr.13: 102807146-102876001 on Build GRCh38
Transcripts
NM_001159596.1 NM_017693.3 NM_001204425.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7195971 c.820A>G p.I274V Chr.13: 102822078-102822078 on Build GRCh38
3466973 c.182C>T p.P61L Chr.13: 102822127-102822127 on Build GRCh38
5790994 c.161G>T p.C54F Chr.13: 102822106-102822106 on Build GRCh38
4455730 c.190A>G p.K64E Chr.13: 102822135-102822135 on Build GRCh38
233881 c.901+9A>G p.? Chr.13: 102822168-102822168 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374411150 Chr.13: 102822238-102822238 on Build GRCh38
rs776139273 Chr.13: 102822075-102822075 on Build GRCh38
rs2274390 Chr.13: 102822032-102822032 on Build GRCh38
rs756398306 Chr.13: 102822181-102822181 on Build GRCh38
rs886646047 Chr.13: 102822055-102822055 on Build GRCh38
rs768305384 Chr.13: 102822187-102822188 on Build GRCh38
rs779638534 Chr.13: 102822015-102822016 on Build GRCh38
rs375743756 Chr.13: 102822112-102822112 on Build GRCh38
rs947340654 Chr.13: 102822331-102822331 on Build GRCh38
rs745953522 Chr.13: 102822216-102822216 on Build GRCh38
rs771196463 Chr.13: 102822000-102822000 on Build GRCh38
rs748470517 Chr.13: 102822030-102822030 on Build GRCh38
rs942233552 Chr.13: 102821976-102821976 on Build GRCh38
rs754220700 Chr.13: 102822194-102822194 on Build GRCh38
rs2274389 Chr.13: 102822340-102822340 on Build GRCh38
rs777957929 Chr.13: 102822188-102822188 on Build GRCh38
rs779393030 Chr.13: 102822206-102822206 on Build GRCh38
rs768283172 Chr.13: 102822073-102822073 on Build GRCh38
rs1016453402 Chr.13: 102822090-102822090 on Build GRCh38
rs751712860 Chr.13: 102822019-102822019 on Build GRCh38
rs980850441 Chr.13: 102822369-102822369 on Build GRCh38
rs375901981 Chr.13: 102822026-102822026 on Build GRCh38
rs956776753 Chr.13: 102822239-102822239 on Build GRCh38
rs749763066 Chr.13: 102822045-102822045 on Build GRCh38
rs571820563 Chr.13: 102822227-102822227 on Build GRCh38
rs112997418 Chr.13: 102822226-102822226 on Build GRCh38
rs773174788 Chr.13: 102822098-102822098 on Build GRCh38
rs762287096 Chr.13: 102822204-102822205 on Build GRCh38
rs572715104 Chr.13: 102822393-102822393 on Build GRCh38
rs73570245 Chr.13: 102822172-102822172 on Build GRCh38
rs367994877 Chr.13: 102822050-102822050 on Build GRCh38
rs906126436 Chr.13: 102821974-102821974 on Build GRCh38
rs926771784 Chr.13: 102821966-102821966 on Build GRCh38
rs761299202 Chr.13: 102822248-102822248 on Build GRCh38
rs757700763 Chr.13: 102822201-102822201 on Build GRCh38
rs144626485 Chr.13: 102822157-102822157 on Build GRCh38
rs1044646497 Chr.13: 102821970-102821970 on Build GRCh38
rs374568565 Chr.13: 102822216-102822217 on Build GRCh38
rs766529406 Chr.13: 102822014-102822014 on Build GRCh38
rs746533294 Chr.13: 102822069-102822069 on Build GRCh38
rs142371210 Chr.13: 102822134-102822134 on Build GRCh38
rs773777490 Chr.13: 102822208-102822209 on Build GRCh38
rs761579996 Chr.13: 102822082-102822082 on Build GRCh38
rs373385824 Chr.13: 102822173-102822173 on Build GRCh38
rs746673201 Chr.13: 102822047-102822049 on Build GRCh38
rs771457493 Chr.13: 102822047-102822047 on Build GRCh38
rs770044669 Chr.13: 102822203-102822204 on Build GRCh38
rs890813732 Chr.13: 102822092-102822092 on Build GRCh38
rs778249093 Chr.13: 102822038-102822038 on Build GRCh38
rs914661762 Chr.13: 102822304-102822304 on Build GRCh38
rs764914763 Chr.13: 102822083-102822083 on Build GRCh38
rs1037877657 Chr.13: 102822009-102822009 on Build GRCh38
rs980106003 Chr.13: 102822335-102822335 on Build GRCh38
rs988893804 Chr.13: 102822285-102822285 on Build GRCh38
rs781275677 Chr.13: 102822029-102822029 on Build GRCh38
rs762938336 Chr.13: 102822105-102822105 on Build GRCh38
rs370072766 Chr.13: 102822043-102822043 on Build GRCh38
rs74540306 Chr.13: 102822271-102822271 on Build GRCh38
rs776116810 Chr.13: 102822202-102822203 on Build GRCh38

View Full Product Details