Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 107169899-107170436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM155A
Entrez Gene ID
728215
Gene Name
family with sequence similarity 155 member A
Gene Aliases
-
UniGene
Hs.535394
Gene Chromosome Location
Chr.13: 107168531-107867255 on Build GRCh38
Transcripts
NM_001080396.2 XM_011521109.2

SNP Detail

SNP ID Chromosome Location
rs945827257 Chr.13: 107170404-107170404 on Build GRCh38
rs191764949 Chr.13: 107170235-107170235 on Build GRCh38
rs898576246 Chr.13: 107170146-107170146 on Build GRCh38
rs964292076 Chr.13: 107170230-107170230 on Build GRCh38
rs1039408507 Chr.13: 107169951-107169951 on Build GRCh38
rs779644692 Chr.13: 107169974-107169975 on Build GRCh38
rs201972990 Chr.13: 107169958-107169958 on Build GRCh38
rs1019844262 Chr.13: 107170219-107170219 on Build GRCh38
rs186874338 Chr.13: 107170234-107170234 on Build GRCh38
rs990273508 Chr.13: 107170402-107170402 on Build GRCh38
rs781725544 Chr.13: 107170126-107170126 on Build GRCh38
rs556029130 Chr.13: 107170014-107170014 on Build GRCh38
rs182481658 Chr.13: 107170058-107170058 on Build GRCh38
rs201214792 Chr.13: 107169959-107169959 on Build GRCh38
rs575731001 Chr.13: 107170019-107170019 on Build GRCh38
rs1026216907 Chr.13: 107170186-107170186 on Build GRCh38
rs748696517 Chr.13: 107170129-107170129 on Build GRCh38
rs951608941 Chr.13: 107170211-107170211 on Build GRCh38
rs925434109 Chr.13: 107170216-107170216 on Build GRCh38
rs924975642 Chr.13: 107170275-107170275 on Build GRCh38
rs11302786 Chr.13: 107169959-107169959 on Build GRCh38
rs892321273 Chr.13: 107170065-107170065 on Build GRCh38
rs768259719 Chr.13: 107170365-107170365 on Build GRCh38
rs1006846481 Chr.13: 107170008-107170010 on Build GRCh38
rs1052261652 Chr.13: 107170025-107170025 on Build GRCh38
rs987390575 Chr.13: 107170215-107170215 on Build GRCh38
rs995587057 Chr.13: 107170160-107170160 on Build GRCh38
rs1051414581 Chr.13: 107169987-107169992 on Build GRCh38
rs898354167 Chr.13: 107170027-107170027 on Build GRCh38
rs561587221 Chr.13: 107170387-107170387 on Build GRCh38
rs114232871 Chr.13: 107170403-107170403 on Build GRCh38
rs757648377 Chr.13: 107170124-107170124 on Build GRCh38
rs371071027 Chr.13: 107170121-107170121 on Build GRCh38
rs951422412 Chr.13: 107170150-107170150 on Build GRCh38
rs987935607 Chr.13: 107170351-107170351 on Build GRCh38
rs1024969456 Chr.13: 107170169-107170169 on Build GRCh38
rs995973383 Chr.13: 107170051-107170051 on Build GRCh38
rs542768918 Chr.13: 107170007-107170007 on Build GRCh38
rs891047009 Chr.13: 107170007-107170008 on Build GRCh38
rs550345335 Chr.13: 107170405-107170405 on Build GRCh38
rs936404000 Chr.13: 107170339-107170339 on Build GRCh38
rs796259521 Chr.13: 107169965-107169965 on Build GRCh38
rs548964311 Chr.13: 107170208-107170211 on Build GRCh38
rs749878251 Chr.13: 107170216-107170219 on Build GRCh38
rs936413798 Chr.13: 107170021-107170021 on Build GRCh38
rs949313984 Chr.13: 107170085-107170085 on Build GRCh38
rs374632749 Chr.13: 107170206-107170206 on Build GRCh38
rs1046282206 Chr.13: 107170102-107170102 on Build GRCh38

View Full Product Details